Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180304673-180305215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
UniGene
Hs.696497
Gene Chromosome Location
Chr.5: 180300690-180353387 on Build GRCh38
Transcripts
NM_005110.3 XM_006714942.2 XM_017010100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356877 c.1801G>A p.A601T Chr.5: 180304813-180304813 on Build GRCh38
6012173 c.1773C>G p.I591M Chr.5: 180304841-180304841 on Build GRCh38
1066779 c.1834G>A p.A612T Chr.5: 180304780-180304780 on Build GRCh38
3614848 c.1824G>T p.Q608H Chr.5: 180304790-180304790 on Build GRCh38
3012267 c.1775T>C p.M592T Chr.5: 180304839-180304839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs988706463 Chr.5: 180304973-180304973 on Build GRCh38
rs989026203 Chr.5: 180304837-180304837 on Build GRCh38
rs372830153 Chr.5: 180304739-180304739 on Build GRCh38
rs766370990 Chr.5: 180304776-180304776 on Build GRCh38
rs567006958 Chr.5: 180304781-180304781 on Build GRCh38
rs777354105 Chr.5: 180304880-180304880 on Build GRCh38
rs572932988 Chr.5: 180305054-180305054 on Build GRCh38
rs913400507 Chr.5: 180305162-180305162 on Build GRCh38
rs918234279 Chr.5: 180305079-180305079 on Build GRCh38
rs746996315 Chr.5: 180305050-180305050 on Build GRCh38
rs938449792 Chr.5: 180305041-180305041 on Build GRCh38
rs969052154 Chr.5: 180305069-180305069 on Build GRCh38
rs370674913 Chr.5: 180304892-180304892 on Build GRCh38
rs143390983 Chr.5: 180304870-180304870 on Build GRCh38
rs374562769 Chr.5: 180304987-180304987 on Build GRCh38
rs966292053 Chr.5: 180305180-180305180 on Build GRCh38
rs544798462 Chr.5: 180305107-180305107 on Build GRCh38
rs558355089 Chr.5: 180305117-180305117 on Build GRCh38
rs867468090 Chr.5: 180304864-180304864 on Build GRCh38
rs369284217 Chr.5: 180304863-180304863 on Build GRCh38
rs766235324 Chr.5: 180304848-180304848 on Build GRCh38
rs757893767 Chr.5: 180304926-180304926 on Build GRCh38
rs968590 Chr.5: 180304749-180304749 on Build GRCh38
rs552798482 Chr.5: 180305028-180305028 on Build GRCh38
rs988972446 Chr.5: 180305145-180305145 on Build GRCh38
rs761525549 Chr.5: 180304741-180304741 on Build GRCh38
rs887609216 Chr.5: 180304891-180304891 on Build GRCh38
rs777948336 Chr.5: 180305190-180305190 on Build GRCh38
rs370090411 Chr.5: 180304814-180304814 on Build GRCh38
rs888610173 Chr.5: 180304853-180304853 on Build GRCh38
rs746950720 Chr.5: 180305084-180305084 on Build GRCh38
rs1039890081 Chr.5: 180305141-180305141 on Build GRCh38
rs768659348 Chr.5: 180304927-180304927 on Build GRCh38
rs552846107 Chr.5: 180304798-180304798 on Build GRCh38
rs755738463 Chr.5: 180304876-180304876 on Build GRCh38
rs754507901 Chr.5: 180304865-180304865 on Build GRCh38
rs759481705 Chr.5: 180304984-180304984 on Build GRCh38
rs956719523 Chr.5: 180304960-180304960 on Build GRCh38
rs532764819 Chr.5: 180304786-180304786 on Build GRCh38
rs377069145 Chr.5: 180304950-180304950 on Build GRCh38
rs780150267 Chr.5: 180304799-180304799 on Build GRCh38
rs927013612 Chr.5: 180305037-180305037 on Build GRCh38
rs965751879 Chr.5: 180304720-180304720 on Build GRCh38
rs538567339 Chr.5: 180304846-180304846 on Build GRCh38
rs994368151 Chr.5: 180304823-180304823 on Build GRCh38
rs558103943 Chr.5: 180304961-180304962 on Build GRCh38
rs776844983 Chr.5: 180304740-180304740 on Build GRCh38
rs992568470 Chr.5: 180305046-180305046 on Build GRCh38
rs376225831 Chr.5: 180304879-180304879 on Build GRCh38
rs780725432 Chr.5: 180304782-180304782 on Build GRCh38
rs777288088 Chr.5: 180304981-180304981 on Build GRCh38
rs979965082 Chr.5: 180304994-180304994 on Build GRCh38
rs773504223 Chr.5: 180304969-180304969 on Build GRCh38
rs969567349 Chr.5: 180304792-180304792 on Build GRCh38
rs976756260 Chr.5: 180304722-180304722 on Build GRCh38
rs758390303 Chr.5: 180304769-180304769 on Build GRCh38
rs1013289844 Chr.5: 180304765-180304765 on Build GRCh38
rs1014808438 Chr.5: 180304732-180304732 on Build GRCh38
rs879381741 Chr.5: 180305189-180305189 on Build GRCh38
rs189898875 Chr.5: 180304874-180304874 on Build GRCh38
rs729652 Chr.5: 180304695-180304695 on Build GRCh38
rs747062916 Chr.5: 180304821-180304821 on Build GRCh38
rs372679978 Chr.5: 180304840-180304840 on Build GRCh38
rs765173820 Chr.5: 180304843-180304843 on Build GRCh38
rs751065674 Chr.5: 180304777-180304777 on Build GRCh38
rs762857383 Chr.5: 180304975-180304975 on Build GRCh38
rs200888012 Chr.5: 180304871-180304871 on Build GRCh38
rs375659031 Chr.5: 180304847-180304847 on Build GRCh38
rs56403937 Chr.5: 180304715-180304715 on Build GRCh38
rs375131566 Chr.5: 180304901-180304901 on Build GRCh38
rs770173247 Chr.5: 180304942-180304942 on Build GRCh38
rs375998002 Chr.5: 180304757-180304757 on Build GRCh38
rs998257021 Chr.5: 180305166-180305166 on Build GRCh38
rs145839746 Chr.5: 180305126-180305126 on Build GRCh38
rs776775664 Chr.5: 180304838-180304838 on Build GRCh38
rs901228799 Chr.5: 180305157-180305157 on Build GRCh38
rs1049117024 Chr.5: 180304890-180304890 on Build GRCh38
rs755378099 Chr.5: 180304822-180304822 on Build GRCh38
rs1049114659 Chr.5: 180305085-180305085 on Build GRCh38
rs751459630 Chr.5: 180304859-180304859 on Build GRCh38
rs1021579676 Chr.5: 180304985-180304985 on Build GRCh38
rs752222791 Chr.5: 180304989-180304989 on Build GRCh38
rs199748661 Chr.5: 180304727-180304727 on Build GRCh38
rs748191781 Chr.5: 180304939-180304939 on Build GRCh38
rs539000015 Chr.5: 180304813-180304813 on Build GRCh38
rs758693715 Chr.5: 180304893-180304893 on Build GRCh38
rs1018400967 Chr.5: 180304717-180304717 on Build GRCh38
rs201814158 Chr.5: 180304743-180304743 on Build GRCh38

View Full Product Details