Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180312258-180312589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFPT2
Entrez Gene ID
9945
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Gene Aliases
GFAT, GFAT 2, GFAT2
UniGene
Hs.696497
Gene Chromosome Location
Chr.5: 180300690-180353387 on Build GRCh38
Transcripts
NM_005110.3 XM_006714942.2 XM_017010100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6103651 c.1518G>T p.E506D Chr.5: 180312458-180312458 on Build GRCh38
4599443 c.1490G>A p.R497Q Chr.5: 180312486-180312486 on Build GRCh38
449464 c.1442G>A p.S481N Chr.5: 180312534-180312534 on Build GRCh38
281358 c.1452C>A p.I484I Chr.5: 180312524-180312524 on Build GRCh38
3919833 c.1521C>T p.I507I Chr.5: 180312455-180312455 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750696966 Chr.5: 180312421-180312421 on Build GRCh38
rs762196067 Chr.5: 180312511-180312511 on Build GRCh38
rs188706919 Chr.5: 180312371-180312371 on Build GRCh38
rs1050771085 Chr.5: 180312427-180312427 on Build GRCh38
rs375030419 Chr.5: 180312514-180312514 on Build GRCh38
rs754832274 Chr.5: 180312541-180312541 on Build GRCh38
rs36046599 Chr.5: 180312451-180312451 on Build GRCh38
rs762953217 Chr.5: 180312532-180312532 on Build GRCh38
rs746813046 Chr.5: 180312441-180312441 on Build GRCh38
rs754711767 Chr.5: 180312447-180312447 on Build GRCh38
rs751766251 Chr.5: 180312538-180312538 on Build GRCh38
rs757413587 Chr.5: 180312398-180312398 on Build GRCh38
rs372123792 Chr.5: 180312379-180312379 on Build GRCh38
rs781251382 Chr.5: 180312450-180312450 on Build GRCh38
rs780815386 Chr.5: 180312548-180312548 on Build GRCh38
rs1026887976 Chr.5: 180312389-180312389 on Build GRCh38
rs761137565 Chr.5: 180312484-180312484 on Build GRCh38
rs200155071 Chr.5: 180312535-180312535 on Build GRCh38
rs758597326 Chr.5: 180312434-180312434 on Build GRCh38
rs761078165 Chr.5: 180312383-180312383 on Build GRCh38
rs941208391 Chr.5: 180312395-180312395 on Build GRCh38
rs780209779 Chr.5: 180312439-180312439 on Build GRCh38
rs750500082 Chr.5: 180312526-180312526 on Build GRCh38
rs962757729 Chr.5: 180312370-180312370 on Build GRCh38
rs777263657 Chr.5: 180312507-180312507 on Build GRCh38
rs770743216 Chr.5: 180312468-180312468 on Build GRCh38
rs765570371 Chr.5: 180312432-180312432 on Build GRCh38
rs764699733 Chr.5: 180312386-180312386 on Build GRCh38
rs982717621 Chr.5: 180312287-180312287 on Build GRCh38
rs769132046 Chr.5: 180312494-180312494 on Build GRCh38
rs1029335865 Chr.5: 180312329-180312329 on Build GRCh38
rs1006656658 Chr.5: 180312363-180312363 on Build GRCh38
rs541208940 Chr.5: 180312512-180312512 on Build GRCh38
rs754238207 Chr.5: 180312387-180312387 on Build GRCh38
rs368813952 Chr.5: 180312452-180312452 on Build GRCh38
rs1016722352 Chr.5: 180312368-180312368 on Build GRCh38
rs932280919 Chr.5: 180312425-180312425 on Build GRCh38
rs752725229 Chr.5: 180312549-180312549 on Build GRCh38
rs921033409 Chr.5: 180312406-180312406 on Build GRCh38
rs902462721 Chr.5: 180312292-180312292 on Build GRCh38
rs541909222 Chr.5: 180312312-180312312 on Build GRCh38
rs908371624 Chr.5: 180312361-180312361 on Build GRCh38
rs765408433 Chr.5: 180312515-180312515 on Build GRCh38
rs372866890 Chr.5: 180312467-180312467 on Build GRCh38
rs374787384 Chr.5: 180312404-180312404 on Build GRCh38
rs181277007 Chr.5: 180312557-180312557 on Build GRCh38
rs774084182 Chr.5: 180312474-180312474 on Build GRCh38

View Full Product Details