Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180601887-180602377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT4
Entrez Gene ID
2324
Gene Name
fms related tyrosine kinase 4
Gene Aliases
FLT-4, FLT41, LMPH1A, PCL, VEGFR-3, VEGFR3
UniGene
Hs.646917
Gene Chromosome Location
Chr.5: 180601506-180650298 on Build GRCh38
Transcripts
NM_182925.4 XM_011534478.2 XM_011534484.2 XM_017009263.1 XM_017009268.1

SNP Detail

SNP ID Chromosome Location
rs1031481292 Chr.5: 180602070-180602070 on Build GRCh38
rs895917593 Chr.5: 180602275-180602275 on Build GRCh38
rs890163740 Chr.5: 180602059-180602059 on Build GRCh38
rs1022884873 Chr.5: 180602160-180602160 on Build GRCh38
rs1049304878 Chr.5: 180602039-180602039 on Build GRCh38
rs115707970 Chr.5: 180602048-180602048 on Build GRCh38
rs919443418 Chr.5: 180601958-180601958 on Build GRCh38
rs911649280 Chr.5: 180602209-180602209 on Build GRCh38
rs113823996 Chr.5: 180602225-180602225 on Build GRCh38
rs927976776 Chr.5: 180602195-180602195 on Build GRCh38
rs781498936 Chr.5: 180602300-180602300 on Build GRCh38
rs955822620 Chr.5: 180602347-180602347 on Build GRCh38
rs545446086 Chr.5: 180602068-180602068 on Build GRCh38
rs978988139 Chr.5: 180602183-180602183 on Build GRCh38
rs980783526 Chr.5: 180602194-180602194 on Build GRCh38
rs868547112 Chr.5: 180602002-180602002 on Build GRCh38
rs138638674 Chr.5: 180602210-180602210 on Build GRCh38
rs987313097 Chr.5: 180602355-180602355 on Build GRCh38
rs915834303 Chr.5: 180601945-180601945 on Build GRCh38
rs996058228 Chr.5: 180602105-180602105 on Build GRCh38
rs959941332 Chr.5: 180601939-180601939 on Build GRCh38
rs766981627 Chr.5: 180602067-180602067 on Build GRCh38
rs77376470 Chr.5: 180602319-180602319 on Build GRCh38
rs991854616 Chr.5: 180601943-180601943 on Build GRCh38
rs1011459899 Chr.5: 180602147-180602147 on Build GRCh38
rs752904103 Chr.5: 180602115-180602115 on Build GRCh38
rs969996764 Chr.5: 180602186-180602186 on Build GRCh38
rs1052857627 Chr.5: 180602343-180602343 on Build GRCh38
rs543202646 Chr.5: 180602260-180602260 on Build GRCh38
rs969030416 Chr.5: 180602301-180602301 on Build GRCh38
rs759006343 Chr.5: 180601938-180601938 on Build GRCh38
rs1035652425 Chr.5: 180602148-180602148 on Build GRCh38
rs1036511584 Chr.5: 180601950-180601950 on Build GRCh38
rs757866838 Chr.5: 180602276-180602276 on Build GRCh38
rs924338399 Chr.5: 180602001-180602001 on Build GRCh38
rs559079273 Chr.5: 180602118-180602118 on Build GRCh38
rs577667926 Chr.5: 180602232-180602232 on Build GRCh38
rs887316887 Chr.5: 180602252-180602252 on Build GRCh38
rs931611765 Chr.5: 180602227-180602227 on Build GRCh38
rs900092629 Chr.5: 180602092-180602092 on Build GRCh38
rs991514868 Chr.5: 180602168-180602168 on Build GRCh38
rs1023281034 Chr.5: 180602179-180602179 on Build GRCh38
rs939458512 Chr.5: 180601949-180601949 on Build GRCh38
rs969378908 Chr.5: 180601916-180601916 on Build GRCh38
rs889406632 Chr.5: 180602046-180602046 on Build GRCh38
rs998582061 Chr.5: 180602324-180602324 on Build GRCh38
rs919391045 Chr.5: 180602226-180602226 on Build GRCh38
rs1007221731 Chr.5: 180602047-180602047 on Build GRCh38
rs924472124 Chr.5: 180602191-180602191 on Build GRCh38
rs930867256 Chr.5: 180602035-180602035 on Build GRCh38
rs887182247 Chr.5: 180602128-180602128 on Build GRCh38
rs981219038 Chr.5: 180601926-180601926 on Build GRCh38
rs752149641 Chr.5: 180602261-180602261 on Build GRCh38
rs563390137 Chr.5: 180602291-180602291 on Build GRCh38
rs755748604 Chr.5: 180602229-180602229 on Build GRCh38

View Full Product Details