Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181004325-181004829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL3
Entrez Gene ID
10917
Gene Name
butyrophilin like 3
Gene Aliases
BTN9.1, BTNLR
UniGene
Hs.225949
Gene Chromosome Location
Chr.5: 180988845-181006727 on Build GRCh38
Transcripts
NM_197975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1066893 c.846C>T p.D282D Chr.5: 181004746-181004746 on Build GRCh38
1185064 c.847G>T p.A283S Chr.5: 181004747-181004747 on Build GRCh38
1066894 c.851G>A p.R284Q Chr.5: 181004751-181004751 on Build GRCh38
4826978 c.836-1G>A p.? Chr.5: 181004735-181004735 on Build GRCh38
1066891 c.825C>T p.H275H Chr.5: 181004433-181004433 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780562662 Chr.5: 181004743-181004743 on Build GRCh38
rs931185990 Chr.5: 181004526-181004526 on Build GRCh38
rs868382641 Chr.5: 181004630-181004630 on Build GRCh38
rs919596560 Chr.5: 181004524-181004524 on Build GRCh38
rs768629666 Chr.5: 181004686-181004687 on Build GRCh38
rs759337826 Chr.5: 181004793-181004793 on Build GRCh38
rs201534771 Chr.5: 181004747-181004747 on Build GRCh38
rs777316895 Chr.5: 181004737-181004737 on Build GRCh38
rs747562004 Chr.5: 181004445-181004445 on Build GRCh38
rs202119732 Chr.5: 181004746-181004746 on Build GRCh38
rs928165088 Chr.5: 181004603-181004603 on Build GRCh38
rs542695748 Chr.5: 181004584-181004584 on Build GRCh38
rs991110130 Chr.5: 181004640-181004640 on Build GRCh38
rs573691992 Chr.5: 181004522-181004522 on Build GRCh38
rs763152483 Chr.5: 181004766-181004766 on Build GRCh38
rs772391437 Chr.5: 181004694-181004695 on Build GRCh38
rs368139118 Chr.5: 181004374-181004374 on Build GRCh38
rs985380294 Chr.5: 181004530-181004530 on Build GRCh38
rs187604443 Chr.5: 181004770-181004770 on Build GRCh38
rs774442663 Chr.5: 181004783-181004783 on Build GRCh38
rs375992218 Chr.5: 181004697-181004697 on Build GRCh38
rs746007917 Chr.5: 181004580-181004580 on Build GRCh38
rs536706389 Chr.5: 181004677-181004677 on Build GRCh38
rs545554746 Chr.5: 181004705-181004705 on Build GRCh38
rs746381247 Chr.5: 181004467-181004467 on Build GRCh38
rs556886756 Chr.5: 181004434-181004434 on Build GRCh38
rs2962301 Chr.5: 181004618-181004618 on Build GRCh38
rs1023164401 Chr.5: 181004624-181004624 on Build GRCh38
rs754839312 Chr.5: 181004719-181004719 on Build GRCh38
rs755663070 Chr.5: 181004446-181004446 on Build GRCh38
rs1032113574 Chr.5: 181004346-181004346 on Build GRCh38
rs372382578 Chr.5: 181004699-181004699 on Build GRCh38
rs939575559 Chr.5: 181004605-181004605 on Build GRCh38
rs970197016 Chr.5: 181004433-181004433 on Build GRCh38
rs983582168 Chr.5: 181004575-181004575 on Build GRCh38
rs373881919 Chr.5: 181004696-181004696 on Build GRCh38
rs561566073 Chr.5: 181004740-181004741 on Build GRCh38
rs143906320 Chr.5: 181004750-181004750 on Build GRCh38
rs911182833 Chr.5: 181004544-181004544 on Build GRCh38
rs201319847 Chr.5: 181004613-181004613 on Build GRCh38
rs1027778369 Chr.5: 181004496-181004496 on Build GRCh38
rs747405558 Chr.5: 181004688-181004688 on Build GRCh38
rs765357378 Chr.5: 181004701-181004701 on Build GRCh38
rs200452211 Chr.5: 181004712-181004712 on Build GRCh38
rs893365296 Chr.5: 181004362-181004362 on Build GRCh38
rs151034827 Chr.5: 181004720-181004720 on Build GRCh38
rs1017557264 Chr.5: 181004797-181004797 on Build GRCh38
rs371013216 Chr.5: 181004711-181004711 on Build GRCh38
rs772617373 Chr.5: 181004687-181004687 on Build GRCh38
rs755788330 Chr.5: 181004726-181004726 on Build GRCh38
rs893254603 Chr.5: 181004761-181004761 on Build GRCh38
rs537155581 Chr.5: 181004373-181004373 on Build GRCh38
rs1036285276 Chr.5: 181004682-181004682 on Build GRCh38
rs868805281 Chr.5: 181004442-181004443 on Build GRCh38
rs1041305569 Chr.5: 181004566-181004566 on Build GRCh38
rs371454208 Chr.5: 181004758-181004758 on Build GRCh38
rs573867040 Chr.5: 181004725-181004725 on Build GRCh38
rs72494582 Chr.5: 181004765-181004765 on Build GRCh38
rs901909762 Chr.5: 181004568-181004568 on Build GRCh38
rs748392659 Chr.5: 181004751-181004751 on Build GRCh38
rs754643161 Chr.5: 181004423-181004423 on Build GRCh38
rs943846544 Chr.5: 181004549-181004549 on Build GRCh38
rs758863164 Chr.5: 181004742-181004742 on Build GRCh38
rs961441307 Chr.5: 181004495-181004495 on Build GRCh38
rs779219892 Chr.5: 181004455-181004455 on Build GRCh38
rs1002976540 Chr.5: 181004633-181004633 on Build GRCh38
rs371953707 Chr.5: 181004683-181004683 on Build GRCh38
rs767337962 Chr.5: 181004799-181004799 on Build GRCh38
rs748792154 Chr.5: 181004740-181004740 on Build GRCh38
rs773226201 Chr.5: 181004704-181004704 on Build GRCh38

View Full Product Details