Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181054160-181054664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_001308245.1 NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_011534444.2 XM_017009113.1 XM_017009114.1 XM_017009115.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3366032 c.890A>C p.K297T Chr.5: 181054242-181054242 on Build GRCh38
3661766 c.887-2A>G p.? Chr.5: 181054237-181054237 on Build GRCh38
3661765 c.887-2A>G p.? Chr.5: 181054237-181054237 on Build GRCh38
3366031 c.890A>C p.K297T Chr.5: 181054242-181054242 on Build GRCh38
4141741 c.907+2T>C p.? Chr.5: 181054261-181054261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs954912010 Chr.5: 181054531-181054531 on Build GRCh38
rs896854809 Chr.5: 181054464-181054464 on Build GRCh38
rs958787188 Chr.5: 181054445-181054445 on Build GRCh38
rs536255801 Chr.5: 181054517-181054517 on Build GRCh38
rs762085586 Chr.5: 181054520-181054520 on Build GRCh38
rs747817656 Chr.5: 181054188-181054188 on Build GRCh38
rs935014467 Chr.5: 181054339-181054339 on Build GRCh38
rs919669520 Chr.5: 181054585-181054585 on Build GRCh38
rs956078290 Chr.5: 181054583-181054583 on Build GRCh38
rs748578897 Chr.5: 181054217-181054218 on Build GRCh38
rs772649074 Chr.5: 181054209-181054209 on Build GRCh38
rs528436767 Chr.5: 181054411-181054411 on Build GRCh38
rs986483068 Chr.5: 181054554-181054554 on Build GRCh38
rs778090392 Chr.5: 181054217-181054219 on Build GRCh38
rs550813857 Chr.5: 181054478-181054478 on Build GRCh38
rs769041214 Chr.5: 181054233-181054233 on Build GRCh38
rs927066118 Chr.5: 181054425-181054425 on Build GRCh38
rs565105670 Chr.5: 181054444-181054444 on Build GRCh38
rs776147638 Chr.5: 181054213-181054213 on Build GRCh38
rs887018934 Chr.5: 181054406-181054406 on Build GRCh38
rs964785660 Chr.5: 181054569-181054569 on Build GRCh38
rs370200512 Chr.5: 181054574-181054574 on Build GRCh38
rs777382119 Chr.5: 181054202-181054202 on Build GRCh38
rs997445788 Chr.5: 181054571-181054571 on Build GRCh38
rs765399807 Chr.5: 181054258-181054258 on Build GRCh38
rs1012918454 Chr.5: 181054449-181054449 on Build GRCh38
rs888269598 Chr.5: 181054483-181054483 on Build GRCh38
rs575447402 Chr.5: 181054640-181054640 on Build GRCh38
rs931169296 Chr.5: 181054587-181054587 on Build GRCh38
rs1006734500 Chr.5: 181054507-181054507 on Build GRCh38
rs368154215 Chr.5: 181054216-181054217 on Build GRCh38
rs187698366 Chr.5: 181054345-181054345 on Build GRCh38
rs914684644 Chr.5: 181054234-181054234 on Build GRCh38
rs924884749 Chr.5: 181054325-181054325 on Build GRCh38
rs898356429 Chr.5: 181054374-181054374 on Build GRCh38
rs763020751 Chr.5: 181054263-181054263 on Build GRCh38
rs546440354 Chr.5: 181054322-181054322 on Build GRCh38
rs78003831 Chr.5: 181054480-181054480 on Build GRCh38
rs1047446945 Chr.5: 181054403-181054403 on Build GRCh38
rs991638175 Chr.5: 181054203-181054203 on Build GRCh38
rs370394591 Chr.5: 181054233-181054233 on Build GRCh38
rs947611508 Chr.5: 181054383-181054383 on Build GRCh38
rs989013093 Chr.5: 181054591-181054591 on Build GRCh38
rs766633253 Chr.5: 181054205-181054209 on Build GRCh38
rs1039513324 Chr.5: 181054584-181054584 on Build GRCh38
rs559398577 Chr.5: 181054365-181054365 on Build GRCh38
rs183329496 Chr.5: 181054191-181054191 on Build GRCh38
rs543147739 Chr.5: 181054231-181054231 on Build GRCh38
rs1056829955 Chr.5: 181054456-181054456 on Build GRCh38
rs769350326 Chr.5: 181054479-181054479 on Build GRCh38
rs59863113 Chr.5: 181054629-181054629 on Build GRCh38
rs995349951 Chr.5: 181054387-181054387 on Build GRCh38
rs201834629 Chr.5: 181054260-181054260 on Build GRCh38
rs776898682 Chr.5: 181054235-181054235 on Build GRCh38
rs767315502 Chr.5: 181054311-181054311 on Build GRCh38
rs199923246 Chr.5: 181054298-181054298 on Build GRCh38
rs989044364 Chr.5: 181054359-181054359 on Build GRCh38
rs1044502490 Chr.5: 181054384-181054384 on Build GRCh38
rs956305474 Chr.5: 181054354-181054354 on Build GRCh38
rs548701756 Chr.5: 181054217-181054217 on Build GRCh38
rs368105479 Chr.5: 181054267-181054267 on Build GRCh38
rs747587013 Chr.5: 181054232-181054232 on Build GRCh38
rs551599642 Chr.5: 181054423-181054423 on Build GRCh38
rs568614431 Chr.5: 181054231-181054232 on Build GRCh38
rs965039587 Chr.5: 181054313-181054313 on Build GRCh38
rs912198200 Chr.5: 181054557-181054557 on Build GRCh38
rs564245856 Chr.5: 181054487-181054487 on Build GRCh38
rs1017723562 Chr.5: 181054264-181054264 on Build GRCh38
rs1021883426 Chr.5: 181054605-181054605 on Build GRCh38
rs942402731 Chr.5: 181054352-181054352 on Build GRCh38
rs748829862 Chr.5: 181054205-181054205 on Build GRCh38
rs912328047 Chr.5: 181054346-181054346 on Build GRCh38
rs372690028 Chr.5: 181054271-181054271 on Build GRCh38
rs754923625 Chr.5: 181054299-181054299 on Build GRCh38
rs77724206 Chr.5: 181054626-181054626 on Build GRCh38
rs762717058 Chr.5: 181054431-181054431 on Build GRCh38
rs547009709 Chr.5: 181054552-181054552 on Build GRCh38
rs762054473 Chr.5: 181054246-181054246 on Build GRCh38
rs531013625 Chr.5: 181054461-181054461 on Build GRCh38

View Full Product Details