Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181055354-181055859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL9
Entrez Gene ID
153579
Gene Name
butyrophilin like 9
Gene Aliases
BTN3, BTN8, VDLS1900
UniGene
Hs.546502
Gene Chromosome Location
Chr.5: 181039875-181061525 on Build GRCh38
Transcripts
NM_001308245.1 NM_152547.4 XM_006714823.3 XM_006714824.3 XM_006714825.3 XM_011534444.2 XM_017009113.1 XM_017009114.1 XM_017009115.1 XM_017009116.1 XM_017009117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4512797 c.916C>T p.Q306* Chr.5: 181055441-181055441 on Build GRCh38
4512798 c.916C>T p.Q306* Chr.5: 181055441-181055441 on Build GRCh38
7357872 c.912G>T p.K304N Chr.5: 181055437-181055437 on Build GRCh38
7357873 c.912G>T p.K304N Chr.5: 181055437-181055437 on Build GRCh38
5823169 c.908-1G>T p.? Chr.5: 181055432-181055432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759547242 Chr.5: 181055551-181055551 on Build GRCh38
rs1015189166 Chr.5: 181055670-181055670 on Build GRCh38
rs763850651 Chr.5: 181055648-181055648 on Build GRCh38
rs781551944 Chr.5: 181055809-181055809 on Build GRCh38
rs1053081260 Chr.5: 181055603-181055603 on Build GRCh38
rs1038931058 Chr.5: 181055835-181055835 on Build GRCh38
rs967868632 Chr.5: 181055700-181055700 on Build GRCh38
rs1039229690 Chr.5: 181055621-181055621 on Build GRCh38
rs373803332 Chr.5: 181055768-181055769 on Build GRCh38
rs752479920 Chr.5: 181055493-181055493 on Build GRCh38
rs1006135068 Chr.5: 181055630-181055630 on Build GRCh38
rs926033616 Chr.5: 181055566-181055566 on Build GRCh38
rs774457661 Chr.5: 181055474-181055474 on Build GRCh38
rs772047402 Chr.5: 181055826-181055828 on Build GRCh38
rs769074522 Chr.5: 181055453-181055453 on Build GRCh38
rs886710729 Chr.5: 181055827-181055827 on Build GRCh38
rs143224692 Chr.5: 181055437-181055437 on Build GRCh38
rs566700313 Chr.5: 181055541-181055541 on Build GRCh38
rs912000074 Chr.5: 181055611-181055611 on Build GRCh38
rs780999341 Chr.5: 181055833-181055833 on Build GRCh38
rs937397874 Chr.5: 181055739-181055739 on Build GRCh38
rs768608671 Chr.5: 181055430-181055430 on Build GRCh38
rs917253369 Chr.5: 181055775-181055775 on Build GRCh38
rs988011544 Chr.5: 181055686-181055686 on Build GRCh38
rs373399452 Chr.5: 181055491-181055491 on Build GRCh38
rs1056019855 Chr.5: 181055476-181055476 on Build GRCh38
rs537229578 Chr.5: 181055492-181055492 on Build GRCh38
rs1043575245 Chr.5: 181055462-181055462 on Build GRCh38
rs915704854 Chr.5: 181055538-181055538 on Build GRCh38
rs768383106 Chr.5: 181055415-181055418 on Build GRCh38
rs113008602 Chr.5: 181055597-181055597 on Build GRCh38
rs770978988 Chr.5: 181055473-181055473 on Build GRCh38
rs374470126 Chr.5: 181055619-181055619 on Build GRCh38
rs187663655 Chr.5: 181055685-181055685 on Build GRCh38
rs773416054 Chr.5: 181055467-181055467 on Build GRCh38
rs781571300 Chr.5: 181055454-181055454 on Build GRCh38
rs1003894294 Chr.5: 181055646-181055646 on Build GRCh38
rs145460487 Chr.5: 181055535-181055535 on Build GRCh38
rs950037861 Chr.5: 181055780-181055780 on Build GRCh38
rs561162170 Chr.5: 181055517-181055517 on Build GRCh38
rs922494214 Chr.5: 181055536-181055536 on Build GRCh38
rs1021300093 Chr.5: 181055693-181055693 on Build GRCh38
rs749922932 Chr.5: 181055808-181055808 on Build GRCh38
rs573376302 Chr.5: 181055756-181055756 on Build GRCh38
rs921989298 Chr.5: 181055785-181055785 on Build GRCh38
rs376363189 Chr.5: 181055505-181055505 on Build GRCh38
rs761241919 Chr.5: 181055418-181055418 on Build GRCh38
rs575423854 Chr.5: 181055755-181055755 on Build GRCh38
rs944684008 Chr.5: 181055765-181055765 on Build GRCh38
rs574247806 Chr.5: 181055596-181055596 on Build GRCh38
rs553128251 Chr.5: 181055672-181055672 on Build GRCh38
rs1039178319 Chr.5: 181055778-181055780 on Build GRCh38
rs148812190 Chr.5: 181055626-181055626 on Build GRCh38
rs532373097 Chr.5: 181055560-181055560 on Build GRCh38
rs926032903 Chr.5: 181055726-181055726 on Build GRCh38
rs773913520 Chr.5: 181055460-181055461 on Build GRCh38
rs558630227 Chr.5: 181055748-181055748 on Build GRCh38
rs553360913 Chr.5: 181055627-181055627 on Build GRCh38
rs76693682 Chr.5: 181055838-181055838 on Build GRCh38
rs979693707 Chr.5: 181055701-181055701 on Build GRCh38
rs887373091 Chr.5: 181055819-181055819 on Build GRCh38
rs748471488 Chr.5: 181055463-181055463 on Build GRCh38
rs748742004 Chr.5: 181055419-181055419 on Build GRCh38
rs11949170 Chr.5: 181055495-181055495 on Build GRCh38
rs11951478 Chr.5: 181055682-181055682 on Build GRCh38
rs11949224 Chr.5: 181055641-181055641 on Build GRCh38
rs11948509 Chr.5: 181055508-181055508 on Build GRCh38
rs748752482 Chr.5: 181055830-181055830 on Build GRCh38
rs568673917 Chr.5: 181055635-181055635 on Build GRCh38
rs759481497 Chr.5: 181055488-181055488 on Build GRCh38
rs777582106 Chr.5: 181055392-181055392 on Build GRCh38
rs372984555 Chr.5: 181055522-181055522 on Build GRCh38
rs951657817 Chr.5: 181055503-181055503 on Build GRCh38
rs937334575 Chr.5: 181055589-181055589 on Build GRCh38
rs78636845 Chr.5: 181055769-181055769 on Build GRCh38
rs913732082 Chr.5: 181055552-181055552 on Build GRCh38
rs1014497899 Chr.5: 181055612-181055612 on Build GRCh38
rs747517924 Chr.5: 181055440-181055440 on Build GRCh38
rs778525251 Chr.5: 181055801-181055801 on Build GRCh38
rs769988503 Chr.5: 181055464-181055464 on Build GRCh38
rs3885614 Chr.5: 181055795-181055795 on Build GRCh38
rs756824735 Chr.5: 181055426-181055426 on Build GRCh38
rs911863549 Chr.5: 181055763-181055763 on Build GRCh38
rs761500479 Chr.5: 181055487-181055487 on Build GRCh38
rs752470872 Chr.5: 181055388-181055388 on Build GRCh38
rs145688786 Chr.5: 181055390-181055390 on Build GRCh38
rs200210432 Chr.5: 181055377-181055377 on Build GRCh38
rs184688226 Chr.5: 181055469-181055469 on Build GRCh38
rs1043113901 Chr.5: 181055565-181055565 on Build GRCh38
rs996580487 Chr.5: 181055660-181055660 on Build GRCh38

View Full Product Details