Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 181241449-181241953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RACK1
Entrez Gene ID
10399
Gene Name
receptor for activated C kinase 1
Gene Aliases
GNB2L1, Gnb2-rs1, H12.3, HLC-7, PIG21
UniGene
Hs.5662
Gene Chromosome Location
Chr.5: 181236928-181243906 on Build GRCh38
Transcripts
NM_006098.4 NR_002592.1

Gene Symbol
SNORD96A
Entrez Gene ID
619571
Gene Name
small nucleolar RNA, C/D box 96A
Gene Aliases
U96A
UniGene
Gene Chromosome Location
Chr.5: 181241818-181241889 on Build GRCh38
Transcripts
NM_006098.4 NR_002592.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5128870 c.348C>T p.D116D Chr.5: 181241573-181241573 on Build GRCh38
5161554 c.283G>A p.G95S Chr.5: 181241638-181241638 on Build GRCh38
5204931 c.357G>T p.Q119H Chr.5: 181241564-181241564 on Build GRCh38
5749939 c.341C>A p.S114Y Chr.5: 181241580-181241580 on Build GRCh38
6346883 c.325C>A p.L109M Chr.5: 181241596-181241596 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878893567 Chr.5: 181241489-181241490 on Build GRCh38
rs773788138 Chr.5: 181241541-181241541 on Build GRCh38
rs11540163 Chr.5: 181241555-181241555 on Build GRCh38
rs112143249 Chr.5: 181241640-181241640 on Build GRCh38
rs753747711 Chr.5: 181241914-181241914 on Build GRCh38
rs751808838 Chr.5: 181241798-181241798 on Build GRCh38
rs560207411 Chr.5: 181241682-181241682 on Build GRCh38
rs781177385 Chr.5: 181241651-181241651 on Build GRCh38
rs764463539 Chr.5: 181241549-181241549 on Build GRCh38
rs745559475 Chr.5: 181241685-181241685 on Build GRCh38
rs773645224 Chr.5: 181241788-181241788 on Build GRCh38
rs41285571 Chr.5: 181241801-181241801 on Build GRCh38
rs762432305 Chr.5: 181241789-181241790 on Build GRCh38
rs773878127 Chr.5: 181241875-181241875 on Build GRCh38
rs745533226 Chr.5: 181241855-181241855 on Build GRCh38
rs375545127 Chr.5: 181241691-181241691 on Build GRCh38
rs769607577 Chr.5: 181241495-181241495 on Build GRCh38
rs368672600 Chr.5: 181241768-181241768 on Build GRCh38
rs140616174 Chr.5: 181241672-181241672 on Build GRCh38
rs752775600 Chr.5: 181241812-181241812 on Build GRCh38
rs542494303 Chr.5: 181241744-181241744 on Build GRCh38
rs1018580146 Chr.5: 181241700-181241700 on Build GRCh38
rs61751567 Chr.5: 181241646-181241646 on Build GRCh38
rs371389536 Chr.5: 181241793-181241793 on Build GRCh38
rs773945055 Chr.5: 181241885-181241885 on Build GRCh38
rs746506944 Chr.5: 181241869-181241869 on Build GRCh38
rs763442802 Chr.5: 181241789-181241789 on Build GRCh38
rs764210871 Chr.5: 181241913-181241913 on Build GRCh38
rs749037214 Chr.5: 181241832-181241832 on Build GRCh38
rs774644330 Chr.5: 181241891-181241891 on Build GRCh38
rs765474107 Chr.5: 181241680-181241681 on Build GRCh38
rs370922076 Chr.5: 181241642-181241642 on Build GRCh38
rs759081911 Chr.5: 181241543-181241543 on Build GRCh38
rs368015408 Chr.5: 181241577-181241577 on Build GRCh38
rs753099415 Chr.5: 181241779-181241780 on Build GRCh38
rs778722333 Chr.5: 181241845-181241845 on Build GRCh38
rs111709706 Chr.5: 181241489-181241489 on Build GRCh38
rs755106833 Chr.5: 181241650-181241650 on Build GRCh38
rs765631889 Chr.5: 181241623-181241623 on Build GRCh38
rs757254897 Chr.5: 181241918-181241918 on Build GRCh38
rs765854338 Chr.5: 181241783-181241783 on Build GRCh38
rs779561096 Chr.5: 181241865-181241865 on Build GRCh38
rs990038496 Chr.5: 181241759-181241759 on Build GRCh38
rs773028960 Chr.5: 181241497-181241497 on Build GRCh38
rs374020184 Chr.5: 181241516-181241516 on Build GRCh38
rs908878214 Chr.5: 181241816-181241816 on Build GRCh38
rs774932583 Chr.5: 181241776-181241776 on Build GRCh38
rs1002131846 Chr.5: 181241728-181241728 on Build GRCh38
rs189371625 Chr.5: 181241481-181241481 on Build GRCh38
rs771682374 Chr.5: 181241859-181241859 on Build GRCh38
rs777527568 Chr.5: 181241653-181241653 on Build GRCh38
rs976945328 Chr.5: 181241786-181241786 on Build GRCh38
rs778668973 Chr.5: 181241921-181241921 on Build GRCh38
rs775220784 Chr.5: 181241703-181241703 on Build GRCh38
rs752751584 Chr.5: 181241910-181241910 on Build GRCh38
rs757028195 Chr.5: 181241837-181241837 on Build GRCh38
rs878915108 Chr.5: 181241485-181241485 on Build GRCh38
rs762269467 Chr.5: 181241601-181241601 on Build GRCh38
rs770385704 Chr.5: 181241871-181241871 on Build GRCh38
rs112513276 Chr.5: 181241490-181241490 on Build GRCh38
rs901614974 Chr.5: 181241486-181241487 on Build GRCh38
rs201611053 Chr.5: 181241679-181241679 on Build GRCh38
rs531624117 Chr.5: 181241735-181241735 on Build GRCh38
rs757712341 Chr.5: 181241926-181241934 on Build GRCh38
rs750141415 Chr.5: 181241884-181241884 on Build GRCh38
rs762370438 Chr.5: 181241777-181241777 on Build GRCh38
rs150391948 Chr.5: 181241751-181241751 on Build GRCh38
rs897766165 Chr.5: 181241908-181241908 on Build GRCh38
rs965658462 Chr.5: 181241701-181241701 on Build GRCh38
rs562517926 Chr.5: 181241693-181241693 on Build GRCh38
rs11540180 Chr.5: 181241588-181241588 on Build GRCh38
rs750877608 Chr.5: 181241627-181241627 on Build GRCh38
rs372862645 Chr.5: 181241680-181241680 on Build GRCh38
rs769640440 Chr.5: 181241702-181241702 on Build GRCh38
rs759787665 Chr.5: 181241893-181241893 on Build GRCh38
rs748931084 Chr.5: 181241510-181241510 on Build GRCh38
rs758636714 Chr.5: 181241639-181241639 on Build GRCh38
rs1050099239 Chr.5: 181241600-181241600 on Build GRCh38
rs748086030 Chr.5: 181241652-181241652 on Build GRCh38
rs1007689709 Chr.5: 181241694-181241694 on Build GRCh38
rs796586350 Chr.5: 181241748-181241748 on Build GRCh38
rs763603695 Chr.5: 181241876-181241876 on Build GRCh38
rs545899546 Chr.5: 181241692-181241692 on Build GRCh38
rs75046714 Chr.5: 181241655-181241655 on Build GRCh38
rs767644958 Chr.5: 181241903-181241903 on Build GRCh38
rs781441209 Chr.5: 181241630-181241630 on Build GRCh38
rs760696981 Chr.5: 181241911-181241911 on Build GRCh38
rs11540186 Chr.5: 181241598-181241598 on Build GRCh38
rs756017493 Chr.5: 181241813-181241813 on Build GRCh38

View Full Product Details