Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 2117331-2117860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMDS
Entrez Gene ID
2762
Gene Name
GDP-mannose 4,6-dehydratase
Gene Aliases
GMD, SDR3E1
UniGene
Hs.144496
Gene Chromosome Location
Chr.6: 1623800-2245634 on Build GRCh38
Transcripts
NM_001253846.1 NM_001500.3 XM_006715066.2 XM_011514500.1 XM_011514502.2 XM_011514503.2 XM_011514505.1 XM_011514506.1 XM_011514507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281398 c.191G>A p.R64Q Chr.6: 2117513-2117513 on Build GRCh38
1076432 c.167G>A p.R56Q Chr.6: 2117537-2117537 on Build GRCh38
1442442 c.166C>T p.R56W Chr.6: 2117538-2117538 on Build GRCh38
7141854 c.191G>T p.R64L Chr.6: 2117513-2117513 on Build GRCh38
3374155 c.227T>C p.I76T Chr.6: 2117477-2117477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs556296220 Chr.6: 2117801-2117801 on Build GRCh38
rs960663057 Chr.6: 2117443-2117443 on Build GRCh38
rs759353627 Chr.6: 2117513-2117513 on Build GRCh38
rs1038726776 Chr.6: 2117749-2117749 on Build GRCh38
rs1048548905 Chr.6: 2117669-2117669 on Build GRCh38
rs771650335 Chr.6: 2117538-2117538 on Build GRCh38
rs769925393 Chr.6: 2117442-2117442 on Build GRCh38
rs970885392 Chr.6: 2117579-2117579 on Build GRCh38
rs779229993 Chr.6: 2117418-2117418 on Build GRCh38
rs763356264 Chr.6: 2117720-2117720 on Build GRCh38
rs933799023 Chr.6: 2117728-2117728 on Build GRCh38
rs573940538 Chr.6: 2117440-2117440 on Build GRCh38
rs539476018 Chr.6: 2117773-2117773 on Build GRCh38
rs184718234 Chr.6: 2117721-2117721 on Build GRCh38
rs751924615 Chr.6: 2117581-2117581 on Build GRCh38
rs375356391 Chr.6: 2117435-2117435 on Build GRCh38
rs764355609 Chr.6: 2117795-2117795 on Build GRCh38
rs887106778 Chr.6: 2117670-2117670 on Build GRCh38
rs751782325 Chr.6: 2117603-2117606 on Build GRCh38
rs576513863 Chr.6: 2117732-2117733 on Build GRCh38
rs773021803 Chr.6: 2117457-2117457 on Build GRCh38
rs780622404 Chr.6: 2117432-2117432 on Build GRCh38
rs369492511 Chr.6: 2117477-2117477 on Build GRCh38
rs34505029 Chr.6: 2117636-2117636 on Build GRCh38
rs1024476807 Chr.6: 2117571-2117571 on Build GRCh38
rs760036051 Chr.6: 2117537-2117537 on Build GRCh38
rs759015784 Chr.6: 2117570-2117570 on Build GRCh38
rs41300831 Chr.6: 2117596-2117596 on Build GRCh38
rs1039864253 Chr.6: 2117628-2117628 on Build GRCh38
rs559146261 Chr.6: 2117605-2117605 on Build GRCh38
rs752801425 Chr.6: 2117419-2117419 on Build GRCh38
rs775220343 Chr.6: 2117519-2117519 on Build GRCh38
rs767024066 Chr.6: 2117518-2117518 on Build GRCh38
rs1026974764 Chr.6: 2117411-2117411 on Build GRCh38
rs770686897 Chr.6: 2117498-2117498 on Build GRCh38
rs896778481 Chr.6: 2117762-2117762 on Build GRCh38
rs77538088 Chr.6: 2117741-2117741 on Build GRCh38
rs979173982 Chr.6: 2117501-2117501 on Build GRCh38
rs935027559 Chr.6: 2117546-2117546 on Build GRCh38
rs915235687 Chr.6: 2117637-2117637 on Build GRCh38
rs969099389 Chr.6: 2117486-2117486 on Build GRCh38
rs763725229 Chr.6: 2117547-2117547 on Build GRCh38
rs200059821 Chr.6: 2117425-2117425 on Build GRCh38
rs995572079 Chr.6: 2117657-2117657 on Build GRCh38
rs569139442 Chr.6: 2117396-2117396 on Build GRCh38
rs1013485988 Chr.6: 2117747-2117747 on Build GRCh38
rs192125145 Chr.6: 2117569-2117569 on Build GRCh38
rs758732804 Chr.6: 2117581-2117582 on Build GRCh38
rs747749404 Chr.6: 2117414-2117414 on Build GRCh38
rs763629342 Chr.6: 2117620-2117620 on Build GRCh38
rs545160741 Chr.6: 2117699-2117699 on Build GRCh38
rs776006541 Chr.6: 2117687-2117687 on Build GRCh38
rs201891715 Chr.6: 2117587-2117587 on Build GRCh38
rs967154729 Chr.6: 2117663-2117663 on Build GRCh38
rs941268680 Chr.6: 2117748-2117748 on Build GRCh38
rs781499907 Chr.6: 2117436-2117436 on Build GRCh38
rs368950978 Chr.6: 2117734-2117735 on Build GRCh38
rs558822379 Chr.6: 2117368-2117368 on Build GRCh38
rs767026759 Chr.6: 2117572-2117572 on Build GRCh38
rs879119553 Chr.6: 2117367-2117367 on Build GRCh38
rs774169486 Chr.6: 2117504-2117504 on Build GRCh38
rs900028226 Chr.6: 2117654-2117654 on Build GRCh38
rs748474322 Chr.6: 2117591-2117591 on Build GRCh38
rs538141051 Chr.6: 2117419-2117419 on Build GRCh38
rs200938074 Chr.6: 2117520-2117520 on Build GRCh38
rs772822725 Chr.6: 2117589-2117589 on Build GRCh38
rs959591824 Chr.6: 2117615-2117615 on Build GRCh38

View Full Product Details