Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 2623840-2624352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC01600
Entrez Gene ID
154386
Gene Name
long intergenic non-protein coding RNA 1600
Gene Aliases
C6orf195, bA145H9.2
UniGene
Hs.680574
Gene Chromosome Location
Chr.6: 2621913-2634603 on Build GRCh38
Transcripts
NR_131168.1

SNP Detail

SNP ID Chromosome Location
rs1006500409 Chr.6: 2624173-2624173 on Build GRCh38
rs376168979 Chr.6: 2624179-2624179 on Build GRCh38
rs972585874 Chr.6: 2624051-2624051 on Build GRCh38
rs73348510 Chr.6: 2624048-2624048 on Build GRCh38
rs567053487 Chr.6: 2624245-2624245 on Build GRCh38
rs986225892 Chr.6: 2624022-2624022 on Build GRCh38
rs571162596 Chr.6: 2624202-2624202 on Build GRCh38
rs937786322 Chr.6: 2624071-2624071 on Build GRCh38
rs889476335 Chr.6: 2624169-2624169 on Build GRCh38
rs552789555 Chr.6: 2624280-2624280 on Build GRCh38
rs768171762 Chr.6: 2623997-2623997 on Build GRCh38
rs564372691 Chr.6: 2624061-2624061 on Build GRCh38
rs562184557 Chr.6: 2624056-2624056 on Build GRCh38
rs1042519881 Chr.6: 2624146-2624146 on Build GRCh38
rs1046432085 Chr.6: 2623878-2623878 on Build GRCh38
rs765445772 Chr.6: 2624187-2624187 on Build GRCh38
rs62391260 Chr.6: 2624287-2624287 on Build GRCh38
rs937772760 Chr.6: 2623925-2623925 on Build GRCh38
rs747923747 Chr.6: 2624219-2624219 on Build GRCh38
rs1041957091 Chr.6: 2624092-2624092 on Build GRCh38
rs151236705 Chr.6: 2623957-2623957 on Build GRCh38
rs543563825 Chr.6: 2624075-2624075 on Build GRCh38
rs867246634 Chr.6: 2624195-2624195 on Build GRCh38
rs183083234 Chr.6: 2624165-2624165 on Build GRCh38
rs567290771 Chr.6: 2623946-2623946 on Build GRCh38
rs533313207 Chr.6: 2624074-2624074 on Build GRCh38
rs78623302 Chr.6: 2624291-2624291 on Build GRCh38
rs904903960 Chr.6: 2623912-2623912 on Build GRCh38
rs188075796 Chr.6: 2624237-2624237 on Build GRCh38
rs761205442 Chr.6: 2624031-2624031 on Build GRCh38
rs946786742 Chr.6: 2624286-2624286 on Build GRCh38
rs971300682 Chr.6: 2624057-2624057 on Build GRCh38
rs758644926 Chr.6: 2624267-2624267 on Build GRCh38
rs571217478 Chr.6: 2623929-2623929 on Build GRCh38
rs938044080 Chr.6: 2623904-2623904 on Build GRCh38
rs1055319283 Chr.6: 2623945-2623945 on Build GRCh38
rs918539150 Chr.6: 2624054-2624054 on Build GRCh38
rs556975699 Chr.6: 2623980-2623980 on Build GRCh38
rs150441521 Chr.6: 2624325-2624325 on Build GRCh38
rs200907994 Chr.6: 2624053-2624053 on Build GRCh38
rs915320753 Chr.6: 2624130-2624130 on Build GRCh38
rs990816029 Chr.6: 2624248-2624248 on Build GRCh38
rs573518564 Chr.6: 2623982-2623982 on Build GRCh38
rs563743627 Chr.6: 2624076-2624076 on Build GRCh38
rs993463534 Chr.6: 2624175-2624175 on Build GRCh38
rs552001988 Chr.6: 2624178-2624178 on Build GRCh38
rs998362443 Chr.6: 2624148-2624148 on Build GRCh38
rs535930458 Chr.6: 2624251-2624251 on Build GRCh38
rs1055243396 Chr.6: 2624121-2624121 on Build GRCh38
rs757820673 Chr.6: 2623869-2623869 on Build GRCh38
rs953629231 Chr.6: 2624001-2624001 on Build GRCh38
rs945031154 Chr.6: 2624081-2624081 on Build GRCh38
rs1043065724 Chr.6: 2623979-2623979 on Build GRCh38
rs542387613 Chr.6: 2624000-2624000 on Build GRCh38
rs9378720 Chr.6: 2624140-2624140 on Build GRCh38
rs1053897136 Chr.6: 2623938-2623938 on Build GRCh38
rs17210205 Chr.6: 2624223-2624223 on Build GRCh38
rs914959707 Chr.6: 2624238-2624238 on Build GRCh38
rs573974612 Chr.6: 2624292-2624292 on Build GRCh38
rs552730751 Chr.6: 2624023-2624023 on Build GRCh38
rs1025442385 Chr.6: 2624210-2624210 on Build GRCh38
rs1150656 Chr.6: 2624028-2624028 on Build GRCh38
rs889278882 Chr.6: 2624142-2624142 on Build GRCh38
rs12213592 Chr.6: 2624147-2624147 on Build GRCh38
rs1012263718 Chr.6: 2623959-2623959 on Build GRCh38
rs970665947 Chr.6: 2624186-2624186 on Build GRCh38
rs997723519 Chr.6: 2624137-2624137 on Build GRCh38
rs571847498 Chr.6: 2624247-2624247 on Build GRCh38
rs949176329 Chr.6: 2623876-2623876 on Build GRCh38
rs953807357 Chr.6: 2624012-2624012 on Build GRCh38
rs926471663 Chr.6: 2624209-2624211 on Build GRCh38
rs559977245 Chr.6: 2624164-2624164 on Build GRCh38
rs906618275 Chr.6: 2623896-2623896 on Build GRCh38

View Full Product Details