Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 3137996-3138518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPHL
Entrez Gene ID
670
Gene Name
biphenyl hydrolase-like (serine hydrolase)
Gene Aliases
BPH-RP, MCNAA, VACVASE
UniGene
Hs.10136
Gene Chromosome Location
Chr.6: 3118376-3153198 on Build GRCh38
Transcripts
NM_001302777.1 NM_004332.3 NR_026648.2 NR_026649.2

SNP Detail

SNP ID Chromosome Location
rs57842152 Chr.6: 3138192-3138192 on Build GRCh38
rs774348298 Chr.6: 3138032-3138032 on Build GRCh38
rs957800875 Chr.6: 3138173-3138173 on Build GRCh38
rs745846572 Chr.6: 3138028-3138029 on Build GRCh38
rs1050415473 Chr.6: 3138229-3138229 on Build GRCh38
rs984448666 Chr.6: 3138290-3138290 on Build GRCh38
rs753814644 Chr.6: 3138357-3138357 on Build GRCh38
rs889792642 Chr.6: 3138435-3138435 on Build GRCh38
rs1013800460 Chr.6: 3138139-3138139 on Build GRCh38
rs748665063 Chr.6: 3138212-3138212 on Build GRCh38
rs557904731 Chr.6: 3138020-3138020 on Build GRCh38
rs565760253 Chr.6: 3138322-3138322 on Build GRCh38
rs1005998063 Chr.6: 3138066-3138066 on Build GRCh38
rs914709096 Chr.6: 3138434-3138434 on Build GRCh38
rs140335945 Chr.6: 3138488-3138488 on Build GRCh38
rs761910412 Chr.6: 3138490-3138490 on Build GRCh38
rs759078724 Chr.6: 3138236-3138236 on Build GRCh38
rs558425543 Chr.6: 3138083-3138083 on Build GRCh38
rs540789276 Chr.6: 3138272-3138272 on Build GRCh38
rs185802602 Chr.6: 3138280-3138280 on Build GRCh38
rs550247137 Chr.6: 3138209-3138209 on Build GRCh38
rs923504866 Chr.6: 3138487-3138487 on Build GRCh38
rs765203407 Chr.6: 3138491-3138491 on Build GRCh38
rs543095531 Chr.6: 3138061-3138061 on Build GRCh38
rs933331374 Chr.6: 3138226-3138226 on Build GRCh38
rs1054886971 Chr.6: 3138303-3138303 on Build GRCh38
rs967970295 Chr.6: 3138443-3138443 on Build GRCh38
rs7765391 Chr.6: 3138265-3138265 on Build GRCh38
rs566266698 Chr.6: 3138121-3138121 on Build GRCh38
rs897660146 Chr.6: 3138260-3138260 on Build GRCh38
rs754948152 Chr.6: 3138125-3138125 on Build GRCh38
rs1006302300 Chr.6: 3138243-3138243 on Build GRCh38
rs977674259 Chr.6: 3138480-3138480 on Build GRCh38
rs577846561 Chr.6: 3138024-3138024 on Build GRCh38
rs146736068 Chr.6: 3138225-3138225 on Build GRCh38
rs993389138 Chr.6: 3138264-3138264 on Build GRCh38
rs775028425 Chr.6: 3138271-3138271 on Build GRCh38
rs1016074857 Chr.6: 3138075-3138075 on Build GRCh38
rs558062369 Chr.6: 3138388-3138388 on Build GRCh38
rs1029053033 Chr.6: 3138037-3138037 on Build GRCh38
rs896319177 Chr.6: 3138122-3138122 on Build GRCh38
rs144270175 Chr.6: 3138220-3138220 on Build GRCh38
rs980491607 Chr.6: 3138127-3138127 on Build GRCh38
rs926370850 Chr.6: 3138144-3138144 on Build GRCh38
rs559095694 Chr.6: 3138131-3138131 on Build GRCh38
rs571496116 Chr.6: 3138444-3138444 on Build GRCh38
rs767652856 Chr.6: 3138163-3138163 on Build GRCh38
rs914779978 Chr.6: 3138208-3138208 on Build GRCh38
rs1037488772 Chr.6: 3138254-3138254 on Build GRCh38
rs989238271 Chr.6: 3138373-3138373 on Build GRCh38
rs549327537 Chr.6: 3138296-3138296 on Build GRCh38
rs1024608931 Chr.6: 3138094-3138094 on Build GRCh38
rs557057738 Chr.6: 3138080-3138080 on Build GRCh38
rs917626170 Chr.6: 3138319-3138319 on Build GRCh38
rs528359944 Chr.6: 3138184-3138184 on Build GRCh38
rs375158141 Chr.6: 3138261-3138261 on Build GRCh38
rs772104232 Chr.6: 3138195-3138196 on Build GRCh38
rs11307799 Chr.6: 3138185-3138185 on Build GRCh38
rs763994832 Chr.6: 3138298-3138298 on Build GRCh38
rs573550165 Chr.6: 3138089-3138089 on Build GRCh38
rs869080182 Chr.6: 3138191-3138192 on Build GRCh38
rs781125136 Chr.6: 3138029-3138029 on Build GRCh38
rs769290375 Chr.6: 3138054-3138054 on Build GRCh38
rs1019708663 Chr.6: 3138462-3138462 on Build GRCh38
rs888920223 Chr.6: 3138062-3138062 on Build GRCh38
rs970801321 Chr.6: 3138095-3138095 on Build GRCh38
rs190279592 Chr.6: 3138387-3138387 on Build GRCh38
rs1032326597 Chr.6: 3138206-3138206 on Build GRCh38
rs1053822741 Chr.6: 3138434-3138436 on Build GRCh38
rs533603048 Chr.6: 3138362-3138362 on Build GRCh38
rs1042590837 Chr.6: 3138355-3138355 on Build GRCh38

View Full Product Details