Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4077263-4077759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM217A
Entrez Gene ID
222826
Gene Name
family with sequence similarity 217 member A
Gene Aliases
C6orf146
UniGene
Hs.657342
Gene Chromosome Location
Chr.6: 4068352-4087344 on Build GRCh38
Transcripts
NM_173563.2 XM_005248951.3 XM_006715025.2 XM_006715026.3 XM_011514413.2 XM_011514414.2 XM_011514415.1 XM_011514416.2 XM_011514417.1 XM_017010480.1 XM_017010481.1 XM_017010482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5893114 c.60+1G>T p.? Chr.6: 4077354-4077354 on Build GRCh38
266146 c.10A>C p.R4R Chr.6: 4077405-4077405 on Build GRCh38
4006500 c.16G>A p.E6K Chr.6: 4077399-4077399 on Build GRCh38
4625791 c.60G>T p.E20D Chr.6: 4077355-4077355 on Build GRCh38
6659500 c.35T>C p.L12P Chr.6: 4077380-4077380 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747316875 Chr.6: 4077445-4077445 on Build GRCh38
rs759394565 Chr.6: 4077481-4077481 on Build GRCh38
rs755681312 Chr.6: 4077409-4077409 on Build GRCh38
rs777057045 Chr.6: 4077458-4077458 on Build GRCh38
rs746327680 Chr.6: 4077699-4077699 on Build GRCh38
rs905287967 Chr.6: 4077538-4077538 on Build GRCh38
rs751328916 Chr.6: 4077379-4077379 on Build GRCh38
rs36205969 Chr.6: 4077670-4077674 on Build GRCh38
rs780703755 Chr.6: 4077317-4077317 on Build GRCh38
rs1003736135 Chr.6: 4077700-4077700 on Build GRCh38
rs761914380 Chr.6: 4077352-4077352 on Build GRCh38
rs77685397 Chr.6: 4077634-4077634 on Build GRCh38
rs772596192 Chr.6: 4077430-4077430 on Build GRCh38
rs778040222 Chr.6: 4077539-4077539 on Build GRCh38
rs532873370 Chr.6: 4077462-4077462 on Build GRCh38
rs762829543 Chr.6: 4077469-4077469 on Build GRCh38
rs192079514 Chr.6: 4077326-4077326 on Build GRCh38
rs149184193 Chr.6: 4077411-4077411 on Build GRCh38
rs868524901 Chr.6: 4077505-4077505 on Build GRCh38
rs899424184 Chr.6: 4077452-4077452 on Build GRCh38
rs751219162 Chr.6: 4077311-4077311 on Build GRCh38
rs376681667 Chr.6: 4077585-4077585 on Build GRCh38
rs867835254 Chr.6: 4077461-4077461 on Build GRCh38
rs747600101 Chr.6: 4077322-4077322 on Build GRCh38
rs970985811 Chr.6: 4077659-4077659 on Build GRCh38
rs765400882 Chr.6: 4077369-4077369 on Build GRCh38
rs763547836 Chr.6: 4077497-4077497 on Build GRCh38
rs568714245 Chr.6: 4077691-4077691 on Build GRCh38
rs554000972 Chr.6: 4077309-4077309 on Build GRCh38
rs772373646 Chr.6: 4077329-4077329 on Build GRCh38
rs140476963 Chr.6: 4077337-4077338 on Build GRCh38
rs773310925 Chr.6: 4077468-4077468 on Build GRCh38
rs370029329 Chr.6: 4077675-4077675 on Build GRCh38
rs755734531 Chr.6: 4077494-4077494 on Build GRCh38
rs138399485 Chr.6: 4077385-4077385 on Build GRCh38
rs1007638057 Chr.6: 4077298-4077298 on Build GRCh38
rs970915165 Chr.6: 4077343-4077343 on Build GRCh38
rs1020200601 Chr.6: 4077291-4077291 on Build GRCh38
rs372907523 Chr.6: 4077677-4077677 on Build GRCh38
rs549419810 Chr.6: 4077484-4077484 on Build GRCh38
rs369629402 Chr.6: 4077589-4077589 on Build GRCh38
rs34188358 Chr.6: 4077515-4077516 on Build GRCh38
rs769970458 Chr.6: 4077434-4077434 on Build GRCh38
rs541801556 Chr.6: 4077377-4077377 on Build GRCh38
rs746375582 Chr.6: 4077327-4077327 on Build GRCh38
rs200457073 Chr.6: 4077324-4077324 on Build GRCh38
rs751317174 Chr.6: 4077478-4077478 on Build GRCh38
rs116381273 Chr.6: 4077498-4077498 on Build GRCh38
rs76866780 Chr.6: 4077331-4077331 on Build GRCh38
rs1007540202 Chr.6: 4077566-4077566 on Build GRCh38
rs112866331 Chr.6: 4077380-4077380 on Build GRCh38
rs201928633 Chr.6: 4077674-4077678 on Build GRCh38
rs752398474 Chr.6: 4077489-4077489 on Build GRCh38
rs574779482 Chr.6: 4077294-4077294 on Build GRCh38
rs768946253 Chr.6: 4077334-4077334 on Build GRCh38
rs76370173 Chr.6: 4077603-4077603 on Build GRCh38
rs754593246 Chr.6: 4077316-4077316 on Build GRCh38
rs767288131 Chr.6: 4077485-4077485 on Build GRCh38
rs947249591 Chr.6: 4077447-4077447 on Build GRCh38
rs538039515 Chr.6: 4077587-4077587 on Build GRCh38
rs666254 Chr.6: 4077301-4077301 on Build GRCh38
rs200025490 Chr.6: 4077359-4077359 on Build GRCh38
rs766283172 Chr.6: 4077473-4077473 on Build GRCh38
rs769001419 Chr.6: 4077449-4077449 on Build GRCh38
rs774602053 Chr.6: 4077671-4077671 on Build GRCh38
rs766126896 Chr.6: 4077378-4077378 on Build GRCh38
rs189094048 Chr.6: 4077431-4077431 on Build GRCh38
rs775897278 Chr.6: 4077330-4077330 on Build GRCh38
rs776860575 Chr.6: 4077344-4077344 on Build GRCh38
rs892487639 Chr.6: 4077600-4077600 on Build GRCh38
rs749521058 Chr.6: 4077609-4077609 on Build GRCh38
rs79815172 Chr.6: 4077604-4077604 on Build GRCh38
rs746406836 Chr.6: 4077414-4077414 on Build GRCh38
rs968614912 Chr.6: 4077663-4077663 on Build GRCh38

View Full Product Details