Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4130680-4131182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C6orf201
Entrez Gene ID
404220
Gene Name
chromosome 6 open reading frame 201
Gene Aliases
dJ1013A10.5
UniGene
Hs.716731
Gene Chromosome Location
Chr.6: 4079197-4130788 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Gene Symbol
ECI2
Entrez Gene ID
10455
Gene Name
enoyl-CoA delta isomerase 2
Gene Aliases
ACBD2, DRS-1, DRS1, HCA88, PECI, dJ1013A10.3
UniGene
Hs.15250
Gene Chromosome Location
Chr.6: 4115693-4135597 on Build GRCh38
Transcripts
NM_001085401.2 NR_104463.1 NR_104464.1 XM_017010861.1 NM_001166010.1 NM_006117.2 NM_206836.2 NR_028588.1 XM_006714957.1 XM_017010170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123835 c.256G>C p.D86H Chr.6: 4130823-4130823 on Build GRCh38
1312195 c.267C>T p.N89N Chr.6: 4130812-4130812 on Build GRCh38
4619865 c.155C>T p.P52L Chr.6: 4130834-4130834 on Build GRCh38
6847929 c.221A>G p.K74R Chr.6: 4130768-4130768 on Build GRCh38
4619864 c.245C>T p.P82L Chr.6: 4130834-4130834 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141043087 Chr.6: 4130856-4130856 on Build GRCh38
rs370215289 Chr.6: 4130889-4130889 on Build GRCh38
rs185741061 Chr.6: 4131151-4131151 on Build GRCh38
rs867082144 Chr.6: 4130952-4130952 on Build GRCh38
rs927557245 Chr.6: 4130723-4130723 on Build GRCh38
rs369147181 Chr.6: 4130733-4130733 on Build GRCh38
rs927184489 Chr.6: 4131043-4131043 on Build GRCh38
rs779946632 Chr.6: 4130903-4130903 on Build GRCh38
rs979425344 Chr.6: 4130793-4130793 on Build GRCh38
rs764938282 Chr.6: 4130811-4130811 on Build GRCh38
rs761460792 Chr.6: 4130810-4130810 on Build GRCh38
rs7751079 Chr.6: 4130973-4130973 on Build GRCh38
rs773755094 Chr.6: 4130762-4130762 on Build GRCh38
rs555642385 Chr.6: 4130844-4130844 on Build GRCh38
rs1005543385 Chr.6: 4130941-4130941 on Build GRCh38
rs146934579 Chr.6: 4130837-4130837 on Build GRCh38
rs151097755 Chr.6: 4130719-4130719 on Build GRCh38
rs776229508 Chr.6: 4130806-4130806 on Build GRCh38
rs752328280 Chr.6: 4131104-4131104 on Build GRCh38
rs757678785 Chr.6: 4130886-4130886 on Build GRCh38
rs746978768 Chr.6: 4130915-4130915 on Build GRCh38
rs777962016 Chr.6: 4130727-4130727 on Build GRCh38
rs112029889 Chr.6: 4130999-4130999 on Build GRCh38
rs11394589 Chr.6: 4131124-4131125 on Build GRCh38
rs1010407931 Chr.6: 4131111-4131111 on Build GRCh38
rs972141351 Chr.6: 4130958-4130958 on Build GRCh38
rs757035827 Chr.6: 4131143-4131143 on Build GRCh38
rs933811111 Chr.6: 4130829-4130829 on Build GRCh38
rs763326832 Chr.6: 4130827-4130827 on Build GRCh38
rs148049194 Chr.6: 4130867-4130867 on Build GRCh38
rs745999661 Chr.6: 4130773-4130773 on Build GRCh38
rs1001694757 Chr.6: 4131057-4131057 on Build GRCh38
rs750063766 Chr.6: 4130825-4130825 on Build GRCh38
rs771257808 Chr.6: 4130908-4130909 on Build GRCh38
rs746056807 Chr.6: 4130897-4130897 on Build GRCh38
rs1050136887 Chr.6: 4130834-4130834 on Build GRCh38
rs557773452 Chr.6: 4131094-4131094 on Build GRCh38
rs897124995 Chr.6: 4130990-4130990 on Build GRCh38
rs930660227 Chr.6: 4130998-4130998 on Build GRCh38
rs762499811 Chr.6: 4130826-4130826 on Build GRCh38
rs1024366813 Chr.6: 4131018-4131018 on Build GRCh38
rs200673444 Chr.6: 4130797-4130797 on Build GRCh38
rs142680577 Chr.6: 4130796-4130796 on Build GRCh38
rs201337545 Chr.6: 4130854-4130854 on Build GRCh38
rs756545311 Chr.6: 4130721-4130721 on Build GRCh38
rs968378614 Chr.6: 4130769-4130769 on Build GRCh38
rs138632460 Chr.6: 4130794-4130794 on Build GRCh38
rs867265688 Chr.6: 4131119-4131119 on Build GRCh38
rs771209268 Chr.6: 4130734-4130734 on Build GRCh38
rs972004324 Chr.6: 4131039-4131039 on Build GRCh38
rs755735197 Chr.6: 4131112-4131112 on Build GRCh38
rs1042120389 Chr.6: 4130804-4130804 on Build GRCh38
rs772224819 Chr.6: 4130899-4130899 on Build GRCh38
rs779271218 Chr.6: 4130771-4130771 on Build GRCh38

View Full Product Details