Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4891634-4892131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL
Entrez Gene ID
9425
Gene Name
chromodomain protein, Y-like
Gene Aliases
CDYL1
UniGene
Hs.269092
Gene Chromosome Location
Chr.6: 4706159-4955544 on Build GRCh38
Transcripts
NM_001143970.1 NM_001143971.1 NM_004824.3 NR_026590.1 XM_011514997.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3628242 c.319C>T p.H107Y Chr.6: 4891845-4891845 on Build GRCh38
6581493 c.565C>G p.L189V Chr.6: 4892091-4892091 on Build GRCh38
1444982 c.216_217insA p.G75fs*13 Chr.6: 4891742-4891743 on Build GRCh38
1444985 c.313T>C p.Y105H Chr.6: 4891839-4891839 on Build GRCh38
5942031 c.448C>G p.P150A Chr.6: 4891974-4891974 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780525041 Chr.6: 4891977-4891977 on Build GRCh38
rs977467313 Chr.6: 4891674-4891674 on Build GRCh38
rs765139049 Chr.6: 4891928-4891928 on Build GRCh38
rs554548467 Chr.6: 4891891-4891891 on Build GRCh38
rs754537308 Chr.6: 4891963-4891963 on Build GRCh38
rs760861481 Chr.6: 4891909-4891909 on Build GRCh38
rs367701620 Chr.6: 4891995-4891995 on Build GRCh38
rs374024219 Chr.6: 4892076-4892076 on Build GRCh38
rs146246309 Chr.6: 4891848-4891848 on Build GRCh38
rs200111001 Chr.6: 4891861-4891861 on Build GRCh38
rs61749071 Chr.6: 4891847-4891847 on Build GRCh38
rs779392958 Chr.6: 4891993-4891993 on Build GRCh38
rs751147723 Chr.6: 4891960-4891960 on Build GRCh38
rs775548129 Chr.6: 4891905-4891905 on Build GRCh38
rs747161343 Chr.6: 4891997-4891997 on Build GRCh38
rs976298118 Chr.6: 4892004-4892004 on Build GRCh38
rs895253776 Chr.6: 4892059-4892059 on Build GRCh38
rs924488456 Chr.6: 4891853-4891853 on Build GRCh38
rs1021491896 Chr.6: 4892067-4892067 on Build GRCh38
rs61747454 Chr.6: 4891781-4891781 on Build GRCh38
rs924269796 Chr.6: 4891680-4891680 on Build GRCh38
rs766037143 Chr.6: 4892020-4892020 on Build GRCh38
rs200764082 Chr.6: 4891937-4891937 on Build GRCh38
rs200651601 Chr.6: 4891860-4891860 on Build GRCh38
rs370862716 Chr.6: 4892003-4892003 on Build GRCh38
rs369230031 Chr.6: 4891790-4891790 on Build GRCh38
rs780347868 Chr.6: 4891996-4891996 on Build GRCh38
rs776094407 Chr.6: 4891677-4891678 on Build GRCh38
rs769971723 Chr.6: 4892107-4892107 on Build GRCh38
rs746275548 Chr.6: 4891994-4891994 on Build GRCh38
rs985994885 Chr.6: 4891968-4891968 on Build GRCh38
rs148279388 Chr.6: 4891811-4891811 on Build GRCh38
rs766462758 Chr.6: 4891867-4891867 on Build GRCh38
rs756025610 Chr.6: 4891683-4891683 on Build GRCh38
rs747676317 Chr.6: 4891691-4891692 on Build GRCh38
rs561066951 Chr.6: 4891685-4891685 on Build GRCh38
rs771312380 Chr.6: 4891890-4891890 on Build GRCh38
rs768787781 Chr.6: 4891999-4891999 on Build GRCh38
rs759040134 Chr.6: 4892046-4892046 on Build GRCh38
rs778746146 Chr.6: 4891718-4891718 on Build GRCh38
rs116726726 Chr.6: 4891895-4891895 on Build GRCh38
rs770399503 Chr.6: 4891789-4891789 on Build GRCh38
rs752257604 Chr.6: 4892053-4892053 on Build GRCh38
rs145622412 Chr.6: 4891824-4891824 on Build GRCh38
rs11965336 Chr.6: 4891693-4891693 on Build GRCh38
rs943564261 Chr.6: 4891707-4891707 on Build GRCh38
rs749903480 Chr.6: 4891796-4891796 on Build GRCh38
rs759709338 Chr.6: 4891802-4891802 on Build GRCh38
rs954380105 Chr.6: 4891855-4891855 on Build GRCh38
rs779653039 Chr.6: 4891757-4891757 on Build GRCh38
rs532050073 Chr.6: 4892018-4892018 on Build GRCh38
rs201622502 Chr.6: 4891955-4891955 on Build GRCh38
rs113031834 Chr.6: 4891884-4891884 on Build GRCh38
rs773252426 Chr.6: 4892009-4892009 on Build GRCh38
rs758795798 Chr.6: 4892077-4892077 on Build GRCh38
rs779849243 Chr.6: 4891866-4891866 on Build GRCh38
rs906767899 Chr.6: 4891998-4891998 on Build GRCh38
rs200953580 Chr.6: 4892052-4892052 on Build GRCh38
rs1011404597 Chr.6: 4892063-4892063 on Build GRCh38
rs757700421 Chr.6: 4891986-4891986 on Build GRCh38
rs142899763 Chr.6: 4891985-4891985 on Build GRCh38
rs554754571 Chr.6: 4891699-4891699 on Build GRCh38
rs994154055 Chr.6: 4891739-4891739 on Build GRCh38
rs1047755992 Chr.6: 4891770-4891770 on Build GRCh38
rs753722279 Chr.6: 4891691-4891691 on Build GRCh38
rs370850783 Chr.6: 4891838-4891838 on Build GRCh38
rs771642793 Chr.6: 4892021-4892023 on Build GRCh38
rs925893817 Chr.6: 4891676-4891676 on Build GRCh38
rs764138201 Chr.6: 4891837-4891837 on Build GRCh38
rs377393556 Chr.6: 4891797-4891797 on Build GRCh38
rs762836740 Chr.6: 4891948-4891948 on Build GRCh38
rs370917045 Chr.6: 4891880-4891880 on Build GRCh38
rs150612356 Chr.6: 4892096-4892096 on Build GRCh38
rs745476824 Chr.6: 4891727-4891727 on Build GRCh38
rs537450737 Chr.6: 4891868-4891868 on Build GRCh38
rs544556054 Chr.6: 4891667-4891667 on Build GRCh38
rs747269304 Chr.6: 4892092-4892092 on Build GRCh38
rs749706969 Chr.6: 4891889-4891889 on Build GRCh38
rs538333704 Chr.6: 4891720-4891720 on Build GRCh38
rs139710301 Chr.6: 4892100-4892100 on Build GRCh38
rs977137388 Chr.6: 4891669-4891669 on Build GRCh38
rs767064076 Chr.6: 4892051-4892051 on Build GRCh38
rs759655988 Chr.6: 4891663-4891663 on Build GRCh38
rs755595055 Chr.6: 4892056-4892056 on Build GRCh38
rs930012575 Chr.6: 4891786-4891786 on Build GRCh38
rs770457472 Chr.6: 4891662-4891663 on Build GRCh38
rs957248962 Chr.6: 4891684-4891684 on Build GRCh38
rs752657919 Chr.6: 4891668-4891668 on Build GRCh38
rs758003601 Chr.6: 4891748-4891748 on Build GRCh38
rs143201617 Chr.6: 4891793-4891793 on Build GRCh38
rs775701266 Chr.6: 4891823-4891823 on Build GRCh38
rs776663806 Chr.6: 4891920-4891920 on Build GRCh38
rs769918692 Chr.6: 4892005-4892005 on Build GRCh38
rs72821458 Chr.6: 4892102-4892102 on Build GRCh38
rs779449574 Chr.6: 4892069-4892069 on Build GRCh38
rs751253394 Chr.6: 4892022-4892022 on Build GRCh38
rs764033923 Chr.6: 4891910-4891910 on Build GRCh38

View Full Product Details