Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4943408-4943911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDYL
Entrez Gene ID
9425
Gene Name
chromodomain protein, Y-like
Gene Aliases
CDYL1
UniGene
Hs.269092
Gene Chromosome Location
Chr.6: 4706159-4955544 on Build GRCh38
Transcripts
NM_001143970.1 NM_001143971.1 NM_004824.3 NR_026590.1 XM_011514997.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7425986 c.1410G>A p.W470* Chr.6: 4943672-4943672 on Build GRCh38
3874630 c.1301T>C p.F434S Chr.6: 4943563-4943563 on Build GRCh38
6213890 c.1493C>T p.S498F Chr.6: 4943755-4943755 on Build GRCh38
451603 c.1447G>A p.D483N Chr.6: 4943709-4943709 on Build GRCh38
1080065 c.1380C>T p.C460C Chr.6: 4943642-4943642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747108171 Chr.6: 4943504-4943504 on Build GRCh38
rs200106765 Chr.6: 4943787-4943788 on Build GRCh38
rs186263393 Chr.6: 4943486-4943486 on Build GRCh38
rs371414089 Chr.6: 4943583-4943583 on Build GRCh38
rs755116251 Chr.6: 4943665-4943665 on Build GRCh38
rs750894935 Chr.6: 4943769-4943769 on Build GRCh38
rs749085579 Chr.6: 4943699-4943699 on Build GRCh38
rs561012988 Chr.6: 4943773-4943774 on Build GRCh38
rs1041056917 Chr.6: 4943832-4943832 on Build GRCh38
rs768807410 Chr.6: 4943509-4943509 on Build GRCh38
rs758770404 Chr.6: 4943624-4943624 on Build GRCh38
rs751845675 Chr.6: 4943772-4943772 on Build GRCh38
rs747215746 Chr.6: 4943653-4943653 on Build GRCh38
rs762803541 Chr.6: 4943565-4943565 on Build GRCh38
rs372837191 Chr.6: 4943696-4943696 on Build GRCh38
rs944674130 Chr.6: 4943831-4943832 on Build GRCh38
rs556935041 Chr.6: 4943675-4943675 on Build GRCh38
rs562541753 Chr.6: 4943772-4943773 on Build GRCh38
rs772936198 Chr.6: 4943560-4943560 on Build GRCh38
rs752560222 Chr.6: 4943765-4943766 on Build GRCh38
rs1056780236 Chr.6: 4943846-4943846 on Build GRCh38
rs754998242 Chr.6: 4943503-4943503 on Build GRCh38
rs765772714 Chr.6: 4943614-4943614 on Build GRCh38
rs748032693 Chr.6: 4943536-4943536 on Build GRCh38
rs758537030 Chr.6: 4943766-4943767 on Build GRCh38
rs567567084 Chr.6: 4943789-4943789 on Build GRCh38
rs1027828684 Chr.6: 4943878-4943878 on Build GRCh38
rs775051909 Chr.6: 4943756-4943756 on Build GRCh38
rs201049668 Chr.6: 4943766-4943766 on Build GRCh38
rs11755468 Chr.6: 4943887-4943887 on Build GRCh38
rs938418404 Chr.6: 4943842-4943842 on Build GRCh38
rs571393565 Chr.6: 4943478-4943478 on Build GRCh38
rs1002516942 Chr.6: 4943473-4943473 on Build GRCh38
rs765716267 Chr.6: 4943766-4943766 on Build GRCh38
rs369415131 Chr.6: 4943678-4943678 on Build GRCh38
rs755308467 Chr.6: 4943775-4943775 on Build GRCh38
rs756239971 Chr.6: 4943806-4943806 on Build GRCh38
rs76863107 Chr.6: 4943770-4943770 on Build GRCh38
rs377263774 Chr.6: 4943528-4943528 on Build GRCh38
rs542941632 Chr.6: 4943727-4943727 on Build GRCh38
rs760150964 Chr.6: 4943765-4943765 on Build GRCh38
rs554967900 Chr.6: 4943771-4943773 on Build GRCh38
rs35797221 Chr.6: 4943787-4943787 on Build GRCh38
rs375737932 Chr.6: 4943669-4943669 on Build GRCh38
rs913805737 Chr.6: 4943526-4943526 on Build GRCh38
rs34649909 Chr.6: 4943771-4943772 on Build GRCh38
rs377158168 Chr.6: 4943770-4943771 on Build GRCh38
rs201124963 Chr.6: 4943657-4943657 on Build GRCh38
rs1046832304 Chr.6: 4943431-4943431 on Build GRCh38
rs898072090 Chr.6: 4943869-4943869 on Build GRCh38
rs796178409 Chr.6: 4943771-4943772 on Build GRCh38
rs781558668 Chr.6: 4943788-4943788 on Build GRCh38
rs1022334900 Chr.6: 4943492-4943492 on Build GRCh38
rs202149861 Chr.6: 4943539-4943539 on Build GRCh38
rs558235391 Chr.6: 4943501-4943501 on Build GRCh38
rs750797501 Chr.6: 4943616-4943616 on Build GRCh38
rs559873803 Chr.6: 4943773-4943773 on Build GRCh38
rs765987450 Chr.6: 4943568-4943568 on Build GRCh38
rs766971402 Chr.6: 4943591-4943591 on Build GRCh38
rs774029556 Chr.6: 4943570-4943570 on Build GRCh38
rs994114541 Chr.6: 4943871-4943871 on Build GRCh38
rs375233596 Chr.6: 4943724-4943724 on Build GRCh38
rs569061824 Chr.6: 4943769-4943770 on Build GRCh38
rs919864663 Chr.6: 4943689-4943689 on Build GRCh38
rs761271355 Chr.6: 4943810-4943810 on Build GRCh38
rs756989501 Chr.6: 4943552-4943552 on Build GRCh38
rs555710686 Chr.6: 4943447-4943447 on Build GRCh38
rs528300502 Chr.6: 4943771-4943771 on Build GRCh38
rs11963443 Chr.6: 4943612-4943612 on Build GRCh38
rs1046949954 Chr.6: 4943723-4943723 on Build GRCh38
rs886879515 Chr.6: 4943446-4943446 on Build GRCh38
rs373430007 Chr.6: 4943481-4943481 on Build GRCh38
rs780162299 Chr.6: 4943642-4943642 on Build GRCh38
rs1056829912 Chr.6: 4943480-4943480 on Build GRCh38
rs771765552 Chr.6: 4943776-4943777 on Build GRCh38
rs1033318193 Chr.6: 4943476-4943476 on Build GRCh38
rs78545686 Chr.6: 4943603-4943603 on Build GRCh38
rs200158214 Chr.6: 4943791-4943791 on Build GRCh38
rs772747436 Chr.6: 4943780-4943781 on Build GRCh38
rs894140594 Chr.6: 4943859-4943859 on Build GRCh38
rs767024470 Chr.6: 4943728-4943728 on Build GRCh38

View Full Product Details