Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7365351-7365851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAGE1
Entrez Gene ID
285782
Gene Name
cancer antigen 1
Gene Aliases
CT3, CT95, CTAG3, bA69L16.7
UniGene
Hs.721486
Gene Chromosome Location
Chr.6: 7326654-7389709 on Build GRCh38
Transcripts
NM_001170692.1 NM_001170693.1 NM_205864.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3629511 c.1696G>A p.D566N Chr.6: 7365476-7365476 on Build GRCh38
5099920 c.2086-9T>C p.? Chr.6: 7365584-7365584 on Build GRCh38
1596464 c.2125G>T p.D709Y Chr.6: 7365536-7365536 on Build GRCh38
1596463 c.2117G>T p.S706I Chr.6: 7365544-7365544 on Build GRCh38
7128655 c.2165A>G p.H722R Chr.6: 7365496-7365496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778196401 Chr.6: 7365531-7365531 on Build GRCh38
rs768324491 Chr.6: 7365580-7365580 on Build GRCh38
rs552820180 Chr.6: 7365783-7365783 on Build GRCh38
rs145708845 Chr.6: 7365500-7365500 on Build GRCh38
rs775261044 Chr.6: 7365569-7365569 on Build GRCh38
rs779165158 Chr.6: 7365820-7365820 on Build GRCh38
rs371788934 Chr.6: 7365521-7365521 on Build GRCh38
rs543196183 Chr.6: 7365509-7365509 on Build GRCh38
rs752069157 Chr.6: 7365506-7365506 on Build GRCh38
rs755405523 Chr.6: 7365418-7365418 on Build GRCh38
rs527881100 Chr.6: 7365515-7365515 on Build GRCh38
rs893070220 Chr.6: 7365472-7365472 on Build GRCh38
rs774495699 Chr.6: 7365451-7365451 on Build GRCh38
rs751422053 Chr.6: 7365489-7365489 on Build GRCh38
rs1016441657 Chr.6: 7365733-7365733 on Build GRCh38
rs998547270 Chr.6: 7365786-7365786 on Build GRCh38
rs772664293 Chr.6: 7365624-7365624 on Build GRCh38
rs934061421 Chr.6: 7365561-7365561 on Build GRCh38
rs200028605 Chr.6: 7365551-7365551 on Build GRCh38
rs200322379 Chr.6: 7365477-7365477 on Build GRCh38
rs761181979 Chr.6: 7365671-7365671 on Build GRCh38
rs201594718 Chr.6: 7365442-7365442 on Build GRCh38
rs776335230 Chr.6: 7365606-7365606 on Build GRCh38
rs746787880 Chr.6: 7365395-7365395 on Build GRCh38
rs773825211 Chr.6: 7365809-7365809 on Build GRCh38
rs372243525 Chr.6: 7365475-7365475 on Build GRCh38
rs764485022 Chr.6: 7365785-7365785 on Build GRCh38
rs959879334 Chr.6: 7365754-7365754 on Build GRCh38
rs374410435 Chr.6: 7365427-7365427 on Build GRCh38
rs1052909355 Chr.6: 7365803-7365803 on Build GRCh38
rs908692325 Chr.6: 7365696-7365696 on Build GRCh38
rs748334719 Chr.6: 7365426-7365426 on Build GRCh38
rs756409000 Chr.6: 7365526-7365526 on Build GRCh38
rs536117402 Chr.6: 7365712-7365712 on Build GRCh38
rs1006197546 Chr.6: 7365715-7365715 on Build GRCh38
rs200680929 Chr.6: 7365494-7365494 on Build GRCh38
rs761391627 Chr.6: 7365607-7365607 on Build GRCh38
rs368412460 Chr.6: 7365443-7365443 on Build GRCh38
rs771776697 Chr.6: 7365467-7365467 on Build GRCh38
rs934315586 Chr.6: 7365412-7365412 on Build GRCh38
rs768115230 Chr.6: 7365770-7365770 on Build GRCh38
rs188475258 Chr.6: 7365413-7365413 on Build GRCh38
rs192919356 Chr.6: 7365571-7365571 on Build GRCh38
rs754124311 Chr.6: 7365805-7365805 on Build GRCh38
rs779077314 Chr.6: 7365544-7365544 on Build GRCh38
rs529635591 Chr.6: 7365457-7365457 on Build GRCh38
rs554296838 Chr.6: 7365811-7365811 on Build GRCh38
rs982983558 Chr.6: 7365643-7365643 on Build GRCh38
rs781557179 Chr.6: 7365420-7365420 on Build GRCh38
rs915587341 Chr.6: 7365778-7365778 on Build GRCh38
rs759880738 Chr.6: 7365449-7365449 on Build GRCh38
rs762334844 Chr.6: 7365763-7365763 on Build GRCh38
rs745731899 Chr.6: 7365452-7365452 on Build GRCh38
rs375800106 Chr.6: 7365538-7365538 on Build GRCh38
rs777822446 Chr.6: 7365428-7365428 on Build GRCh38
rs941612100 Chr.6: 7365706-7365706 on Build GRCh38
rs776245516 Chr.6: 7365478-7365478 on Build GRCh38
rs767798625 Chr.6: 7365517-7365517 on Build GRCh38
rs547599964 Chr.6: 7365784-7365784 on Build GRCh38
rs775306425 Chr.6: 7365474-7365474 on Build GRCh38
rs768125928 Chr.6: 7365375-7365375 on Build GRCh38
rs764743876 Chr.6: 7365618-7365618 on Build GRCh38
rs772006207 Chr.6: 7365566-7365566 on Build GRCh38
rs565163363 Chr.6: 7365448-7365449 on Build GRCh38

View Full Product Details