Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10529337-10529848 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCNT2
Entrez Gene ID
2651
Gene Name
glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group)
Gene Aliases
CCAT, CTRCT13, GCNT2C, GCNT5, IGNT, II, NACGT1, NAGCT1, ULG3, bA360O19.2, bA421M1.1
UniGene
Hs.519884
Gene Chromosome Location
Chr.6: 10521283-10629368 on Build GRCh38
Transcripts
NM_145649.4 XM_005248999.2 XM_006715052.3 XM_011514468.2 XM_017010732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7146317 c.732G>T p.V244V Chr.6: 10529643-10529643 on Build GRCh38
5635962 c.672C>T p.H224H Chr.6: 10529583-10529583 on Build GRCh38
3721909 c.474G>A p.S158S Chr.6: 10529385-10529385 on Build GRCh38
5571111 c.829G>A p.V277I Chr.6: 10529740-10529740 on Build GRCh38
7141873 c.777G>T p.V259V Chr.6: 10529688-10529688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771800934 Chr.6: 10529551-10529551 on Build GRCh38
rs753095004 Chr.6: 10529480-10529480 on Build GRCh38
rs779063703 Chr.6: 10529766-10529766 on Build GRCh38
rs774530016 Chr.6: 10529431-10529432 on Build GRCh38
rs972573195 Chr.6: 10529654-10529654 on Build GRCh38
rs913588782 Chr.6: 10529744-10529744 on Build GRCh38
rs575251024 Chr.6: 10529520-10529520 on Build GRCh38
rs767987662 Chr.6: 10529650-10529650 on Build GRCh38
rs541962993 Chr.6: 10529460-10529460 on Build GRCh38
rs772454113 Chr.6: 10529715-10529715 on Build GRCh38
rs774604411 Chr.6: 10529710-10529710 on Build GRCh38
rs761096313 Chr.6: 10529720-10529720 on Build GRCh38
rs937874777 Chr.6: 10529787-10529787 on Build GRCh38
rs888354864 Chr.6: 10529576-10529576 on Build GRCh38
rs745931655 Chr.6: 10529611-10529611 on Build GRCh38
rs200052359 Chr.6: 10529771-10529771 on Build GRCh38
rs746871443 Chr.6: 10529599-10529599 on Build GRCh38
rs532935757 Chr.6: 10529739-10529739 on Build GRCh38
rs757985984 Chr.6: 10529668-10529668 on Build GRCh38
rs139794913 Chr.6: 10529803-10529803 on Build GRCh38
rs759596259 Chr.6: 10529558-10529558 on Build GRCh38
rs779642200 Chr.6: 10529426-10529426 on Build GRCh38
rs1046528944 Chr.6: 10529776-10529776 on Build GRCh38
rs774692488 Chr.6: 10529641-10529641 on Build GRCh38
rs147103060 Chr.6: 10529604-10529604 on Build GRCh38
rs540411690 Chr.6: 10529573-10529573 on Build GRCh38
rs186235421 Chr.6: 10529548-10529548 on Build GRCh38
rs750509497 Chr.6: 10529374-10529374 on Build GRCh38
rs758159782 Chr.6: 10529593-10529593 on Build GRCh38
rs754060575 Chr.6: 10529469-10529469 on Build GRCh38
rs771822241 Chr.6: 10529773-10529773 on Build GRCh38
rs767843763 Chr.6: 10529597-10529598 on Build GRCh38
rs752139864 Chr.6: 10529725-10529725 on Build GRCh38
rs776850375 Chr.6: 10529556-10529556 on Build GRCh38
rs949664812 Chr.6: 10529774-10529774 on Build GRCh38
rs141557910 Chr.6: 10529553-10529553 on Build GRCh38
rs986818634 Chr.6: 10529475-10529475 on Build GRCh38
rs377397535 Chr.6: 10529709-10529709 on Build GRCh38
rs757942928 Chr.6: 10529686-10529687 on Build GRCh38
rs975887471 Chr.6: 10529700-10529700 on Build GRCh38
rs962108296 Chr.6: 10529399-10529399 on Build GRCh38
rs758740184 Chr.6: 10529484-10529484 on Build GRCh38
rs748961646 Chr.6: 10529439-10529439 on Build GRCh38
rs764671795 Chr.6: 10529666-10529666 on Build GRCh38
rs760802120 Chr.6: 10529462-10529462 on Build GRCh38
rs955070677 Chr.6: 10529683-10529683 on Build GRCh38
rs764932357 Chr.6: 10529647-10529647 on Build GRCh38
rs9466560 Chr.6: 10529754-10529754 on Build GRCh38
rs767344843 Chr.6: 10529647-10529647 on Build GRCh38
rs148004068 Chr.6: 10529743-10529743 on Build GRCh38
rs761578619 Chr.6: 10529457-10529457 on Build GRCh38
rs781147997 Chr.6: 10529742-10529742 on Build GRCh38
rs753082290 Chr.6: 10529402-10529402 on Build GRCh38
rs564014638 Chr.6: 10529555-10529555 on Build GRCh38
rs547127442 Chr.6: 10529789-10529789 on Build GRCh38
rs150542637 Chr.6: 10529802-10529802 on Build GRCh38
rs376038250 Chr.6: 10529523-10529523 on Build GRCh38
rs867792819 Chr.6: 10529572-10529573 on Build GRCh38
rs755986508 Chr.6: 10529822-10529822 on Build GRCh38
rs984752836 Chr.6: 10529395-10529395 on Build GRCh38
rs182849633 Chr.6: 10529731-10529731 on Build GRCh38
rs887798958 Chr.6: 10529528-10529528 on Build GRCh38
rs762582303 Chr.6: 10529583-10529583 on Build GRCh38
rs753331219 Chr.6: 10529575-10529575 on Build GRCh38
rs137853339 Chr.6: 10529416-10529416 on Build GRCh38
rs370727036 Chr.6: 10529735-10529735 on Build GRCh38
rs892103872 Chr.6: 10529531-10529531 on Build GRCh38
rs561470405 Chr.6: 10529394-10529394 on Build GRCh38
rs923129212 Chr.6: 10529707-10529707 on Build GRCh38
rs551445279 Chr.6: 10529755-10529755 on Build GRCh38
rs764632923 Chr.6: 10529485-10529485 on Build GRCh38
rs76555271 Chr.6: 10529760-10529760 on Build GRCh38
rs781238161 Chr.6: 10529678-10529678 on Build GRCh38
rs780003599 Chr.6: 10529703-10529703 on Build GRCh38
rs761163111 Chr.6: 10529775-10529775 on Build GRCh38
rs375575478 Chr.6: 10529758-10529758 on Build GRCh38
rs751381070 Chr.6: 10529675-10529675 on Build GRCh38
rs769797010 Chr.6: 10529614-10529614 on Build GRCh38
rs777630944 Chr.6: 10529674-10529674 on Build GRCh38
rs762413480 Chr.6: 10529573-10529574 on Build GRCh38
rs752553216 Chr.6: 10529670-10529670 on Build GRCh38
rs772360747 Chr.6: 10529772-10529772 on Build GRCh38
rs778485764 Chr.6: 10529536-10529536 on Build GRCh38
rs368830625 Chr.6: 10529809-10529809 on Build GRCh38
rs553334210 Chr.6: 10529393-10529393 on Build GRCh38
rs923450700 Chr.6: 10529435-10529435 on Build GRCh38
rs137853340 Chr.6: 10529594-10529594 on Build GRCh38
rs561742156 Chr.6: 10529595-10529595 on Build GRCh38
rs763720351 Chr.6: 10529800-10529800 on Build GRCh38
rs766980413 Chr.6: 10529723-10529723 on Build GRCh38
rs775617613 Chr.6: 10529563-10529563 on Build GRCh38
rs749451195 Chr.6: 10529708-10529708 on Build GRCh38
rs762221929 Chr.6: 10529642-10529642 on Build GRCh38
rs575843044 Chr.6: 10529362-10529362 on Build GRCh38
rs143988975 Chr.6: 10529653-10529653 on Build GRCh38
rs775733706 Chr.6: 10529636-10529636 on Build GRCh38
rs760893627 Chr.6: 10529381-10529381 on Build GRCh38
rs767428497 Chr.6: 10529373-10529373 on Build GRCh38
rs777237277 Chr.6: 10529793-10529793 on Build GRCh38
rs747596532 Chr.6: 10529374-10529376 on Build GRCh38
rs751289332 Chr.6: 10529740-10529740 on Build GRCh38
rs923329043 Chr.6: 10529397-10529397 on Build GRCh38
rs766594355 Chr.6: 10529463-10529463 on Build GRCh38
rs150206842 Chr.6: 10529478-10529478 on Build GRCh38
rs764389975 Chr.6: 10529565-10529565 on Build GRCh38
rs768277361 Chr.6: 10529444-10529444 on Build GRCh38
rs201295923 Chr.6: 10529446-10529446 on Build GRCh38
rs773715942 Chr.6: 10529719-10529719 on Build GRCh38
rs367912474 Chr.6: 10529799-10529799 on Build GRCh38
rs755572823 Chr.6: 10529503-10529503 on Build GRCh38
rs778203608 Chr.6: 10529404-10529404 on Build GRCh38
rs759043571 Chr.6: 10529659-10529659 on Build GRCh38
rs779353582 Chr.6: 10529398-10529398 on Build GRCh38
rs773968312 Chr.6: 10529605-10529605 on Build GRCh38
rs757766007 Chr.6: 10529567-10529567 on Build GRCh38
rs143363982 Chr.6: 10529689-10529689 on Build GRCh38
rs772030668 Chr.6: 10529459-10529459 on Build GRCh38
rs769335145 Chr.6: 10529413-10529413 on Build GRCh38
rs368989684 Chr.6: 10529368-10529368 on Build GRCh38
rs769855709 Chr.6: 10529702-10529702 on Build GRCh38
rs772673036 Chr.6: 10529564-10529564 on Build GRCh38
rs141698805 Chr.6: 10529761-10529761 on Build GRCh38
rs779553822 Chr.6: 10529512-10529512 on Build GRCh38
rs750171358 Chr.6: 10529820-10529820 on Build GRCh38
rs766682666 Chr.6: 10529384-10529384 on Build GRCh38
rs759847093 Chr.6: 10529470-10529470 on Build GRCh38
rs373886978 Chr.6: 10529640-10529640 on Build GRCh38
rs752093697 Chr.6: 10529495-10529495 on Build GRCh38
rs780096446 Chr.6: 10529757-10529757 on Build GRCh38
rs199904355 Chr.6: 10529562-10529562 on Build GRCh38
rs763984017 Chr.6: 10529770-10529771 on Build GRCh38
rs758943227 Chr.6: 10529403-10529403 on Build GRCh38
rs752005643 Chr.6: 10529566-10529566 on Build GRCh38
rs752310750 Chr.6: 10529666-10529668 on Build GRCh38
rs754150475 Chr.6: 10529385-10529385 on Build GRCh38
rs972133234 Chr.6: 10529417-10529417 on Build GRCh38
rs752332523 Chr.6: 10529667-10529667 on Build GRCh38
rs754493637 Chr.6: 10529582-10529582 on Build GRCh38
rs757161386 Chr.6: 10529677-10529677 on Build GRCh38
rs747951151 Chr.6: 10529546-10529546 on Build GRCh38
rs958722438 Chr.6: 10529382-10529382 on Build GRCh38
rs759030699 Chr.6: 10529621-10529653 on Build GRCh38
rs776129708 Chr.6: 10529602-10529603 on Build GRCh38
rs998552380 Chr.6: 10529811-10529811 on Build GRCh38
rs373238065 Chr.6: 10529488-10529488 on Build GRCh38

View Full Product Details