Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10873499-10874005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCM2
Entrez Gene ID
9247
Gene Name
glial cells missing homolog 2
Gene Aliases
GCMB, hGCMb
UniGene
Hs.227098
Gene Chromosome Location
Chr.6: 10873223-10881865 on Build GRCh38
Transcripts
NM_004752.3

SNP Detail

SNP ID Chromosome Location
rs971674088 Chr.6: 10873931-10873931 on Build GRCh38
rs1016155800 Chr.6: 10873816-10873816 on Build GRCh38
rs201822843 Chr.6: 10873961-10873961 on Build GRCh38
rs989640336 Chr.6: 10873956-10873956 on Build GRCh38
rs111930987 Chr.6: 10873755-10873755 on Build GRCh38
rs981856109 Chr.6: 10873877-10873877 on Build GRCh38
rs191527371 Chr.6: 10873792-10873792 on Build GRCh38
rs866777825 Chr.6: 10873778-10873778 on Build GRCh38
rs746217594 Chr.6: 10873945-10873945 on Build GRCh38
rs562115657 Chr.6: 10873687-10873687 on Build GRCh38
rs1023325886 Chr.6: 10873848-10873848 on Build GRCh38
rs990891642 Chr.6: 10873831-10873831 on Build GRCh38
rs938017667 Chr.6: 10873586-10873586 on Build GRCh38
rs746425059 Chr.6: 10873896-10873896 on Build GRCh38
rs923712442 Chr.6: 10873928-10873928 on Build GRCh38
rs796803799 Chr.6: 10873712-10873712 on Build GRCh38
rs1037252420 Chr.6: 10873722-10873722 on Build GRCh38
rs963362203 Chr.6: 10873820-10873820 on Build GRCh38
rs970870134 Chr.6: 10873866-10873866 on Build GRCh38
rs928029949 Chr.6: 10873570-10873570 on Build GRCh38
rs924141636 Chr.6: 10873529-10873529 on Build GRCh38
rs770348814 Chr.6: 10873950-10873950 on Build GRCh38
rs6942384 Chr.6: 10873555-10873555 on Build GRCh38
rs892761284 Chr.6: 10873875-10873878 on Build GRCh38
rs140170781 Chr.6: 10873662-10873662 on Build GRCh38
rs909853850 Chr.6: 10873699-10873699 on Build GRCh38
rs1025744720 Chr.6: 10873773-10873773 on Build GRCh38
rs1039746276 Chr.6: 10873714-10873714 on Build GRCh38
rs533104557 Chr.6: 10873782-10873782 on Build GRCh38
rs1020191072 Chr.6: 10873883-10873883 on Build GRCh38
rs541026596 Chr.6: 10873707-10873707 on Build GRCh38
rs116313934 Chr.6: 10873721-10873721 on Build GRCh38
rs935515272 Chr.6: 10873546-10873546 on Build GRCh38
rs866313660 Chr.6: 10873565-10873565 on Build GRCh38
rs941625822 Chr.6: 10873706-10873706 on Build GRCh38
rs1006568724 Chr.6: 10873805-10873805 on Build GRCh38
rs551033921 Chr.6: 10873720-10873720 on Build GRCh38
rs982223858 Chr.6: 10873569-10873569 on Build GRCh38
rs1054069218 Chr.6: 10873812-10873812 on Build GRCh38
rs758323182 Chr.6: 10873583-10873583 on Build GRCh38
rs529312846 Chr.6: 10873703-10873703 on Build GRCh38

View Full Product Details