Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10981003-10981548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL2
Entrez Gene ID
54898
Gene Name
ELOVL fatty acid elongase 2
Gene Aliases
SSC2
UniGene
Hs.656436
Gene Chromosome Location
Chr.6: 10980759-11044391 on Build GRCh38
Transcripts
NM_017770.3 XM_011514716.2 XM_011514717.2

SNP Detail

SNP ID Chromosome Location
rs778037706 Chr.6: 10981187-10981187 on Build GRCh38
rs1015058472 Chr.6: 10981208-10981208 on Build GRCh38
rs927908531 Chr.6: 10981397-10981397 on Build GRCh38
rs1052574457 Chr.6: 10981276-10981276 on Build GRCh38
rs3734396 Chr.6: 10981265-10981265 on Build GRCh38
rs1028624875 Chr.6: 10981188-10981188 on Build GRCh38
rs560760984 Chr.6: 10981251-10981251 on Build GRCh38
rs537092360 Chr.6: 10981108-10981108 on Build GRCh38
rs904438926 Chr.6: 10981140-10981140 on Build GRCh38
rs1052552250 Chr.6: 10981441-10981441 on Build GRCh38
rs536053583 Chr.6: 10981506-10981506 on Build GRCh38
rs935497341 Chr.6: 10981275-10981276 on Build GRCh38
rs139850682 Chr.6: 10981088-10981088 on Build GRCh38
rs549769938 Chr.6: 10981507-10981507 on Build GRCh38
rs995961961 Chr.6: 10981419-10981419 on Build GRCh38
rs981186244 Chr.6: 10981364-10981364 on Build GRCh38
rs149992735 Chr.6: 10981206-10981206 on Build GRCh38
rs543196945 Chr.6: 10981465-10981465 on Build GRCh38
rs943961198 Chr.6: 10981190-10981190 on Build GRCh38
rs868531107 Chr.6: 10981246-10981246 on Build GRCh38
rs769352659 Chr.6: 10981371-10981373 on Build GRCh38
rs951199991 Chr.6: 10981100-10981100 on Build GRCh38
rs183896935 Chr.6: 10981226-10981226 on Build GRCh38
rs1027062263 Chr.6: 10981427-10981427 on Build GRCh38
rs1021986288 Chr.6: 10981113-10981113 on Build GRCh38
rs1042889276 Chr.6: 10981475-10981475 on Build GRCh38
rs769252777 Chr.6: 10981452-10981452 on Build GRCh38
rs996280227 Chr.6: 10981177-10981177 on Build GRCh38
rs1027192263 Chr.6: 10981089-10981089 on Build GRCh38
rs1023664605 Chr.6: 10981044-10981044 on Build GRCh38
rs536427327 Chr.6: 10981083-10981083 on Build GRCh38
rs747579070 Chr.6: 10981373-10981373 on Build GRCh38
rs1010063072 Chr.6: 10981094-10981094 on Build GRCh38
rs1013801953 Chr.6: 10981292-10981292 on Build GRCh38
rs954464194 Chr.6: 10981189-10981189 on Build GRCh38
rs896727852 Chr.6: 10981288-10981288 on Build GRCh38
rs765033763 Chr.6: 10981285-10981286 on Build GRCh38
rs905194447 Chr.6: 10981395-10981395 on Build GRCh38
rs939432927 Chr.6: 10981434-10981434 on Build GRCh38
rs969846869 Chr.6: 10981053-10981053 on Build GRCh38
rs550407768 Chr.6: 10981316-10981316 on Build GRCh38
rs892537553 Chr.6: 10981461-10981461 on Build GRCh38
rs890091193 Chr.6: 10981511-10981511 on Build GRCh38
rs554404200 Chr.6: 10981085-10981085 on Build GRCh38
rs570304183 Chr.6: 10981495-10981495 on Build GRCh38
rs867377581 Chr.6: 10981429-10981429 on Build GRCh38
rs948320208 Chr.6: 10981340-10981340 on Build GRCh38
rs191462351 Chr.6: 10981212-10981212 on Build GRCh38
rs962214113 Chr.6: 10981218-10981218 on Build GRCh38
rs777181509 Chr.6: 10981471-10981471 on Build GRCh38

View Full Product Details