Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 10981440-10981950 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL2
Entrez Gene ID
54898
Gene Name
ELOVL fatty acid elongase 2
Gene Aliases
SSC2
UniGene
Hs.656436
Gene Chromosome Location
Chr.6: 10980759-11044391 on Build GRCh38
Transcripts
NM_017770.3 XM_011514716.2 XM_011514717.2

SNP Detail

SNP ID Chromosome Location
rs762453676 Chr.6: 10981736-10981736 on Build GRCh38
rs548366073 Chr.6: 10981813-10981813 on Build GRCh38
rs1028235328 Chr.6: 10981870-10981870 on Build GRCh38
rs200599828 Chr.6: 10981700-10981700 on Build GRCh38
rs931905062 Chr.6: 10981821-10981821 on Build GRCh38
rs997134884 Chr.6: 10981865-10981865 on Build GRCh38
rs978436649 Chr.6: 10981925-10981925 on Build GRCh38
rs924344514 Chr.6: 10981691-10981691 on Build GRCh38
rs565951205 Chr.6: 10981774-10981774 on Build GRCh38
rs192834243 Chr.6: 10981855-10981855 on Build GRCh38
rs536053583 Chr.6: 10981506-10981506 on Build GRCh38
rs905475081 Chr.6: 10981749-10981749 on Build GRCh38
rs975377047 Chr.6: 10981533-10981533 on Build GRCh38
rs866176037 Chr.6: 10981833-10981833 on Build GRCh38
rs887177994 Chr.6: 10981789-10981789 on Build GRCh38
rs762450792 Chr.6: 10981669-10981669 on Build GRCh38
rs961766075 Chr.6: 10981646-10981646 on Build GRCh38
rs564898933 Chr.6: 10981536-10981536 on Build GRCh38
rs111843465 Chr.6: 10981779-10981779 on Build GRCh38
rs549769938 Chr.6: 10981507-10981507 on Build GRCh38
rs981020968 Chr.6: 10981709-10981709 on Build GRCh38
rs547317816 Chr.6: 10981719-10981719 on Build GRCh38
rs45621340 Chr.6: 10981818-10981818 on Build GRCh38
rs988138785 Chr.6: 10981737-10981737 on Build GRCh38
rs890091193 Chr.6: 10981511-10981511 on Build GRCh38
rs914023978 Chr.6: 10981757-10981757 on Build GRCh38
rs768220353 Chr.6: 10981723-10981723 on Build GRCh38
rs543196945 Chr.6: 10981465-10981465 on Build GRCh38
rs570304183 Chr.6: 10981495-10981495 on Build GRCh38
rs3823014 Chr.6: 10981610-10981610 on Build GRCh38
rs558603555 Chr.6: 10981898-10981898 on Build GRCh38
rs978870392 Chr.6: 10981687-10981687 on Build GRCh38
rs879914612 Chr.6: 10981706-10981706 on Build GRCh38
rs995447006 Chr.6: 10981663-10981663 on Build GRCh38
rs969062803 Chr.6: 10981904-10981904 on Build GRCh38
rs578037830 Chr.6: 10981860-10981860 on Build GRCh38
rs1043285506 Chr.6: 10981814-10981814 on Build GRCh38
rs1027787517 Chr.6: 10981666-10981666 on Build GRCh38
rs1042889276 Chr.6: 10981475-10981475 on Build GRCh38
rs1048898566 Chr.6: 10981782-10981788 on Build GRCh38
rs375148857 Chr.6: 10981840-10981840 on Build GRCh38
rs1008556315 Chr.6: 10981879-10981879 on Build GRCh38
rs777181509 Chr.6: 10981471-10981471 on Build GRCh38
rs1035977876 Chr.6: 10981922-10981922 on Build GRCh38

View Full Product Details