Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12296248-12296759 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
UniGene
Hs.511899
Gene Chromosome Location
Chr.6: 12256464-12297194 on Build GRCh38
Transcripts
NM_001168319.1 NM_001955.4 XM_011514330.2 XM_011514331.2 XM_011514332.2 XM_017010331.1

SNP Detail

SNP ID Chromosome Location
rs974939197 Chr.6: 12296569-12296569 on Build GRCh38
rs907518493 Chr.6: 12296608-12296608 on Build GRCh38
rs1023045945 Chr.6: 12296388-12296388 on Build GRCh38
rs1032153921 Chr.6: 12296469-12296469 on Build GRCh38
rs996017876 Chr.6: 12296375-12296375 on Build GRCh38
rs539153026 Chr.6: 12296338-12296338 on Build GRCh38
rs989033958 Chr.6: 12296484-12296484 on Build GRCh38
rs940639056 Chr.6: 12296633-12296633 on Build GRCh38
rs538409865 Chr.6: 12296653-12296653 on Build GRCh38
rs766557431 Chr.6: 12296359-12296359 on Build GRCh38
rs185307338 Chr.6: 12296564-12296564 on Build GRCh38
rs553081608 Chr.6: 12296339-12296339 on Build GRCh38
rs41332046 Chr.6: 12296308-12296308 on Build GRCh38
rs1045266557 Chr.6: 12296731-12296731 on Build GRCh38
rs1012072985 Chr.6: 12296722-12296722 on Build GRCh38
rs1021355314 Chr.6: 12296542-12296543 on Build GRCh38
rs997838490 Chr.6: 12296416-12296416 on Build GRCh38
rs189274493 Chr.6: 12296345-12296345 on Build GRCh38
rs374886874 Chr.6: 12296379-12296379 on Build GRCh38
rs773410128 Chr.6: 12296719-12296719 on Build GRCh38
rs1004731360 Chr.6: 12296354-12296354 on Build GRCh38
rs970792658 Chr.6: 12296543-12296543 on Build GRCh38
rs576029963 Chr.6: 12296650-12296650 on Build GRCh38
rs941809618 Chr.6: 12296369-12296369 on Build GRCh38
rs578133364 Chr.6: 12296312-12296312 on Build GRCh38
rs910065667 Chr.6: 12296642-12296642 on Build GRCh38
rs970496140 Chr.6: 12296392-12296392 on Build GRCh38
rs1002884201 Chr.6: 12296423-12296423 on Build GRCh38
rs1036140818 Chr.6: 12296705-12296705 on Build GRCh38
rs142576582 Chr.6: 12296360-12296361 on Build GRCh38
rs11969143 Chr.6: 12296532-12296532 on Build GRCh38
rs1035652528 Chr.6: 12296431-12296431 on Build GRCh38
rs956294314 Chr.6: 12296442-12296442 on Build GRCh38
rs559273970 Chr.6: 12296602-12296602 on Build GRCh38
rs1051383197 Chr.6: 12296603-12296603 on Build GRCh38
rs886368413 Chr.6: 12296344-12296344 on Build GRCh38
rs182061188 Chr.6: 12296358-12296358 on Build GRCh38
rs899031542 Chr.6: 12296660-12296660 on Build GRCh38
rs186847861 Chr.6: 12296273-12296273 on Build GRCh38
rs112399848 Chr.6: 12296467-12296467 on Build GRCh38
rs544104028 Chr.6: 12296542-12296542 on Build GRCh38
rs41328054 Chr.6: 12296505-12296505 on Build GRCh38
rs554956466 Chr.6: 12296530-12296530 on Build GRCh38
rs140005633 Chr.6: 12296724-12296724 on Build GRCh38
rs150657531 Chr.6: 12296599-12296599 on Build GRCh38
rs897521262 Chr.6: 12296391-12296391 on Build GRCh38
rs1007306656 Chr.6: 12296443-12296443 on Build GRCh38

View Full Product Details