Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12297053-12297549 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDN1
Entrez Gene ID
1906
Gene Name
endothelin 1
Gene Aliases
ARCND3, ET1, HDLCQ7, PPET1, QME
UniGene
Hs.511899
Gene Chromosome Location
Chr.6: 12256464-12297194 on Build GRCh38
Transcripts
NM_001168319.1 NM_001955.4 XM_011514330.2 XM_011514331.2 XM_011514332.2 XM_017010331.1

SNP Detail

SNP ID Chromosome Location
rs973044321 Chr.6: 12297307-12297307 on Build GRCh38
rs557675903 Chr.6: 12297476-12297476 on Build GRCh38
rs981083496 Chr.6: 12297312-12297312 on Build GRCh38
rs945196831 Chr.6: 12297252-12297252 on Build GRCh38
rs891843175 Chr.6: 12297130-12297130 on Build GRCh38
rs895173638 Chr.6: 12297332-12297332 on Build GRCh38
rs1043083787 Chr.6: 12297418-12297418 on Build GRCh38
rs534434058 Chr.6: 12297277-12297277 on Build GRCh38
rs915124314 Chr.6: 12297456-12297456 on Build GRCh38
rs1010234988 Chr.6: 12297131-12297131 on Build GRCh38
rs747436362 Chr.6: 12297207-12297207 on Build GRCh38
rs10478730 Chr.6: 12297276-12297276 on Build GRCh38
rs762332377 Chr.6: 12297313-12297313 on Build GRCh38
rs938777413 Chr.6: 12297108-12297108 on Build GRCh38
rs1042617211 Chr.6: 12297259-12297262 on Build GRCh38
rs972901237 Chr.6: 12297398-12297398 on Build GRCh38
rs961931048 Chr.6: 12297360-12297360 on Build GRCh38
rs891895683 Chr.6: 12297382-12297382 on Build GRCh38
rs554893339 Chr.6: 12297424-12297424 on Build GRCh38
rs1057272360 Chr.6: 12297368-12297368 on Build GRCh38
rs1053912115 Chr.6: 12297493-12297493 on Build GRCh38
rs373209923 Chr.6: 12297310-12297310 on Build GRCh38
rs552776161 Chr.6: 12297311-12297311 on Build GRCh38
rs1042195874 Chr.6: 12297494-12297494 on Build GRCh38
rs141168313 Chr.6: 12297237-12297237 on Build GRCh38
rs1030254886 Chr.6: 12297401-12297401 on Build GRCh38
rs987218627 Chr.6: 12297264-12297264 on Build GRCh38
rs778612709 Chr.6: 12297110-12297110 on Build GRCh38
rs189972505 Chr.6: 12297402-12297402 on Build GRCh38
rs773171181 Chr.6: 12297373-12297373 on Build GRCh38
rs1029189156 Chr.6: 12297253-12297253 on Build GRCh38
rs955023598 Chr.6: 12297258-12297258 on Build GRCh38
rs1053748687 Chr.6: 12297241-12297242 on Build GRCh38
rs763033387 Chr.6: 12297438-12297438 on Build GRCh38
rs182937647 Chr.6: 12297453-12297453 on Build GRCh38
rs544091619 Chr.6: 12297466-12297466 on Build GRCh38
rs10478731 Chr.6: 12297333-12297333 on Build GRCh38
rs1017150685 Chr.6: 12297344-12297344 on Build GRCh38
rs4714383 Chr.6: 12297249-12297249 on Build GRCh38
rs552938301 Chr.6: 12297271-12297271 on Build GRCh38
rs904506592 Chr.6: 12297422-12297422 on Build GRCh38

View Full Product Details