Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13787238-13787737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUR1
Entrez Gene ID
63933
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
C6orf79, CCDC90A, FMP32
UniGene
Hs.214043
Gene Chromosome Location
Chr.6: 13786549-13814994 on Build GRCh38
Transcripts
NM_001031713.3 XM_011514802.1

SNP Detail

SNP ID Chromosome Location
rs149871670 Chr.6: 13787602-13787602 on Build GRCh38
rs767923709 Chr.6: 13787640-13787640 on Build GRCh38
rs1030139496 Chr.6: 13787477-13787477 on Build GRCh38
rs993762793 Chr.6: 13787469-13787469 on Build GRCh38
rs372516479 Chr.6: 13787643-13787643 on Build GRCh38
rs919832225 Chr.6: 13787323-13787323 on Build GRCh38
rs904064902 Chr.6: 13787470-13787470 on Build GRCh38
rs1017605569 Chr.6: 13787520-13787520 on Build GRCh38
rs966967083 Chr.6: 13787587-13787587 on Build GRCh38
rs776129164 Chr.6: 13787648-13787648 on Build GRCh38
rs938429836 Chr.6: 13787299-13787299 on Build GRCh38
rs929254696 Chr.6: 13787646-13787646 on Build GRCh38
rs1011345984 Chr.6: 13787694-13787694 on Build GRCh38
rs575492972 Chr.6: 13787313-13787313 on Build GRCh38
rs915656992 Chr.6: 13787620-13787620 on Build GRCh38
rs923386844 Chr.6: 13787370-13787370 on Build GRCh38
rs967084278 Chr.6: 13787705-13787705 on Build GRCh38
rs747779530 Chr.6: 13787585-13787585 on Build GRCh38
rs952660135 Chr.6: 13787338-13787338 on Build GRCh38
rs774148580 Chr.6: 13787364-13787364 on Build GRCh38
rs3734671 Chr.6: 13787560-13787560 on Build GRCh38
rs139113435 Chr.6: 13787499-13787499 on Build GRCh38
rs112540233 Chr.6: 13787601-13787603 on Build GRCh38
rs893875429 Chr.6: 13787689-13787689 on Build GRCh38
rs376405891 Chr.6: 13787461-13787461 on Build GRCh38
rs890259885 Chr.6: 13787479-13787479 on Build GRCh38
rs369922310 Chr.6: 13787615-13787615 on Build GRCh38
rs16874329 Chr.6: 13787608-13787608 on Build GRCh38
rs79564501 Chr.6: 13787637-13787637 on Build GRCh38
rs1060117 Chr.6: 13787695-13787695 on Build GRCh38
rs928389936 Chr.6: 13787298-13787298 on Build GRCh38
rs773305438 Chr.6: 13787483-13787483 on Build GRCh38
rs3832437 Chr.6: 13787331-13787331 on Build GRCh38
rs542603133 Chr.6: 13787592-13787592 on Build GRCh38
rs1038058212 Chr.6: 13787437-13787437 on Build GRCh38
rs1013789839 Chr.6: 13787524-13787524 on Build GRCh38
rs768433201 Chr.6: 13787433-13787433 on Build GRCh38
rs567727749 Chr.6: 13787702-13787702 on Build GRCh38
rs536719891 Chr.6: 13787512-13787512 on Build GRCh38
rs986503299 Chr.6: 13787702-13787703 on Build GRCh38
rs892619502 Chr.6: 13787521-13787521 on Build GRCh38
rs371700434 Chr.6: 13787605-13787607 on Build GRCh38
rs577421303 Chr.6: 13787381-13787381 on Build GRCh38
rs866318854 Chr.6: 13787452-13787452 on Build GRCh38
rs941664827 Chr.6: 13787362-13787362 on Build GRCh38
rs1007608072 Chr.6: 13787498-13787498 on Build GRCh38
rs1050488824 Chr.6: 13787409-13787409 on Build GRCh38
rs772632238 Chr.6: 13787612-13787612 on Build GRCh38
rs982287815 Chr.6: 13787267-13787267 on Build GRCh38
rs191397603 Chr.6: 13787678-13787678 on Build GRCh38
rs980495634 Chr.6: 13787305-13787305 on Build GRCh38
rs897728655 Chr.6: 13787454-13787454 on Build GRCh38
rs958317269 Chr.6: 13787544-13787544 on Build GRCh38
rs554720846 Chr.6: 13787447-13787447 on Build GRCh38
rs146211351 Chr.6: 13787361-13787361 on Build GRCh38
rs531990602 Chr.6: 13787606-13787606 on Build GRCh38
rs779980721 Chr.6: 13787340-13787340 on Build GRCh38
rs931968765 Chr.6: 13787386-13787386 on Build GRCh38
rs761190746 Chr.6: 13787658-13787658 on Build GRCh38
rs3734672 Chr.6: 13787346-13787346 on Build GRCh38
rs932089240 Chr.6: 13787685-13787685 on Build GRCh38
rs539904587 Chr.6: 13787440-13787440 on Build GRCh38
rs1052581722 Chr.6: 13787511-13787511 on Build GRCh38
rs1021652525 Chr.6: 13787575-13787575 on Build GRCh38
rs918894203 Chr.6: 13787642-13787642 on Build GRCh38
rs969445000 Chr.6: 13787297-13787297 on Build GRCh38
rs1007345954 Chr.6: 13787675-13787675 on Build GRCh38

View Full Product Details