Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13787839-13788384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCUR1
Entrez Gene ID
63933
Gene Name
mitochondrial calcium uniporter regulator 1
Gene Aliases
C6orf79, CCDC90A, FMP32
UniGene
Hs.214043
Gene Chromosome Location
Chr.6: 13786549-13814994 on Build GRCh38
Transcripts
NM_001031713.3 XM_011514802.1

SNP Detail

SNP ID Chromosome Location
rs529766945 Chr.6: 13788158-13788158 on Build GRCh38
rs915086752 Chr.6: 13788130-13788130 on Build GRCh38
rs1012482213 Chr.6: 13787958-13787958 on Build GRCh38
rs144525307 Chr.6: 13788040-13788040 on Build GRCh38
rs528022141 Chr.6: 13788178-13788178 on Build GRCh38
rs1017115986 Chr.6: 13788001-13788001 on Build GRCh38
rs570007774 Chr.6: 13788355-13788355 on Build GRCh38
rs888228769 Chr.6: 13788018-13788018 on Build GRCh38
rs973181342 Chr.6: 13788007-13788007 on Build GRCh38
rs530171402 Chr.6: 13788269-13788269 on Build GRCh38
rs936647279 Chr.6: 13787902-13787902 on Build GRCh38
rs1026090441 Chr.6: 13788218-13788218 on Build GRCh38
rs953563120 Chr.6: 13787864-13787864 on Build GRCh38
rs548008194 Chr.6: 13788180-13788180 on Build GRCh38
rs1204302 Chr.6: 13788175-13788175 on Build GRCh38
rs974690638 Chr.6: 13788314-13788314 on Build GRCh38
rs188939314 Chr.6: 13788069-13788069 on Build GRCh38
rs758487694 Chr.6: 13787933-13787933 on Build GRCh38
rs567806280 Chr.6: 13788211-13788211 on Build GRCh38
rs3207315 Chr.6: 13788094-13788094 on Build GRCh38
rs893604802 Chr.6: 13787915-13787915 on Build GRCh38
rs978312709 Chr.6: 13788350-13788350 on Build GRCh38
rs372499626 Chr.6: 13788337-13788337 on Build GRCh38
rs911800872 Chr.6: 13787897-13787897 on Build GRCh38
rs975034144 Chr.6: 13788089-13788089 on Build GRCh38
rs550013603 Chr.6: 13788341-13788341 on Build GRCh38
rs1038822474 Chr.6: 13787906-13787906 on Build GRCh38
rs1003540645 Chr.6: 13787932-13787932 on Build GRCh38
rs773897160 Chr.6: 13787925-13787925 on Build GRCh38
rs964312123 Chr.6: 13788292-13788292 on Build GRCh38
rs545683095 Chr.6: 13788173-13788173 on Build GRCh38
rs1035301638 Chr.6: 13787934-13787934 on Build GRCh38
rs879408982 Chr.6: 13787885-13787886 on Build GRCh38
rs963171743 Chr.6: 13788002-13788002 on Build GRCh38
rs938203193 Chr.6: 13787994-13787994 on Build GRCh38
rs987156161 Chr.6: 13788257-13788257 on Build GRCh38
rs943233253 Chr.6: 13787900-13787900 on Build GRCh38
rs553728352 Chr.6: 13788036-13788036 on Build GRCh38
rs950451394 Chr.6: 13788247-13788247 on Build GRCh38
rs750477868 Chr.6: 13787916-13787916 on Build GRCh38
rs994738622 Chr.6: 13788217-13788217 on Build GRCh38
rs970947184 Chr.6: 13788206-13788206 on Build GRCh38
rs943305392 Chr.6: 13788080-13788080 on Build GRCh38
rs938187251 Chr.6: 13788243-13788243 on Build GRCh38
rs1042831413 Chr.6: 13787917-13787917 on Build GRCh38
rs3757254 Chr.6: 13788020-13788020 on Build GRCh38
rs557655142 Chr.6: 13787920-13787920 on Build GRCh38
rs1028681041 Chr.6: 13788131-13788131 on Build GRCh38
rs953889637 Chr.6: 13788136-13788136 on Build GRCh38
rs1017566025 Chr.6: 13788210-13788210 on Build GRCh38
rs563823481 Chr.6: 13788202-13788202 on Build GRCh38
rs147811464 Chr.6: 13788151-13788151 on Build GRCh38
rs577548330 Chr.6: 13787951-13787951 on Build GRCh38
rs575177324 Chr.6: 13787892-13787892 on Build GRCh38
rs761486172 Chr.6: 13787999-13787999 on Build GRCh38
rs926831328 Chr.6: 13787973-13787973 on Build GRCh38
rs987082594 Chr.6: 13788049-13788049 on Build GRCh38

View Full Product Details