Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 15533089-15533578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTNBP1
Entrez Gene ID
84062
Gene Name
dystrobrevin binding protein 1
Gene Aliases
BLOC1S8, DBND, HPS7, My031, SDY
UniGene
Hs.571148
Gene Chromosome Location
Chr.6: 15522801-15663058 on Build GRCh38
Transcripts
NM_001271667.1 NM_001271668.1 NM_001271669.1 NM_032122.4 NM_183040.2 NR_036448.1 XM_005249447.4 XM_011514936.2 XM_011514937.2 XM_017011348.1 XM_017011349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1441805 c.664C>T p.R222* Chr.6: 15533243-15533243 on Build GRCh38
3920955 c.561C>G p.T187T Chr.6: 15533346-15533346 on Build GRCh38
6650041 c.528G>A p.E176E Chr.6: 15533379-15533379 on Build GRCh38
5746523 c.634C>A p.L212M Chr.6: 15533276-15533276 on Build GRCh38
4443538 c.583C>T p.R195W Chr.6: 15533324-15533324 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs111308544 Chr.6: 15533546-15533546 on Build GRCh38
rs755709396 Chr.6: 15533246-15533246 on Build GRCh38
rs773151898 Chr.6: 15533284-15533284 on Build GRCh38
rs761227062 Chr.6: 15533400-15533400 on Build GRCh38
rs764601456 Chr.6: 15533427-15533427 on Build GRCh38
rs767576216 Chr.6: 15533231-15533231 on Build GRCh38
rs202135348 Chr.6: 15533202-15533202 on Build GRCh38
rs529662568 Chr.6: 15533179-15533179 on Build GRCh38
rs532321679 Chr.6: 15533194-15533194 on Build GRCh38
rs757905508 Chr.6: 15533446-15533446 on Build GRCh38
rs560914300 Chr.6: 15533177-15533177 on Build GRCh38
rs764648832 Chr.6: 15533344-15533344 on Build GRCh38
rs534257582 Chr.6: 15533233-15533233 on Build GRCh38
rs923034063 Chr.6: 15533129-15533130 on Build GRCh38
rs866664336 Chr.6: 15533180-15533180 on Build GRCh38
rs75380691 Chr.6: 15533192-15533192 on Build GRCh38
rs984335749 Chr.6: 15533114-15533114 on Build GRCh38
rs374563522 Chr.6: 15533418-15533418 on Build GRCh38
rs192166343 Chr.6: 15533185-15533185 on Build GRCh38
rs774323219 Chr.6: 15533220-15533220 on Build GRCh38
rs138074958 Chr.6: 15533352-15533352 on Build GRCh38
rs367543104 Chr.6: 15533253-15533253 on Build GRCh38
rs999303527 Chr.6: 15533533-15533533 on Build GRCh38
rs1043551751 Chr.6: 15533548-15533548 on Build GRCh38
rs998441607 Chr.6: 15533503-15533503 on Build GRCh38
rs758823248 Chr.6: 15533347-15533348 on Build GRCh38
rs903750438 Chr.6: 15533302-15533302 on Build GRCh38
rs183791745 Chr.6: 15533119-15533119 on Build GRCh38
rs964548288 Chr.6: 15533120-15533120 on Build GRCh38
rs944761352 Chr.6: 15533501-15533501 on Build GRCh38
rs556813776 Chr.6: 15533351-15533351 on Build GRCh38
rs983696355 Chr.6: 15533497-15533497 on Build GRCh38
rs769506243 Chr.6: 15533208-15533208 on Build GRCh38
rs751238048 Chr.6: 15533323-15533324 on Build GRCh38
rs753238222 Chr.6: 15533242-15533242 on Build GRCh38
rs759738848 Chr.6: 15533324-15533324 on Build GRCh38
rs775297230 Chr.6: 15533214-15533214 on Build GRCh38
rs776893674 Chr.6: 15533323-15533323 on Build GRCh38
rs999153661 Chr.6: 15533178-15533178 on Build GRCh38
rs199687646 Chr.6: 15533439-15533439 on Build GRCh38
rs760702919 Chr.6: 15533286-15533286 on Build GRCh38
rs765629999 Chr.6: 15533332-15533332 on Build GRCh38
rs771935509 Chr.6: 15533281-15533281 on Build GRCh38
rs139290329 Chr.6: 15533125-15533125 on Build GRCh38
rs747886064 Chr.6: 15533373-15533373 on Build GRCh38
rs759665568 Chr.6: 15533405-15533405 on Build GRCh38
rs367702294 Chr.6: 15533243-15533243 on Build GRCh38
rs991953810 Chr.6: 15533186-15533186 on Build GRCh38
rs748965197 Chr.6: 15533354-15533354 on Build GRCh38
rs373188215 Chr.6: 15533193-15533193 on Build GRCh38
rs186662424 Chr.6: 15533139-15533139 on Build GRCh38
rs147011671 Chr.6: 15533254-15533254 on Build GRCh38
rs750339551 Chr.6: 15533500-15533500 on Build GRCh38
rs768375302 Chr.6: 15533216-15533216 on Build GRCh38
rs868661401 Chr.6: 15533298-15533298 on Build GRCh38
rs765320024 Chr.6: 15533409-15533409 on Build GRCh38
rs200731587 Chr.6: 15533386-15533386 on Build GRCh38
rs1030661643 Chr.6: 15533346-15533346 on Build GRCh38
rs952268286 Chr.6: 15533469-15533469 on Build GRCh38
rs370724534 Chr.6: 15533235-15533235 on Build GRCh38
rs755341865 Chr.6: 15533237-15533237 on Build GRCh38
rs933048754 Chr.6: 15533133-15533133 on Build GRCh38
rs755318275 Chr.6: 15533200-15533200 on Build GRCh38
rs766488666 Chr.6: 15533314-15533314 on Build GRCh38
rs969552575 Chr.6: 15533435-15533435 on Build GRCh38
rs11753919 Chr.6: 15533502-15533502 on Build GRCh38
rs1030767949 Chr.6: 15533540-15533540 on Build GRCh38
rs16876589 Chr.6: 15533266-15533266 on Build GRCh38
rs112527545 Chr.6: 15533411-15533411 on Build GRCh38
rs142211938 Chr.6: 15533361-15533361 on Build GRCh38
rs189450651 Chr.6: 15533526-15533526 on Build GRCh38
rs184939400 Chr.6: 15533229-15533229 on Build GRCh38
rs75704383 Chr.6: 15533375-15533375 on Build GRCh38
rs540744446 Chr.6: 15533171-15533171 on Build GRCh38
rs1005788664 Chr.6: 15533207-15533207 on Build GRCh38
rs536290179 Chr.6: 15533339-15533339 on Build GRCh38
rs989148427 Chr.6: 15533555-15533555 on Build GRCh38
rs201229929 Chr.6: 15533201-15533201 on Build GRCh38
rs754516188 Chr.6: 15533364-15533364 on Build GRCh38
rs76349207 Chr.6: 15533245-15533245 on Build GRCh38
rs752816673 Chr.6: 15533197-15533219 on Build GRCh38
rs907893777 Chr.6: 15533212-15533212 on Build GRCh38
rs747937400 Chr.6: 15533277-15533277 on Build GRCh38
rs771033313 Chr.6: 15533376-15533376 on Build GRCh38
rs778637428 Chr.6: 15533367-15533367 on Build GRCh38
rs761919569 Chr.6: 15533191-15533191 on Build GRCh38
rs745511215 Chr.6: 15533205-15533205 on Build GRCh38
rs752939004 Chr.6: 15533334-15533334 on Build GRCh38
rs932078499 Chr.6: 15533532-15533532 on Build GRCh38
rs1005528477 Chr.6: 15533491-15533491 on Build GRCh38
rs61739410 Chr.6: 15533238-15533238 on Build GRCh38
rs752149784 Chr.6: 15533349-15533349 on Build GRCh38
rs572983818 Chr.6: 15533159-15533159 on Build GRCh38
rs1046667159 Chr.6: 15533390-15533390 on Build GRCh38
rs996476048 Chr.6: 15533134-15533134 on Build GRCh38

View Full Product Details