Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16238474-16238969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPR
Entrez Gene ID
2766
Gene Name
guanosine monophosphate reductase
Gene Aliases
GMPR 1, GMPR1
UniGene
Hs.484741
Gene Chromosome Location
Chr.6: 16238580-16295549 on Build GRCh38
Transcripts
NM_006877.3 XM_011514508.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6343871 c.87+6G>T p.? Chr.6: 16238786-16238786 on Build GRCh38
7238689 c.87+3G>T p.? Chr.6: 16238783-16238783 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933387602 Chr.6: 16238770-16238770 on Build GRCh38
rs1022694328 Chr.6: 16238525-16238525 on Build GRCh38
rs948177199 Chr.6: 16238922-16238922 on Build GRCh38
rs549342881 Chr.6: 16238619-16238619 on Build GRCh38
rs775639429 Chr.6: 16238815-16238815 on Build GRCh38
rs1054235122 Chr.6: 16238677-16238677 on Build GRCh38
rs565767956 Chr.6: 16238690-16238690 on Build GRCh38
rs11538836 Chr.6: 16238601-16238601 on Build GRCh38
rs984972264 Chr.6: 16238618-16238618 on Build GRCh38
rs1032772231 Chr.6: 16238497-16238497 on Build GRCh38
rs771842768 Chr.6: 16238723-16238723 on Build GRCh38
rs574231468 Chr.6: 16238733-16238733 on Build GRCh38
rs755887221 Chr.6: 16238662-16238663 on Build GRCh38
rs906998969 Chr.6: 16238647-16238647 on Build GRCh38
rs768736207 Chr.6: 16238814-16238815 on Build GRCh38
rs763336945 Chr.6: 16238672-16238672 on Build GRCh38
rs548550304 Chr.6: 16238679-16238679 on Build GRCh38
rs569282829 Chr.6: 16238625-16238625 on Build GRCh38
rs917990034 Chr.6: 16238916-16238916 on Build GRCh38
rs1025958812 Chr.6: 16238705-16238705 on Build GRCh38
rs10046457 Chr.6: 16238506-16238506 on Build GRCh38
rs377446870 Chr.6: 16238717-16238717 on Build GRCh38
rs558324868 Chr.6: 16238660-16238660 on Build GRCh38
rs527448934 Chr.6: 16238513-16238513 on Build GRCh38
rs556914341 Chr.6: 16238782-16238782 on Build GRCh38
rs1002604317 Chr.6: 16238911-16238911 on Build GRCh38
rs933356190 Chr.6: 16238643-16238643 on Build GRCh38
rs11538834 Chr.6: 16238673-16238673 on Build GRCh38
rs950904158 Chr.6: 16238609-16238609 on Build GRCh38
rs998699550 Chr.6: 16238670-16238670 on Build GRCh38
rs893044901 Chr.6: 16238864-16238864 on Build GRCh38
rs747036222 Chr.6: 16238737-16238737 on Build GRCh38
rs965281627 Chr.6: 16238738-16238738 on Build GRCh38
rs764531027 Chr.6: 16238688-16238688 on Build GRCh38
rs536919884 Chr.6: 16238754-16238754 on Build GRCh38
rs776740248 Chr.6: 16238748-16238748 on Build GRCh38
rs971497397 Chr.6: 16238567-16238567 on Build GRCh38
rs1056496884 Chr.6: 16238907-16238907 on Build GRCh38
rs545424770 Chr.6: 16238894-16238894 on Build GRCh38
rs935310240 Chr.6: 16238853-16238853 on Build GRCh38
rs923939817 Chr.6: 16238837-16238837 on Build GRCh38
rs919546957 Chr.6: 16238586-16238586 on Build GRCh38
rs554383030 Chr.6: 16238710-16238710 on Build GRCh38
rs903115718 Chr.6: 16238655-16238656 on Build GRCh38
rs576467698 Chr.6: 16238833-16238833 on Build GRCh38
rs974936327 Chr.6: 16238744-16238744 on Build GRCh38
rs1042353950 Chr.6: 16238648-16238649 on Build GRCh38
rs923354191 Chr.6: 16238749-16238749 on Build GRCh38
rs11538837 Chr.6: 16238604-16238604 on Build GRCh38
rs770762636 Chr.6: 16238745-16238745 on Build GRCh38
rs538263192 Chr.6: 16238655-16238655 on Build GRCh38
rs676 Chr.6: 16238735-16238735 on Build GRCh38
rs547617845 Chr.6: 16238566-16238566 on Build GRCh38
rs779775612 Chr.6: 16238698-16238698 on Build GRCh38
rs943501389 Chr.6: 16238629-16238629 on Build GRCh38
rs529970027 Chr.6: 16238613-16238613 on Build GRCh38
rs923372740 Chr.6: 16238640-16238640 on Build GRCh38
rs376410884 Chr.6: 16238792-16238792 on Build GRCh38
rs770034068 Chr.6: 16238798-16238798 on Build GRCh38
rs534125759 Chr.6: 16238694-16238694 on Build GRCh38
rs981120959 Chr.6: 16238577-16238577 on Build GRCh38
rs753381835 Chr.6: 16238700-16238700 on Build GRCh38
rs886137026 Chr.6: 16238928-16238928 on Build GRCh38
rs754720859 Chr.6: 16238707-16238707 on Build GRCh38
rs567440012 Chr.6: 16238582-16238582 on Build GRCh38
rs566959983 Chr.6: 16238919-16238919 on Build GRCh38
rs892996250 Chr.6: 16238693-16238693 on Build GRCh38
rs749182206 Chr.6: 16238815-16238815 on Build GRCh38
rs565232840 Chr.6: 16238937-16238937 on Build GRCh38
rs779885429 Chr.6: 16238750-16238751 on Build GRCh38
rs778672609 Chr.6: 16238711-16238711 on Build GRCh38
rs1023168427 Chr.6: 16238696-16238696 on Build GRCh38
rs758308093 Chr.6: 16238840-16238840 on Build GRCh38
rs1039559627 Chr.6: 16238638-16238638 on Build GRCh38

View Full Product Details