Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16295271-16295779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPR
Entrez Gene ID
2766
Gene Name
guanosine monophosphate reductase
Gene Aliases
GMPR 1, GMPR1
UniGene
Hs.484741
Gene Chromosome Location
Chr.6: 16238580-16295549 on Build GRCh38
Transcripts
NM_006877.3 XM_011514508.2

SNP Detail

SNP ID Chromosome Location
rs541238106 Chr.6: 16295700-16295701 on Build GRCh38
rs925430439 Chr.6: 16295371-16295371 on Build GRCh38
rs1006048858 Chr.6: 16295515-16295515 on Build GRCh38
rs1032469045 Chr.6: 16295656-16295656 on Build GRCh38
rs937060543 Chr.6: 16295456-16295456 on Build GRCh38
rs976142517 Chr.6: 16295338-16295338 on Build GRCh38
rs534640295 Chr.6: 16295666-16295666 on Build GRCh38
rs1019964250 Chr.6: 16295399-16295399 on Build GRCh38
rs901118090 Chr.6: 16295704-16295704 on Build GRCh38
rs949678975 Chr.6: 16295411-16295411 on Build GRCh38
rs968476593 Chr.6: 16295482-16295482 on Build GRCh38
rs754745765 Chr.6: 16295712-16295712 on Build GRCh38
rs750806150 Chr.6: 16295636-16295636 on Build GRCh38
rs982782255 Chr.6: 16295696-16295696 on Build GRCh38
rs368617839 Chr.6: 16295648-16295648 on Build GRCh38
rs986228149 Chr.6: 16295361-16295361 on Build GRCh38
rs1007631705 Chr.6: 16295644-16295644 on Build GRCh38
rs386697487 Chr.6: 16295324-16295326 on Build GRCh38
rs1034119765 Chr.6: 16295321-16295321 on Build GRCh38
rs940873813 Chr.6: 16295303-16295303 on Build GRCh38
rs554534340 Chr.6: 16295677-16295677 on Build GRCh38
rs563414594 Chr.6: 16295307-16295307 on Build GRCh38
rs547763519 Chr.6: 16295385-16295385 on Build GRCh38
rs1042427 Chr.6: 16295324-16295324 on Build GRCh38
rs536733227 Chr.6: 16295557-16295557 on Build GRCh38
rs180701200 Chr.6: 16295602-16295602 on Build GRCh38
rs56410915 Chr.6: 16295326-16295326 on Build GRCh38
rs6723 Chr.6: 16295358-16295358 on Build GRCh38
rs937019645 Chr.6: 16295576-16295576 on Build GRCh38
rs958186494 Chr.6: 16295658-16295658 on Build GRCh38
rs1029039417 Chr.6: 16295337-16295337 on Build GRCh38
rs549970916 Chr.6: 16295592-16295592 on Build GRCh38
rs1045366814 Chr.6: 16295412-16295412 on Build GRCh38
rs970978580 Chr.6: 16295669-16295669 on Build GRCh38
rs896039719 Chr.6: 16295300-16295300 on Build GRCh38
rs763865533 Chr.6: 16295438-16295438 on Build GRCh38
rs906919140 Chr.6: 16295296-16295296 on Build GRCh38
rs1030755255 Chr.6: 16295349-16295349 on Build GRCh38
rs528021798 Chr.6: 16295372-16295372 on Build GRCh38
rs9383150 Chr.6: 16295660-16295660 on Build GRCh38
rs937030765 Chr.6: 16295718-16295718 on Build GRCh38
rs1014816358 Chr.6: 16295330-16295330 on Build GRCh38
rs367732937 Chr.6: 16295730-16295730 on Build GRCh38
rs72840228 Chr.6: 16295480-16295480 on Build GRCh38
rs180022 Chr.6: 16295630-16295630 on Build GRCh38
rs552140825 Chr.6: 16295683-16295683 on Build GRCh38
rs751167932 Chr.6: 16295481-16295484 on Build GRCh38
rs899329777 Chr.6: 16295645-16295645 on Build GRCh38
rs1036228804 Chr.6: 16295596-16295596 on Build GRCh38
rs962348979 Chr.6: 16295333-16295333 on Build GRCh38
rs1001692102 Chr.6: 16295315-16295315 on Build GRCh38
rs887592158 Chr.6: 16295507-16295507 on Build GRCh38
rs764953084 Chr.6: 16295716-16295716 on Build GRCh38
rs914194165 Chr.6: 16295392-16295392 on Build GRCh38
rs909000060 Chr.6: 16295756-16295756 on Build GRCh38
rs953533471 Chr.6: 16295643-16295643 on Build GRCh38
rs897707100 Chr.6: 16295605-16295605 on Build GRCh38
rs926989459 Chr.6: 16295549-16295549 on Build GRCh38
rs567579733 Chr.6: 16295524-16295524 on Build GRCh38
rs774685190 Chr.6: 16295350-16295350 on Build GRCh38
rs992483168 Chr.6: 16295583-16295583 on Build GRCh38

View Full Product Details