Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16303581-16304124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
UniGene
Hs.434961
Gene Chromosome Location
Chr.6: 16299112-16761490 on Build GRCh38
Transcripts
NM_000332.3 NM_001128164.1

SNP Detail

SNP ID Chromosome Location
rs966608531 Chr.6: 16303637-16303637 on Build GRCh38
rs924147950 Chr.6: 16303970-16303970 on Build GRCh38
rs755297442 Chr.6: 16303820-16303820 on Build GRCh38
rs1006445805 Chr.6: 16303785-16303785 on Build GRCh38
rs963238012 Chr.6: 16303877-16303877 on Build GRCh38
rs964415763 Chr.6: 16303800-16303800 on Build GRCh38
rs568655259 Chr.6: 16303861-16303861 on Build GRCh38
rs371882314 Chr.6: 16303628-16303628 on Build GRCh38
rs777409809 Chr.6: 16303915-16303915 on Build GRCh38
rs1016029435 Chr.6: 16303795-16303795 on Build GRCh38
rs368346295 Chr.6: 16303617-16303617 on Build GRCh38
rs936871494 Chr.6: 16304050-16304050 on Build GRCh38
rs746721075 Chr.6: 16303927-16303927 on Build GRCh38
rs554595627 Chr.6: 16303678-16303678 on Build GRCh38
rs760959130 Chr.6: 16303628-16303629 on Build GRCh38
rs770484750 Chr.6: 16304004-16304004 on Build GRCh38
rs182264870 Chr.6: 16303730-16303730 on Build GRCh38
rs1022204851 Chr.6: 16303611-16303611 on Build GRCh38
rs72840230 Chr.6: 16304091-16304091 on Build GRCh38
rs930231919 Chr.6: 16303990-16303990 on Build GRCh38
rs574172713 Chr.6: 16303729-16303729 on Build GRCh38
rs918802805 Chr.6: 16303986-16303986 on Build GRCh38
rs911590228 Chr.6: 16304016-16304016 on Build GRCh38
rs974246056 Chr.6: 16303946-16303946 on Build GRCh38
rs576199358 Chr.6: 16304027-16304027 on Build GRCh38
rs145017547 Chr.6: 16303771-16303771 on Build GRCh38

View Full Product Details