Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 16305156-16305661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN1
Entrez Gene ID
6310
Gene Name
ataxin 1
Gene Aliases
ATX1, D6S504E, SCA1
UniGene
Hs.434961
Gene Chromosome Location
Chr.6: 16299112-16761490 on Build GRCh38
Transcripts
NM_000332.3 NM_001128164.1

SNP Detail

SNP ID Chromosome Location
rs923653691 Chr.6: 16305483-16305483 on Build GRCh38
rs1047212378 Chr.6: 16305200-16305200 on Build GRCh38
rs77916634 Chr.6: 16305275-16305275 on Build GRCh38
rs1044736451 Chr.6: 16305584-16305584 on Build GRCh38
rs865991334 Chr.6: 16305299-16305299 on Build GRCh38
rs1045030031 Chr.6: 16305524-16305524 on Build GRCh38
rs141220673 Chr.6: 16305199-16305199 on Build GRCh38
rs898520336 Chr.6: 16305196-16305196 on Build GRCh38
rs983276982 Chr.6: 16305462-16305462 on Build GRCh38
rs770505492 Chr.6: 16305516-16305516 on Build GRCh38
rs543843264 Chr.6: 16305609-16305609 on Build GRCh38
rs887168257 Chr.6: 16305594-16305594 on Build GRCh38
rs979074180 Chr.6: 16305443-16305443 on Build GRCh38
rs1003787656 Chr.6: 16305630-16305630 on Build GRCh38
rs1052632021 Chr.6: 16305555-16305557 on Build GRCh38
rs962703263 Chr.6: 16305504-16305504 on Build GRCh38
rs746465133 Chr.6: 16305440-16305440 on Build GRCh38
rs1010608284 Chr.6: 16305578-16305578 on Build GRCh38
rs745410897 Chr.6: 16305522-16305522 on Build GRCh38
rs909926643 Chr.6: 16305465-16305465 on Build GRCh38
rs893529973 Chr.6: 16305561-16305561 on Build GRCh38
rs544657091 Chr.6: 16305540-16305540 on Build GRCh38
rs56071504 Chr.6: 16305272-16305272 on Build GRCh38
rs951527666 Chr.6: 16305439-16305439 on Build GRCh38
rs562967724 Chr.6: 16305602-16305602 on Build GRCh38
rs967719453 Chr.6: 16305430-16305430 on Build GRCh38
rs1023908629 Chr.6: 16305295-16305295 on Build GRCh38
rs76672287 Chr.6: 16305274-16305274 on Build GRCh38
rs964618209 Chr.6: 16305297-16305297 on Build GRCh38
rs372773233 Chr.6: 16305274-16305275 on Build GRCh38
rs12524151 Chr.6: 16305637-16305637 on Build GRCh38
rs376832186 Chr.6: 16305514-16305514 on Build GRCh38
rs144945682 Chr.6: 16305360-16305360 on Build GRCh38
rs1035593519 Chr.6: 16305415-16305415 on Build GRCh38
rs948009558 Chr.6: 16305183-16305183 on Build GRCh38
rs947948732 Chr.6: 16305523-16305523 on Build GRCh38
rs759886723 Chr.6: 16305587-16305587 on Build GRCh38
rs905484953 Chr.6: 16305595-16305595 on Build GRCh38
rs878992657 Chr.6: 16305275-16305275 on Build GRCh38
rs1008362639 Chr.6: 16305225-16305225 on Build GRCh38
rs576336022 Chr.6: 16305275-16305276 on Build GRCh38
rs183433679 Chr.6: 16305204-16305204 on Build GRCh38
rs561583528 Chr.6: 16305378-16305378 on Build GRCh38
rs917787600 Chr.6: 16305517-16305517 on Build GRCh38
rs780150460 Chr.6: 16305547-16305547 on Build GRCh38
rs1019800329 Chr.6: 16305257-16305257 on Build GRCh38
rs1045460896 Chr.6: 16305190-16305190 on Build GRCh38
rs1035243618 Chr.6: 16305637-16305638 on Build GRCh38
rs919917933 Chr.6: 16305528-16305528 on Build GRCh38

View Full Product Details