Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 20534379-20534893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKAL1
Entrez Gene ID
54901
Gene Name
CDK5 regulatory subunit associated protein 1 like 1
Gene Aliases
-
UniGene
Hs.657604
Gene Chromosome Location
Chr.6: 20534457-21232404 on Build GRCh38
Transcripts
NM_017774.3 XM_011514718.1 XM_011514719.2 XM_017010986.1 XM_017010987.1

SNP Detail

SNP ID Chromosome Location
rs868601974 Chr.6: 20534427-20534427 on Build GRCh38
rs928988123 Chr.6: 20534554-20534554 on Build GRCh38
rs149181889 Chr.6: 20534420-20534420 on Build GRCh38
rs567820490 Chr.6: 20534488-20534488 on Build GRCh38
rs994068065 Chr.6: 20534867-20534867 on Build GRCh38
rs933497880 Chr.6: 20534851-20534851 on Build GRCh38
rs949802303 Chr.6: 20534532-20534532 on Build GRCh38
rs763917248 Chr.6: 20534508-20534508 on Build GRCh38
rs923563519 Chr.6: 20534739-20534739 on Build GRCh38
rs538764883 Chr.6: 20534546-20534546 on Build GRCh38
rs57228197 Chr.6: 20534740-20534740 on Build GRCh38
rs888000211 Chr.6: 20534569-20534569 on Build GRCh38
rs1040618444 Chr.6: 20534500-20534500 on Build GRCh38
rs918911879 Chr.6: 20534551-20534551 on Build GRCh38
rs1001249976 Chr.6: 20534662-20534662 on Build GRCh38
rs887935984 Chr.6: 20534446-20534446 on Build GRCh38
rs895195828 Chr.6: 20534828-20534828 on Build GRCh38
rs1046258689 Chr.6: 20534857-20534857 on Build GRCh38
rs11963450 Chr.6: 20534743-20534743 on Build GRCh38
rs957327280 Chr.6: 20534517-20534517 on Build GRCh38
rs41271303 Chr.6: 20534577-20534577 on Build GRCh38
rs536213002 Chr.6: 20534668-20534668 on Build GRCh38
rs1039183313 Chr.6: 20534582-20534582 on Build GRCh38
rs957482441 Chr.6: 20534682-20534682 on Build GRCh38
rs563308663 Chr.6: 20534424-20534424 on Build GRCh38
rs985633366 Chr.6: 20534660-20534660 on Build GRCh38
rs1022745627 Chr.6: 20534714-20534714 on Build GRCh38
rs895793166 Chr.6: 20534539-20534539 on Build GRCh38
rs115926208 Chr.6: 20534528-20534528 on Build GRCh38
rs971233186 Chr.6: 20534728-20534728 on Build GRCh38
rs961284618 Chr.6: 20534565-20534565 on Build GRCh38
rs577876243 Chr.6: 20534756-20534756 on Build GRCh38
rs563188326 Chr.6: 20534761-20534761 on Build GRCh38
rs548741094 Chr.6: 20534869-20534869 on Build GRCh38
rs140741461 Chr.6: 20534687-20534687 on Build GRCh38
rs568301085 Chr.6: 20534496-20534496 on Build GRCh38
rs368377774 Chr.6: 20534408-20534408 on Build GRCh38
rs991702767 Chr.6: 20534527-20534527 on Build GRCh38
rs997602138 Chr.6: 20534495-20534495 on Build GRCh38
rs184975546 Chr.6: 20534760-20534760 on Build GRCh38
rs985007727 Chr.6: 20534471-20534471 on Build GRCh38
rs901997600 Chr.6: 20534843-20534843 on Build GRCh38
rs993757767 Chr.6: 20534429-20534429 on Build GRCh38
rs116186936 Chr.6: 20534734-20534734 on Build GRCh38
rs756115717 Chr.6: 20534769-20534769 on Build GRCh38
rs1033539855 Chr.6: 20534529-20534529 on Build GRCh38
rs1008118491 Chr.6: 20534466-20534466 on Build GRCh38
rs370587873 Chr.6: 20534605-20534605 on Build GRCh38
rs902492125 Chr.6: 20534653-20534653 on Build GRCh38
rs756145264 Chr.6: 20534615-20534615 on Build GRCh38
rs762847250 Chr.6: 20534501-20534501 on Build GRCh38
rs1027054830 Chr.6: 20534636-20534636 on Build GRCh38
rs1045337927 Chr.6: 20534419-20534419 on Build GRCh38
rs1017802554 Chr.6: 20534467-20534467 on Build GRCh38
rs780176942 Chr.6: 20534779-20534779 on Build GRCh38
rs1057041443 Chr.6: 20534531-20534531 on Build GRCh38
rs984832318 Chr.6: 20534751-20534751 on Build GRCh38
rs936660755 Chr.6: 20534780-20534780 on Build GRCh38
rs530534468 Chr.6: 20534768-20534768 on Build GRCh38
rs775229872 Chr.6: 20534474-20534474 on Build GRCh38
rs545943567 Chr.6: 20534473-20534473 on Build GRCh38
rs1024872661 Chr.6: 20534454-20534454 on Build GRCh38
rs997590876 Chr.6: 20534608-20534608 on Build GRCh38
rs976182716 Chr.6: 20534735-20534735 on Build GRCh38
rs943613718 Chr.6: 20534794-20534794 on Build GRCh38
rs530395154 Chr.6: 20534433-20534433 on Build GRCh38
rs534986100 Chr.6: 20534513-20534513 on Build GRCh38
rs1042596351 Chr.6: 20534862-20534862 on Build GRCh38
rs931050241 Chr.6: 20534470-20534470 on Build GRCh38
rs549652347 Chr.6: 20534469-20534469 on Build GRCh38
rs952519142 Chr.6: 20534646-20534646 on Build GRCh38
rs1013393777 Chr.6: 20534553-20534553 on Build GRCh38
rs1049808835 Chr.6: 20534434-20534434 on Build GRCh38
rs894988854 Chr.6: 20534544-20534544 on Build GRCh38
rs1055629815 Chr.6: 20534826-20534826 on Build GRCh38
rs143352371 Chr.6: 20534555-20534555 on Build GRCh38
rs1021077231 Chr.6: 20534644-20534644 on Build GRCh38
rs966733307 Chr.6: 20534497-20534497 on Build GRCh38
rs1032356435 Chr.6: 20534663-20534663 on Build GRCh38
rs180859908 Chr.6: 20534518-20534518 on Build GRCh38
rs1039335200 Chr.6: 20534812-20534812 on Build GRCh38
rs1045491505 Chr.6: 20534542-20534542 on Build GRCh38
rs961043545 Chr.6: 20534459-20534459 on Build GRCh38

View Full Product Details