Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24702789-24703311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT13
Entrez Gene ID
55856
Gene Name
acyl-CoA thioesterase 13
Gene Aliases
HT012, PNAS-27, THEM2
UniGene
Hs.731605
Gene Chromosome Location
Chr.6: 24667035-24705069 on Build GRCh38
Transcripts
NM_001160094.1 NM_018473.3

SNP Detail

SNP ID Chromosome Location
rs903614460 Chr.6: 24702942-24702947 on Build GRCh38
rs559156470 Chr.6: 24702953-24702953 on Build GRCh38
rs142934663 Chr.6: 24702912-24702913 on Build GRCh38
rs1031097943 Chr.6: 24702959-24702959 on Build GRCh38
rs1029998868 Chr.6: 24703158-24703158 on Build GRCh38
rs971310937 Chr.6: 24702965-24702965 on Build GRCh38
rs1057052763 Chr.6: 24703052-24703052 on Build GRCh38
rs186573159 Chr.6: 24703116-24703116 on Build GRCh38
rs1056319 Chr.6: 24702818-24702818 on Build GRCh38
rs942782693 Chr.6: 24703012-24703012 on Build GRCh38
rs897798944 Chr.6: 24702844-24702844 on Build GRCh38
rs565595544 Chr.6: 24702825-24702825 on Build GRCh38
rs942123835 Chr.6: 24703039-24703039 on Build GRCh38
rs773760023 Chr.6: 24702914-24702915 on Build GRCh38
rs964019440 Chr.6: 24703183-24703183 on Build GRCh38
rs948934227 Chr.6: 24703075-24703075 on Build GRCh38
rs902826674 Chr.6: 24702855-24702855 on Build GRCh38
rs982346307 Chr.6: 24702968-24702968 on Build GRCh38
rs369791083 Chr.6: 24703183-24703186 on Build GRCh38
rs554347870 Chr.6: 24703020-24703020 on Build GRCh38
rs933838676 Chr.6: 24703031-24703031 on Build GRCh38
rs909907360 Chr.6: 24703007-24703007 on Build GRCh38
rs1036780799 Chr.6: 24702842-24702842 on Build GRCh38
rs760622659 Chr.6: 24703276-24703276 on Build GRCh38
rs573456174 Chr.6: 24702815-24702815 on Build GRCh38
rs575591311 Chr.6: 24703048-24703048 on Build GRCh38
rs150319382 Chr.6: 24703108-24703108 on Build GRCh38
rs551131408 Chr.6: 24703029-24703029 on Build GRCh38
rs938725746 Chr.6: 24703049-24703049 on Build GRCh38
rs938953046 Chr.6: 24702858-24702858 on Build GRCh38
rs984071002 Chr.6: 24703001-24703001 on Build GRCh38
rs543055316 Chr.6: 24702828-24702828 on Build GRCh38
rs192450304 Chr.6: 24702872-24702872 on Build GRCh38
rs778856218 Chr.6: 24703145-24703145 on Build GRCh38
rs774612551 Chr.6: 24703013-24703013 on Build GRCh38
rs773321186 Chr.6: 24703246-24703255 on Build GRCh38
rs892194419 Chr.6: 24703127-24703127 on Build GRCh38
rs984374964 Chr.6: 24703160-24703160 on Build GRCh38
rs537313771 Chr.6: 24703051-24703051 on Build GRCh38
rs909876121 Chr.6: 24703169-24703169 on Build GRCh38
rs374586952 Chr.6: 24702971-24702971 on Build GRCh38
rs934120528 Chr.6: 24703238-24703238 on Build GRCh38
rs1024476831 Chr.6: 24703111-24703111 on Build GRCh38
rs771865254 Chr.6: 24703196-24703196 on Build GRCh38
rs182304133 Chr.6: 24702951-24702951 on Build GRCh38
rs894771609 Chr.6: 24703071-24703071 on Build GRCh38
rs371628628 Chr.6: 24702876-24702876 on Build GRCh38
rs16889632 Chr.6: 24702826-24702826 on Build GRCh38
rs904659857 Chr.6: 24703104-24703104 on Build GRCh38
rs529025317 Chr.6: 24702944-24702944 on Build GRCh38
rs866513733 Chr.6: 24703032-24703032 on Build GRCh38
rs797015671 Chr.6: 24703180-24703183 on Build GRCh38
rs116166221 Chr.6: 24703044-24703044 on Build GRCh38
rs1032708006 Chr.6: 24702941-24702941 on Build GRCh38
rs1023797021 Chr.6: 24702964-24702964 on Build GRCh38
rs1030280412 Chr.6: 24703009-24703009 on Build GRCh38
rs760566751 Chr.6: 24703285-24703285 on Build GRCh38
rs749330291 Chr.6: 24703091-24703091 on Build GRCh38
rs552700909 Chr.6: 24702923-24702929 on Build GRCh38
rs796408116 Chr.6: 24702882-24702882 on Build GRCh38
rs550740549 Chr.6: 24703103-24703103 on Build GRCh38
rs901038196 Chr.6: 24702821-24702821 on Build GRCh38
rs1004132175 Chr.6: 24703135-24703135 on Build GRCh38
rs918851718 Chr.6: 24702994-24702994 on Build GRCh38
rs929948036 Chr.6: 24702996-24702996 on Build GRCh38
rs16889633 Chr.6: 24703214-24703214 on Build GRCh38
rs956375640 Chr.6: 24702990-24702990 on Build GRCh38
rs1021746682 Chr.6: 24702930-24702930 on Build GRCh38
rs748096554 Chr.6: 24703156-24703156 on Build GRCh38
rs553489511 Chr.6: 24703133-24703133 on Build GRCh38
rs557212655 Chr.6: 24702894-24702894 on Build GRCh38

View Full Product Details