Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 24703840-24704383 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOT13
Entrez Gene ID
55856
Gene Name
acyl-CoA thioesterase 13
Gene Aliases
HT012, PNAS-27, THEM2
UniGene
Hs.731605
Gene Chromosome Location
Chr.6: 24667035-24705069 on Build GRCh38
Transcripts
NM_001160094.1 NM_018473.3

SNP Detail

SNP ID Chromosome Location
rs745582723 Chr.6: 24704162-24704162 on Build GRCh38
rs985621022 Chr.6: 24703895-24703896 on Build GRCh38
rs943061192 Chr.6: 24703902-24703902 on Build GRCh38
rs547325290 Chr.6: 24703891-24703891 on Build GRCh38
rs544702505 Chr.6: 24703937-24703937 on Build GRCh38
rs568632923 Chr.6: 24704026-24704026 on Build GRCh38
rs898799126 Chr.6: 24703936-24703937 on Build GRCh38
rs921393685 Chr.6: 24704112-24704112 on Build GRCh38
rs1035958128 Chr.6: 24704317-24704317 on Build GRCh38
rs1018474069 Chr.6: 24704286-24704286 on Build GRCh38
rs189605917 Chr.6: 24704053-24704053 on Build GRCh38
rs760818585 Chr.6: 24704237-24704237 on Build GRCh38
rs955617317 Chr.6: 24704243-24704244 on Build GRCh38
rs569217896 Chr.6: 24704320-24704320 on Build GRCh38
rs1054978375 Chr.6: 24704221-24704221 on Build GRCh38
rs1014076213 Chr.6: 24704025-24704026 on Build GRCh38
rs566150886 Chr.6: 24703959-24703959 on Build GRCh38
rs908923039 Chr.6: 24704072-24704072 on Build GRCh38
rs998879891 Chr.6: 24703963-24703963 on Build GRCh38
rs57839214 Chr.6: 24704243-24704243 on Build GRCh38
rs891364048 Chr.6: 24704161-24704161 on Build GRCh38
rs767230609 Chr.6: 24704242-24704243 on Build GRCh38
rs547026602 Chr.6: 24704158-24704158 on Build GRCh38
rs552509436 Chr.6: 24703902-24703902 on Build GRCh38
rs932854115 Chr.6: 24704129-24704129 on Build GRCh38
rs566867244 Chr.6: 24704190-24704190 on Build GRCh38
rs1000016540 Chr.6: 24704175-24704175 on Build GRCh38
rs985629276 Chr.6: 24704091-24704091 on Build GRCh38
rs368353960 Chr.6: 24704077-24704077 on Build GRCh38
rs557405253 Chr.6: 24704124-24704124 on Build GRCh38
rs770861931 Chr.6: 24704035-24704035 on Build GRCh38
rs1053598 Chr.6: 24704310-24704310 on Build GRCh38
rs575682531 Chr.6: 24704233-24704233 on Build GRCh38
rs139572889 Chr.6: 24704026-24704026 on Build GRCh38
rs990271308 Chr.6: 24704027-24704027 on Build GRCh38
rs1022794975 Chr.6: 24704028-24704028 on Build GRCh38
rs150884401 Chr.6: 24703997-24703997 on Build GRCh38
rs532680533 Chr.6: 24704242-24704242 on Build GRCh38
rs910342877 Chr.6: 24703900-24703900 on Build GRCh38
rs943113580 Chr.6: 24704103-24704104 on Build GRCh38
rs895722893 Chr.6: 24704009-24704009 on Build GRCh38
rs201997792 Chr.6: 24704243-24704243 on Build GRCh38
rs963226680 Chr.6: 24704292-24704292 on Build GRCh38
rs905674263 Chr.6: 24704168-24704168 on Build GRCh38
rs983061589 Chr.6: 24704059-24704059 on Build GRCh38
rs911359785 Chr.6: 24704100-24704100 on Build GRCh38
rs1051756495 Chr.6: 24704130-24704130 on Build GRCh38
rs1031705387 Chr.6: 24704008-24704008 on Build GRCh38
rs891873115 Chr.6: 24704224-24704224 on Build GRCh38
rs908849873 Chr.6: 24704328-24704328 on Build GRCh38
rs538452021 Chr.6: 24704338-24704338 on Build GRCh38
rs928584000 Chr.6: 24704049-24704049 on Build GRCh38
rs891097959 Chr.6: 24703956-24703956 on Build GRCh38
rs922862410 Chr.6: 24703891-24703892 on Build GRCh38
rs746730647 Chr.6: 24704032-24704032 on Build GRCh38
rs941809987 Chr.6: 24703906-24703906 on Build GRCh38
rs577131873 Chr.6: 24704086-24704091 on Build GRCh38
rs115202710 Chr.6: 24704340-24704340 on Build GRCh38
rs529901002 Chr.6: 24704140-24704140 on Build GRCh38
rs1038501276 Chr.6: 24703915-24703915 on Build GRCh38
rs572644469 Chr.6: 24703908-24703908 on Build GRCh38
rs769315241 Chr.6: 24704197-24704197 on Build GRCh38
rs900033470 Chr.6: 24703920-24703920 on Build GRCh38
rs758008612 Chr.6: 24704085-24704086 on Build GRCh38
rs1047383249 Chr.6: 24704133-24704133 on Build GRCh38
rs1053104233 Chr.6: 24704228-24704228 on Build GRCh38
rs949982934 Chr.6: 24704354-24704354 on Build GRCh38
rs776408601 Chr.6: 24704075-24704075 on Build GRCh38
rs767872697 Chr.6: 24704265-24704265 on Build GRCh38
rs536100662 Chr.6: 24704111-24704111 on Build GRCh38
rs970268874 Chr.6: 24704298-24704298 on Build GRCh38
rs773269931 Chr.6: 24704066-24704066 on Build GRCh38
rs989446948 Chr.6: 24703876-24703876 on Build GRCh38
rs969922620 Chr.6: 24704034-24704034 on Build GRCh38
rs983367292 Chr.6: 24704321-24704321 on Build GRCh38
rs887516677 Chr.6: 24703879-24703879 on Build GRCh38
rs758216526 Chr.6: 24703948-24703948 on Build GRCh38
rs956260049 Chr.6: 24704065-24704065 on Build GRCh38
rs568744317 Chr.6: 24704343-24704343 on Build GRCh38
rs572912297 Chr.6: 24704313-24704313 on Build GRCh38
rs1048130150 Chr.6: 24703965-24703965 on Build GRCh38

View Full Product Details