Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26376653-26377151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001197246.2 NM_001197247.2 NM_001197248.2 NM_001197249.2 NM_007047.4 XM_005248827.3 XM_005248831.3 XM_005248832.3 XM_006714979.3 XM_006714980.3 XM_006714981.3 XM_006714982.3 XM_011514267.2 XM_011514268.2 XM_011514269.2 XM_011514270.2 XM_011514271.2 XM_017010211.1 XM_017010212.1 XM_017010213.1 XM_017010214.1 XM_017010215.1 XM_017010216.1 XM_017010217.1

SNP Detail

SNP ID Chromosome Location
rs538852052 Chr.6: 26376695-26376695 on Build GRCh38
rs965538370 Chr.6: 26376678-26376678 on Build GRCh38
rs987420332 Chr.6: 26376890-26376890 on Build GRCh38
rs1026559186 Chr.6: 26376764-26376764 on Build GRCh38
rs748350204 Chr.6: 26376859-26376859 on Build GRCh38
rs926080805 Chr.6: 26376834-26376834 on Build GRCh38
rs943052136 Chr.6: 26376931-26376931 on Build GRCh38
rs754160156 Chr.6: 26376720-26376720 on Build GRCh38
rs1033045661 Chr.6: 26377004-26377004 on Build GRCh38
rs746814366 Chr.6: 26376809-26376809 on Build GRCh38
rs551291128 Chr.6: 26376727-26376727 on Build GRCh38
rs111568057 Chr.6: 26376841-26376841 on Build GRCh38
rs144256099 Chr.6: 26377085-26377085 on Build GRCh38
rs149826859 Chr.6: 26376952-26376952 on Build GRCh38
rs1024566208 Chr.6: 26376973-26376973 on Build GRCh38
rs749166240 Chr.6: 26377117-26377117 on Build GRCh38
rs939217832 Chr.6: 26376757-26376757 on Build GRCh38
rs953802096 Chr.6: 26376696-26376696 on Build GRCh38
rs560612660 Chr.6: 26376944-26376944 on Build GRCh38
rs573540670 Chr.6: 26376770-26376770 on Build GRCh38
rs988874702 Chr.6: 26377045-26377045 on Build GRCh38
rs554597576 Chr.6: 26377014-26377014 on Build GRCh38
rs984252834 Chr.6: 26376700-26376700 on Build GRCh38
rs573381973 Chr.6: 26376816-26376816 on Build GRCh38
rs9366654 Chr.6: 26376800-26376800 on Build GRCh38
rs974434222 Chr.6: 26377104-26377104 on Build GRCh38
rs927880142 Chr.6: 26376755-26376755 on Build GRCh38
rs559200552 Chr.6: 26377103-26377103 on Build GRCh38
rs139636053 Chr.6: 26376988-26376988 on Build GRCh38
rs567196970 Chr.6: 26377018-26377018 on Build GRCh38
rs536918568 Chr.6: 26376739-26376739 on Build GRCh38
rs943008753 Chr.6: 26377089-26377089 on Build GRCh38
rs544875091 Chr.6: 26376870-26376870 on Build GRCh38
rs1028508504 Chr.6: 26376889-26376889 on Build GRCh38
rs189748143 Chr.6: 26377067-26377067 on Build GRCh38
rs1053089637 Chr.6: 26377084-26377084 on Build GRCh38
rs145787608 Chr.6: 26376979-26376979 on Build GRCh38
rs957476873 Chr.6: 26377025-26377025 on Build GRCh38
rs201823546 Chr.6: 26376884-26376884 on Build GRCh38
rs930785031 Chr.6: 26376945-26376945 on Build GRCh38
rs987755544 Chr.6: 26377079-26377079 on Build GRCh38
rs144182156 Chr.6: 26376733-26376733 on Build GRCh38
rs920322934 Chr.6: 26377115-26377115 on Build GRCh38
rs182593871 Chr.6: 26376745-26376745 on Build GRCh38
rs534814181 Chr.6: 26376726-26376726 on Build GRCh38
rs1014889098 Chr.6: 26376967-26376967 on Build GRCh38
rs995043033 Chr.6: 26376763-26376763 on Build GRCh38
rs1021179097 Chr.6: 26377048-26377048 on Build GRCh38
rs184882276 Chr.6: 26376895-26376895 on Build GRCh38
rs565443220 Chr.6: 26376842-26376842 on Build GRCh38
rs909594899 Chr.6: 26376966-26376966 on Build GRCh38
rs957860566 Chr.6: 26376835-26376835 on Build GRCh38
rs1001526572 Chr.6: 26376995-26376995 on Build GRCh38
rs536220251 Chr.6: 26377019-26377019 on Build GRCh38
rs886627205 Chr.6: 26376965-26376965 on Build GRCh38
rs536406747 Chr.6: 26376749-26376749 on Build GRCh38
rs945263974 Chr.6: 26376882-26376882 on Build GRCh38
rs750902615 Chr.6: 26376802-26376802 on Build GRCh38
rs775902280 Chr.6: 26376677-26376677 on Build GRCh38
rs535955889 Chr.6: 26377009-26377009 on Build GRCh38
rs998784197 Chr.6: 26376875-26376875 on Build GRCh38
rs111353898 Chr.6: 26376706-26376706 on Build GRCh38
rs373770692 Chr.6: 26376868-26376868 on Build GRCh38
rs561138666 Chr.6: 26376871-26376871 on Build GRCh38
rs536004008 Chr.6: 26376790-26376790 on Build GRCh38
rs547033648 Chr.6: 26376691-26376691 on Build GRCh38
rs946509159 Chr.6: 26377035-26377035 on Build GRCh38
rs1051891246 Chr.6: 26376715-26376715 on Build GRCh38
rs954325393 Chr.6: 26376935-26376935 on Build GRCh38
rs552823491 Chr.6: 26377039-26377039 on Build GRCh38
rs1043134578 Chr.6: 26376814-26376814 on Build GRCh38
rs575868397 Chr.6: 26377123-26377123 on Build GRCh38
rs546826656 Chr.6: 26376941-26376941 on Build GRCh38
rs886570916 Chr.6: 26377124-26377124 on Build GRCh38
rs976945455 Chr.6: 26376686-26376686 on Build GRCh38

View Full Product Details