Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26405826-26406322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
UniGene
Hs.191510
Gene Chromosome Location
Chr.6: 26402237-26415216 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5979145 c.136G>A p.E46K Chr.6: 26405959-26405959 on Build GRCh38
5912380 c.250G>A p.E84K Chr.6: 26406073-26406073 on Build GRCh38
5996731 c.402T>C p.Y134Y Chr.6: 26406225-26406225 on Build GRCh38
1076782 c.346G>A p.V116I Chr.6: 26406169-26406169 on Build GRCh38
5979144 c.136G>A p.E46K Chr.6: 26405959-26405959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763829130 Chr.6: 26406067-26406067 on Build GRCh38
rs755894627 Chr.6: 26406098-26406098 on Build GRCh38
rs893261617 Chr.6: 26406266-26406266 on Build GRCh38
rs769977541 Chr.6: 26405885-26405885 on Build GRCh38
rs755260389 Chr.6: 26406104-26406104 on Build GRCh38
rs200982319 Chr.6: 26405964-26405964 on Build GRCh38
rs1026694815 Chr.6: 26405882-26405882 on Build GRCh38
rs749420100 Chr.6: 26405893-26405893 on Build GRCh38
rs779020584 Chr.6: 26406014-26406014 on Build GRCh38
rs753615560 Chr.6: 26406101-26406101 on Build GRCh38
rs150905778 Chr.6: 26406092-26406092 on Build GRCh38
rs367894457 Chr.6: 26406054-26406054 on Build GRCh38
rs992506375 Chr.6: 26405853-26405853 on Build GRCh38
rs1042136313 Chr.6: 26406291-26406291 on Build GRCh38
rs549341227 Chr.6: 26406097-26406097 on Build GRCh38
rs2393650 Chr.6: 26406168-26406168 on Build GRCh38
rs762580249 Chr.6: 26405937-26405937 on Build GRCh38
rs138258145 Chr.6: 26406099-26406099 on Build GRCh38
rs527607062 Chr.6: 26405903-26405903 on Build GRCh38
rs969820507 Chr.6: 26406115-26406115 on Build GRCh38
rs773515134 Chr.6: 26405914-26405914 on Build GRCh38
rs994794725 Chr.6: 26406069-26406069 on Build GRCh38
rs755062420 Chr.6: 26406011-26406011 on Build GRCh38
rs199781860 Chr.6: 26406055-26406055 on Build GRCh38
rs564000550 Chr.6: 26405959-26405959 on Build GRCh38
rs936292956 Chr.6: 26405943-26405943 on Build GRCh38
rs772492852 Chr.6: 26406018-26406018 on Build GRCh38
rs778170905 Chr.6: 26406133-26406133 on Build GRCh38
rs372233454 Chr.6: 26405913-26405913 on Build GRCh38
rs369084587 Chr.6: 26405967-26405967 on Build GRCh38
rs540919899 Chr.6: 26405904-26405904 on Build GRCh38
rs139011114 Chr.6: 26405975-26405975 on Build GRCh38
rs527834024 Chr.6: 26406286-26406286 on Build GRCh38
rs750226632 Chr.6: 26405966-26405966 on Build GRCh38
rs371237088 Chr.6: 26405895-26405895 on Build GRCh38
rs926171888 Chr.6: 26406124-26406124 on Build GRCh38
rs763634366 Chr.6: 26405950-26405950 on Build GRCh38
rs111540572 Chr.6: 26406196-26406196 on Build GRCh38
rs749473171 Chr.6: 26405991-26405991 on Build GRCh38
rs766298182 Chr.6: 26406295-26406295 on Build GRCh38
rs768674856 Chr.6: 26405902-26405902 on Build GRCh38
rs761066899 Chr.6: 26405918-26405918 on Build GRCh38
rs777223423 Chr.6: 26405908-26405909 on Build GRCh38
rs757956747 Chr.6: 26405858-26405858 on Build GRCh38
rs751505681 Chr.6: 26405869-26405869 on Build GRCh38
rs149895116 Chr.6: 26406020-26406020 on Build GRCh38
rs376221996 Chr.6: 26406157-26406157 on Build GRCh38
rs779075658 Chr.6: 26406112-26406112 on Build GRCh38
rs764843210 Chr.6: 26405923-26405923 on Build GRCh38
rs201217877 Chr.6: 26406134-26406134 on Build GRCh38
rs998206119 Chr.6: 26405989-26405989 on Build GRCh38
rs758452573 Chr.6: 26406117-26406117 on Build GRCh38
rs768468170 Chr.6: 26406160-26406160 on Build GRCh38
rs377117570 Chr.6: 26405965-26405965 on Build GRCh38
rs757082273 Chr.6: 26405873-26405873 on Build GRCh38
rs771438333 Chr.6: 26406138-26406138 on Build GRCh38
rs780278847 Chr.6: 26405990-26405990 on Build GRCh38
rs771284361 Chr.6: 26406045-26406045 on Build GRCh38
rs368555498 Chr.6: 26405861-26405861 on Build GRCh38
rs780796693 Chr.6: 26405877-26405877 on Build GRCh38
rs773029993 Chr.6: 26406224-26406224 on Build GRCh38
rs780292357 Chr.6: 26405891-26405891 on Build GRCh38
rs752346032 Chr.6: 26405934-26405934 on Build GRCh38
rs746233858 Chr.6: 26406158-26406158 on Build GRCh38
rs557288785 Chr.6: 26406240-26406240 on Build GRCh38
rs748253847 Chr.6: 26406016-26406016 on Build GRCh38
rs748221919 Chr.6: 26406116-26406116 on Build GRCh38
rs774075940 Chr.6: 26406081-26406081 on Build GRCh38
rs537072716 Chr.6: 26406225-26406225 on Build GRCh38
rs767404028 Chr.6: 26406094-26406094 on Build GRCh38
rs950454895 Chr.6: 26405848-26405848 on Build GRCh38
rs3857550 Chr.6: 26406027-26406027 on Build GRCh38
rs1026663094 Chr.6: 26406076-26406076 on Build GRCh38
rs184245591 Chr.6: 26405845-26405845 on Build GRCh38

View Full Product Details