Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26407629-26408126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
UniGene
Hs.191510
Gene Chromosome Location
Chr.6: 26402237-26415216 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6173405 c.645G>T p.G215G Chr.6: 26407882-26407882 on Build GRCh38
396191 c.688G>T p.E230* Chr.6: 26407925-26407925 on Build GRCh38
1076784 c.643G>A p.G215R Chr.6: 26407880-26407880 on Build GRCh38
1154634 c.705T>C p.I235I Chr.6: 26407942-26407942 on Build GRCh38
5595691 c.451C>T p.H151Y Chr.6: 26407688-26407688 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200236658 Chr.6: 26407828-26407828 on Build GRCh38
rs764220613 Chr.6: 26407758-26407758 on Build GRCh38
rs756553049 Chr.6: 26407820-26407820 on Build GRCh38
rs530314249 Chr.6: 26408084-26408084 on Build GRCh38
rs1027618310 Chr.6: 26408012-26408012 on Build GRCh38
rs777508118 Chr.6: 26407940-26407940 on Build GRCh38
rs771608988 Chr.6: 26407694-26407694 on Build GRCh38
rs779021158 Chr.6: 26407946-26407946 on Build GRCh38
rs770236343 Chr.6: 26407673-26407673 on Build GRCh38
rs376863801 Chr.6: 26407722-26407722 on Build GRCh38
rs756262847 Chr.6: 26407738-26407738 on Build GRCh38
rs781770259 Chr.6: 26407801-26407801 on Build GRCh38
rs775425163 Chr.6: 26407889-26407889 on Build GRCh38
rs533940581 Chr.6: 26407754-26407754 on Build GRCh38
rs571412787 Chr.6: 26407736-26407736 on Build GRCh38
rs749133967 Chr.6: 26407957-26407957 on Build GRCh38
rs958742727 Chr.6: 26408095-26408095 on Build GRCh38
rs1057932 Chr.6: 26407845-26407845 on Build GRCh38
rs144114619 Chr.6: 26407917-26407917 on Build GRCh38
rs180776414 Chr.6: 26407659-26407659 on Build GRCh38
rs759408823 Chr.6: 26407869-26407869 on Build GRCh38
rs746594545 Chr.6: 26407942-26407942 on Build GRCh38
rs567816736 Chr.6: 26407652-26407652 on Build GRCh38
rs909854506 Chr.6: 26407778-26407778 on Build GRCh38
rs999174057 Chr.6: 26407893-26407893 on Build GRCh38
rs774720485 Chr.6: 26407963-26407963 on Build GRCh38
rs1057933 Chr.6: 26407908-26407908 on Build GRCh38
rs996134212 Chr.6: 26407683-26407683 on Build GRCh38
rs754276670 Chr.6: 26407919-26407919 on Build GRCh38
rs141319367 Chr.6: 26407949-26407949 on Build GRCh38
rs746284813 Chr.6: 26407672-26407672 on Build GRCh38
rs769608272 Chr.6: 26407878-26407878 on Build GRCh38
rs776552214 Chr.6: 26407859-26407859 on Build GRCh38
rs970771079 Chr.6: 26408048-26408048 on Build GRCh38
rs773463629 Chr.6: 26407991-26407991 on Build GRCh38
rs776498634 Chr.6: 26407728-26407728 on Build GRCh38
rs373188059 Chr.6: 26407714-26407714 on Build GRCh38
rs369843054 Chr.6: 26407788-26407788 on Build GRCh38
rs900071015 Chr.6: 26407682-26407682 on Build GRCh38
rs774667179 Chr.6: 26407901-26407901 on Build GRCh38
rs780410774 Chr.6: 26407821-26407821 on Build GRCh38
rs981233742 Chr.6: 26408081-26408081 on Build GRCh38
rs769562626 Chr.6: 26407933-26407933 on Build GRCh38
rs887396739 Chr.6: 26408017-26408017 on Build GRCh38
rs770468308 Chr.6: 26407819-26407819 on Build GRCh38
rs746048084 Chr.6: 26408034-26408034 on Build GRCh38
rs149149636 Chr.6: 26407918-26407918 on Build GRCh38
rs760146651 Chr.6: 26407717-26407717 on Build GRCh38
rs762628652 Chr.6: 26407896-26407896 on Build GRCh38
rs1018897056 Chr.6: 26407666-26407666 on Build GRCh38
rs769813059 Chr.6: 26407952-26407952 on Build GRCh38
rs1016991633 Chr.6: 26407966-26407966 on Build GRCh38
rs117508907 Chr.6: 26407691-26407691 on Build GRCh38
rs531138964 Chr.6: 26407988-26407988 on Build GRCh38
rs745642915 Chr.6: 26407829-26407830 on Build GRCh38
rs775272201 Chr.6: 26407954-26407954 on Build GRCh38
rs1025008914 Chr.6: 26407753-26407753 on Build GRCh38
rs77196683 Chr.6: 26407654-26407654 on Build GRCh38
rs151227699 Chr.6: 26407829-26407829 on Build GRCh38
rs561643430 Chr.6: 26408009-26408009 on Build GRCh38
rs544451736 Chr.6: 26408008-26408008 on Build GRCh38
rs147375005 Chr.6: 26407690-26407690 on Build GRCh38
rs373041261 Chr.6: 26407958-26407958 on Build GRCh38
rs961694188 Chr.6: 26407978-26407978 on Build GRCh38
rs1004168021 Chr.6: 26408021-26408021 on Build GRCh38
rs201789912 Chr.6: 26407916-26407916 on Build GRCh38
rs973409411 Chr.6: 26407805-26407805 on Build GRCh38
rs762071540 Chr.6: 26407968-26407968 on Build GRCh38
rs148613503 Chr.6: 26407815-26407815 on Build GRCh38
rs764942378 Chr.6: 26407751-26407751 on Build GRCh38
rs140260146 Chr.6: 26407903-26407903 on Build GRCh38
rs764430378 Chr.6: 26407900-26407900 on Build GRCh38
rs138215264 Chr.6: 26407783-26407783 on Build GRCh38
rs746703088 Chr.6: 26407777-26407779 on Build GRCh38
rs112574152 Chr.6: 26407824-26407824 on Build GRCh38
rs147150065 Chr.6: 26407716-26407716 on Build GRCh38
rs542657364 Chr.6: 26407855-26407855 on Build GRCh38
rs756180920 Chr.6: 26407655-26407655 on Build GRCh38
rs557579831 Chr.6: 26407727-26407727 on Build GRCh38
rs756770560 Chr.6: 26407948-26407948 on Build GRCh38
rs781095568 Chr.6: 26407828-26407829 on Build GRCh38
rs999834969 Chr.6: 26407660-26407660 on Build GRCh38
rs538727485 Chr.6: 26407713-26407713 on Build GRCh38
rs546621070 Chr.6: 26408087-26408087 on Build GRCh38
rs778950432 Chr.6: 26407819-26407819 on Build GRCh38

View Full Product Details