Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26411462-26411957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A1
Entrez Gene ID
11119
Gene Name
butyrophilin subfamily 3 member A1
Gene Aliases
BT3.1, BTF5, BTN3.1, CD277
UniGene
Hs.191510
Gene Chromosome Location
Chr.6: 26402237-26415216 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
NM_001145008.1 NM_001145009.1 NM_007048.5 NM_194441.2 XM_005248833.2 XM_005248834.3 XM_006714983.2 XM_011514268.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7088784 c.992-1G>A p.? Chr.6: 26411554-26411554 on Build GRCh38
7313407 c.1003G>T p.A335S Chr.6: 26411566-26411566 on Build GRCh38
3777395 c.992-1G>A p.? Chr.6: 26411554-26411554 on Build GRCh38
7313408 c.1003G>T p.A335S Chr.6: 26411566-26411566 on Build GRCh38
3860994 c.1004C>T p.A335V Chr.6: 26411567-26411567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767629073 Chr.6: 26411542-26411542 on Build GRCh38
rs771655111 Chr.6: 26411687-26411687 on Build GRCh38
rs756388157 Chr.6: 26411628-26411628 on Build GRCh38
rs555838811 Chr.6: 26411792-26411792 on Build GRCh38
rs765516907 Chr.6: 26411592-26411592 on Build GRCh38
rs368630675 Chr.6: 26411526-26411526 on Build GRCh38
rs527423717 Chr.6: 26411577-26411577 on Build GRCh38
rs376097422 Chr.6: 26411519-26411519 on Build GRCh38
rs12194411 Chr.6: 26411852-26411852 on Build GRCh38
rs933114274 Chr.6: 26411593-26411593 on Build GRCh38
rs770847986 Chr.6: 26411582-26411582 on Build GRCh38
rs942197763 Chr.6: 26411916-26411916 on Build GRCh38
rs1050614178 Chr.6: 26411597-26411597 on Build GRCh38
rs867850017 Chr.6: 26411708-26411708 on Build GRCh38
rs539665764 Chr.6: 26411689-26411689 on Build GRCh38
rs754704911 Chr.6: 26411547-26411547 on Build GRCh38
rs957741258 Chr.6: 26411761-26411761 on Build GRCh38
rs748550635 Chr.6: 26411833-26411833 on Build GRCh38
rs556703265 Chr.6: 26411694-26411694 on Build GRCh38
rs12194412 Chr.6: 26411853-26411853 on Build GRCh38
rs779195300 Chr.6: 26411599-26411599 on Build GRCh38
rs910312456 Chr.6: 26411886-26411886 on Build GRCh38
rs778410079 Chr.6: 26411552-26411552 on Build GRCh38
rs987858734 Chr.6: 26411535-26411535 on Build GRCh38
rs745423783 Chr.6: 26411682-26411682 on Build GRCh38
rs578021769 Chr.6: 26411871-26411871 on Build GRCh38
rs192941342 Chr.6: 26411797-26411797 on Build GRCh38
rs757709470 Chr.6: 26411525-26411525 on Build GRCh38
rs904987218 Chr.6: 26411762-26411762 on Build GRCh38
rs200674588 Chr.6: 26411587-26411587 on Build GRCh38
rs747002828 Chr.6: 26411580-26411580 on Build GRCh38
rs931117145 Chr.6: 26411915-26411915 on Build GRCh38
rs912943907 Chr.6: 26411794-26411794 on Build GRCh38
rs1012971757 Chr.6: 26411757-26411757 on Build GRCh38
rs547099840 Chr.6: 26411634-26411634 on Build GRCh38
rs750593163 Chr.6: 26411580-26411580 on Build GRCh38
rs750852378 Chr.6: 26411624-26411624 on Build GRCh38
rs1057154183 Chr.6: 26411716-26411716 on Build GRCh38
rs751676073 Chr.6: 26411603-26411603 on Build GRCh38
rs780502361 Chr.6: 26411518-26411518 on Build GRCh38
rs370669442 Chr.6: 26411543-26411543 on Build GRCh38
rs542848241 Chr.6: 26411709-26411709 on Build GRCh38
rs760962813 Chr.6: 26411626-26411633 on Build GRCh38
rs768346667 Chr.6: 26411776-26411776 on Build GRCh38
rs986421672 Chr.6: 26411826-26411826 on Build GRCh38
rs912518603 Chr.6: 26411841-26411841 on Build GRCh38
rs768110606 Chr.6: 26411620-26411620 on Build GRCh38
rs747719629 Chr.6: 26411568-26411568 on Build GRCh38
rs1032253264 Chr.6: 26411783-26411783 on Build GRCh38
rs386698237 Chr.6: 26411852-26411853 on Build GRCh38
rs927071483 Chr.6: 26411489-26411489 on Build GRCh38
rs756713597 Chr.6: 26411596-26411596 on Build GRCh38
rs183680768 Chr.6: 26411884-26411884 on Build GRCh38
rs28571747 Chr.6: 26411759-26411759 on Build GRCh38
rs12214924 Chr.6: 26411667-26411667 on Build GRCh38
rs1028126256 Chr.6: 26411672-26411672 on Build GRCh38
rs12214976 Chr.6: 26411735-26411735 on Build GRCh38
rs891007984 Chr.6: 26411610-26411610 on Build GRCh38
rs757907368 Chr.6: 26411614-26411614 on Build GRCh38
rs776528991 Chr.6: 26411584-26411584 on Build GRCh38
rs985860208 Chr.6: 26411861-26411861 on Build GRCh38
rs900608230 Chr.6: 26411656-26411656 on Build GRCh38
rs574630664 Chr.6: 26411567-26411567 on Build GRCh38

View Full Product Details