Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26458347-26458846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN2A1
Entrez Gene ID
11120
Gene Name
butyrophilin subfamily 2 member A1
Gene Aliases
BK14H9.1, BT2.1, BTF1, BTN2.1, DJ3E1.1
UniGene
Hs.159028
Gene Chromosome Location
Chr.6: 26457904-26476621 on Build GRCh38
Transcripts
NM_001197233.2 NM_001197234.2 NM_007049.4 NM_078476.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2858425 c.8C>T p.S3L Chr.6: 26458644-26458644 on Build GRCh38
4666385 c.41_42insCCT p.L20_S21insL Chr.6: 26458677-26458678 on Build GRCh38
6348780 c.16G>T p.A6S Chr.6: 26458652-26458652 on Build GRCh38
6348779 c.16G>T p.A6S Chr.6: 26458652-26458652 on Build GRCh38
5564253 c.33G>A p.R11R Chr.6: 26458669-26458669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747773721 Chr.6: 26458698-26458698 on Build GRCh38
rs765094199 Chr.6: 26458676-26458676 on Build GRCh38
rs759871915 Chr.6: 26458617-26458617 on Build GRCh38
rs552157771 Chr.6: 26458750-26458750 on Build GRCh38
rs373156116 Chr.6: 26458759-26458759 on Build GRCh38
rs1039345385 Chr.6: 26458415-26458415 on Build GRCh38
rs2393662 Chr.6: 26458470-26458470 on Build GRCh38
rs749668726 Chr.6: 26458696-26458698 on Build GRCh38
rs908431710 Chr.6: 26458524-26458524 on Build GRCh38
rs975082642 Chr.6: 26458431-26458431 on Build GRCh38
rs776749750 Chr.6: 26458719-26458719 on Build GRCh38
rs753622901 Chr.6: 26458609-26458609 on Build GRCh38
rs746481774 Chr.6: 26458715-26458715 on Build GRCh38
rs1026606445 Chr.6: 26458458-26458458 on Build GRCh38
rs548365439 Chr.6: 26458468-26458468 on Build GRCh38
rs996555951 Chr.6: 26458708-26458708 on Build GRCh38
rs911698191 Chr.6: 26458387-26458387 on Build GRCh38
rs770545022 Chr.6: 26458717-26458717 on Build GRCh38
rs757349495 Chr.6: 26458674-26458677 on Build GRCh38
rs568723410 Chr.6: 26458754-26458754 on Build GRCh38
rs867566268 Chr.6: 26458473-26458473 on Build GRCh38
rs377102930 Chr.6: 26458395-26458395 on Build GRCh38
rs769991377 Chr.6: 26458678-26458683 on Build GRCh38
rs146399224 Chr.6: 26458643-26458643 on Build GRCh38
rs3857553 Chr.6: 26458466-26458466 on Build GRCh38
rs963751435 Chr.6: 26458429-26458429 on Build GRCh38
rs200786224 Chr.6: 26458626-26458626 on Build GRCh38
rs765509634 Chr.6: 26458620-26458620 on Build GRCh38
rs777982807 Chr.6: 26458494-26458494 on Build GRCh38
rs903364192 Chr.6: 26458410-26458410 on Build GRCh38
rs1015955258 Chr.6: 26458796-26458796 on Build GRCh38
rs780380718 Chr.6: 26458691-26458691 on Build GRCh38
rs1047423345 Chr.6: 26458480-26458480 on Build GRCh38
rs751615832 Chr.6: 26458680-26458680 on Build GRCh38
rs1024028411 Chr.6: 26458525-26458525 on Build GRCh38
rs1013901577 Chr.6: 26458763-26458763 on Build GRCh38
rs774410521 Chr.6: 26458758-26458758 on Build GRCh38
rs745797236 Chr.6: 26458732-26458732 on Build GRCh38
rs199747313 Chr.6: 26458637-26458637 on Build GRCh38
rs561166376 Chr.6: 26458499-26458499 on Build GRCh38
rs554227538 Chr.6: 26458677-26458678 on Build GRCh38
rs142203065 Chr.6: 26458639-26458639 on Build GRCh38
rs756533999 Chr.6: 26458690-26458690 on Build GRCh38
rs764085206 Chr.6: 26458679-26458679 on Build GRCh38
rs201060519 Chr.6: 26458597-26458597 on Build GRCh38
rs760536633 Chr.6: 26458667-26458667 on Build GRCh38
rs140375890 Chr.6: 26458677-26458677 on Build GRCh38
rs771693277 Chr.6: 26458699-26458699 on Build GRCh38
rs1041188606 Chr.6: 26458683-26458683 on Build GRCh38
rs952346462 Chr.6: 26458496-26458498 on Build GRCh38
rs752021152 Chr.6: 26458635-26458635 on Build GRCh38
rs781698683 Chr.6: 26458684-26458684 on Build GRCh38
rs920502862 Chr.6: 26458671-26458671 on Build GRCh38
rs970660574 Chr.6: 26458818-26458818 on Build GRCh38
rs771575009 Chr.6: 26458591-26458591 on Build GRCh38
rs943235405 Chr.6: 26458411-26458411 on Build GRCh38
rs777181223 Chr.6: 26458714-26458714 on Build GRCh38
rs757726145 Chr.6: 26458681-26458681 on Build GRCh38
rs776590945 Chr.6: 26458672-26458672 on Build GRCh38
rs531848316 Chr.6: 26458408-26458408 on Build GRCh38
rs913203002 Chr.6: 26458638-26458638 on Build GRCh38
rs772724912 Chr.6: 26458661-26458661 on Build GRCh38
rs2393663 Chr.6: 26458500-26458500 on Build GRCh38
rs775288227 Chr.6: 26458751-26458751 on Build GRCh38
rs769724341 Chr.6: 26458749-26458749 on Build GRCh38
rs771543600 Chr.6: 26458660-26458660 on Build GRCh38
rs970615429 Chr.6: 26458529-26458529 on Build GRCh38
rs761276076 Chr.6: 26458590-26458590 on Build GRCh38
rs139658946 Chr.6: 26458670-26458670 on Build GRCh38
rs1001741896 Chr.6: 26458550-26458550 on Build GRCh38
rs7750281 Chr.6: 26458787-26458787 on Build GRCh38
rs777895127 Chr.6: 26458627-26458627 on Build GRCh38
rs756497385 Chr.6: 26458678-26458678 on Build GRCh38
rs1038899519 Chr.6: 26458668-26458668 on Build GRCh38
rs766104679 Chr.6: 26458603-26458603 on Build GRCh38
rs777769829 Chr.6: 26458633-26458634 on Build GRCh38
rs1033143002 Chr.6: 26458789-26458789 on Build GRCh38
rs200522594 Chr.6: 26458762-26458762 on Build GRCh38
rs372951914 Chr.6: 26458775-26458775 on Build GRCh38
rs761847776 Chr.6: 26458766-26458766 on Build GRCh38
rs778572796 Chr.6: 26458644-26458644 on Build GRCh38
rs751394675 Chr.6: 26458647-26458654 on Build GRCh38
rs899108912 Chr.6: 26458445-26458445 on Build GRCh38
rs759454614 Chr.6: 26458673-26458673 on Build GRCh38
rs747828235 Chr.6: 26458655-26458655 on Build GRCh38

View Full Product Details