Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26508416-26508919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN1A1
Entrez Gene ID
696
Gene Name
butyrophilin subfamily 1 member A1
Gene Aliases
BT, BTN, BTN1
UniGene
Hs.153058
Gene Chromosome Location
Chr.6: 26499290-26510425 on Build GRCh38
Transcripts
NM_001732.2 XM_005249340.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6513825 c.952C>A p.L318I Chr.6: 26508545-26508545 on Build GRCh38
2858534 c.1006G>C p.E336Q Chr.6: 26508599-26508599 on Build GRCh38
5808680 c.1052A>G p.E351G Chr.6: 26508645-26508645 on Build GRCh38
6484894 c.1038T>A p.C346* Chr.6: 26508631-26508631 on Build GRCh38
2858542 c.1170G>T p.E390D Chr.6: 26508763-26508763 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773129427 Chr.6: 26508798-26508798 on Build GRCh38
rs747345849 Chr.6: 26508607-26508607 on Build GRCh38
rs778400918 Chr.6: 26508713-26508713 on Build GRCh38
rs900851330 Chr.6: 26508446-26508446 on Build GRCh38
rs1017131548 Chr.6: 26508621-26508621 on Build GRCh38
rs371312377 Chr.6: 26508809-26508809 on Build GRCh38
rs765357924 Chr.6: 26508709-26508709 on Build GRCh38
rs755100540 Chr.6: 26508687-26508687 on Build GRCh38
rs267600914 Chr.6: 26508832-26508832 on Build GRCh38
rs536850553 Chr.6: 26508584-26508584 on Build GRCh38
rs201452649 Chr.6: 26508627-26508627 on Build GRCh38
rs757554363 Chr.6: 26508724-26508724 on Build GRCh38
rs867829785 Chr.6: 26508872-26508872 on Build GRCh38
rs747726672 Chr.6: 26508861-26508861 on Build GRCh38
rs768247954 Chr.6: 26508756-26508756 on Build GRCh38
rs778241200 Chr.6: 26508585-26508585 on Build GRCh38
rs747221306 Chr.6: 26508825-26508825 on Build GRCh38
rs369973508 Chr.6: 26508855-26508855 on Build GRCh38
rs754677050 Chr.6: 26508854-26508854 on Build GRCh38
rs780321806 Chr.6: 26508704-26508704 on Build GRCh38
rs774987970 Chr.6: 26508613-26508613 on Build GRCh38
rs760377425 Chr.6: 26508660-26508660 on Build GRCh38
rs775105638 Chr.6: 26508455-26508455 on Build GRCh38
rs374953117 Chr.6: 26508835-26508835 on Build GRCh38
rs774265636 Chr.6: 26508485-26508485 on Build GRCh38
rs763111469 Chr.6: 26508836-26508836 on Build GRCh38
rs778436239 Chr.6: 26508769-26508769 on Build GRCh38
rs764387639 Chr.6: 26508540-26508543 on Build GRCh38
rs138457017 Chr.6: 26508779-26508779 on Build GRCh38
rs368401858 Chr.6: 26508530-26508530 on Build GRCh38
rs770576601 Chr.6: 26508811-26508811 on Build GRCh38
rs764761095 Chr.6: 26508845-26508845 on Build GRCh38
rs1040065853 Chr.6: 26508490-26508490 on Build GRCh38
rs765523860 Chr.6: 26508822-26508822 on Build GRCh38
rs202063334 Chr.6: 26508852-26508852 on Build GRCh38
rs753825745 Chr.6: 26508477-26508477 on Build GRCh38
rs759635479 Chr.6: 26508816-26508816 on Build GRCh38
rs753942003 Chr.6: 26508676-26508676 on Build GRCh38
rs557646919 Chr.6: 26508519-26508519 on Build GRCh38
rs901579443 Chr.6: 26508496-26508496 on Build GRCh38
rs775011189 Chr.6: 26508607-26508608 on Build GRCh38
rs766691983 Chr.6: 26508667-26508667 on Build GRCh38
rs765355838 Chr.6: 26508457-26508458 on Build GRCh38
rs754008731 Chr.6: 26508545-26508545 on Build GRCh38
rs773043636 Chr.6: 26508642-26508642 on Build GRCh38
rs756404007 Chr.6: 26508535-26508535 on Build GRCh38
rs188923166 Chr.6: 26508869-26508869 on Build GRCh38
rs755009512 Chr.6: 26508569-26508569 on Build GRCh38
rs143821520 Chr.6: 26508752-26508752 on Build GRCh38
rs748891499 Chr.6: 26508619-26508619 on Build GRCh38
rs757651220 Chr.6: 26508851-26508851 on Build GRCh38
rs542740353 Chr.6: 26508710-26508710 on Build GRCh38
rs769533934 Chr.6: 26508611-26508611 on Build GRCh38
rs748831182 Chr.6: 26508753-26508753 on Build GRCh38
rs760523424 Chr.6: 26508802-26508802 on Build GRCh38
rs1035055406 Chr.6: 26508648-26508648 on Build GRCh38
rs28463517 Chr.6: 26508727-26508727 on Build GRCh38
rs1015737982 Chr.6: 26508829-26508829 on Build GRCh38
rs761702210 Chr.6: 26508640-26508640 on Build GRCh38
rs752997989 Chr.6: 26508480-26508482 on Build GRCh38
rs990817013 Chr.6: 26508652-26508652 on Build GRCh38
rs751939842 Chr.6: 26508846-26508846 on Build GRCh38
rs1980600 Chr.6: 26508501-26508501 on Build GRCh38
rs374903527 Chr.6: 26508722-26508722 on Build GRCh38
rs867351766 Chr.6: 26508821-26508821 on Build GRCh38
rs748609018 Chr.6: 26508570-26508570 on Build GRCh38
rs868024333 Chr.6: 26508815-26508815 on Build GRCh38
rs777779954 Chr.6: 26508883-26508883 on Build GRCh38
rs148996980 Chr.6: 26508641-26508641 on Build GRCh38
rs752873211 Chr.6: 26508827-26508827 on Build GRCh38
rs757074593 Chr.6: 26508505-26508505 on Build GRCh38
rs866094593 Chr.6: 26508758-26508758 on Build GRCh38
rs368135520 Chr.6: 26508794-26508794 on Build GRCh38
rs142696565 Chr.6: 26508543-26508543 on Build GRCh38
rs199986830 Chr.6: 26508712-26508712 on Build GRCh38
rs371101297 Chr.6: 26508463-26508463 on Build GRCh38
rs375023084 Chr.6: 26508473-26508473 on Build GRCh38

View Full Product Details