Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36686410-36686917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1A
Entrez Gene ID
1026
Gene Name
cyclin dependent kinase inhibitor 1A
Gene Aliases
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
UniGene
Hs.370771
Gene Chromosome Location
Chr.6: 36676460-36687339 on Build GRCh38
Transcripts
NM_000389.4 NM_001220777.1 NM_001220778.1 NM_001291549.1 NM_078467.2

SNP Detail

SNP ID Chromosome Location
rs375384268 Chr.6: 36686850-36686850 on Build GRCh38
rs139090547 Chr.6: 36686645-36686645 on Build GRCh38
rs908342499 Chr.6: 36686765-36686765 on Build GRCh38
rs757862523 Chr.6: 36686468-36686468 on Build GRCh38
rs991560468 Chr.6: 36686718-36686718 on Build GRCh38
rs566590564 Chr.6: 36686676-36686676 on Build GRCh38
rs1035067351 Chr.6: 36686554-36686554 on Build GRCh38
rs772094412 Chr.6: 36686717-36686717 on Build GRCh38
rs774512258 Chr.6: 36686508-36686508 on Build GRCh38
rs894366694 Chr.6: 36686437-36686437 on Build GRCh38
rs1009737725 Chr.6: 36686614-36686614 on Build GRCh38
rs544057426 Chr.6: 36686611-36686611 on Build GRCh38
rs967514962 Chr.6: 36686650-36686650 on Build GRCh38
rs941387040 Chr.6: 36686455-36686455 on Build GRCh38
rs567103693 Chr.6: 36686713-36686713 on Build GRCh38
rs529198430 Chr.6: 36686580-36686580 on Build GRCh38
rs1048397424 Chr.6: 36686463-36686463 on Build GRCh38
rs1014932119 Chr.6: 36686532-36686532 on Build GRCh38
rs557652835 Chr.6: 36686769-36686769 on Build GRCh38
rs1038653449 Chr.6: 36686848-36686848 on Build GRCh38
rs551145694 Chr.6: 36686673-36686673 on Build GRCh38
rs990366166 Chr.6: 36686595-36686595 on Build GRCh38
rs939842159 Chr.6: 36686854-36686854 on Build GRCh38
rs540666701 Chr.6: 36686491-36686491 on Build GRCh38
rs933848673 Chr.6: 36686742-36686742 on Build GRCh38
rs562367477 Chr.6: 36686640-36686640 on Build GRCh38
rs1021650885 Chr.6: 36686620-36686620 on Build GRCh38
rs370572240 Chr.6: 36686464-36686464 on Build GRCh38
rs184742749 Chr.6: 36686454-36686454 on Build GRCh38
rs995702977 Chr.6: 36686441-36686441 on Build GRCh38
rs1004007357 Chr.6: 36686516-36686516 on Build GRCh38
rs997905489 Chr.6: 36686569-36686569 on Build GRCh38
rs540064877 Chr.6: 36686870-36686870 on Build GRCh38
rs548971366 Chr.6: 36686696-36686696 on Build GRCh38
rs74801436 Chr.6: 36686747-36686747 on Build GRCh38
rs181534480 Chr.6: 36686669-36686669 on Build GRCh38
rs980065929 Chr.6: 36686652-36686652 on Build GRCh38
rs1059247 Chr.6: 36686822-36686822 on Build GRCh38
rs968318210 Chr.6: 36686621-36686621 on Build GRCh38
rs186592256 Chr.6: 36686679-36686679 on Build GRCh38
rs6457937 Chr.6: 36686574-36686574 on Build GRCh38
rs189202045 Chr.6: 36686467-36686467 on Build GRCh38
rs10046116 Chr.6: 36686684-36686684 on Build GRCh38
rs367706888 Chr.6: 36686439-36686439 on Build GRCh38
rs191000568 Chr.6: 36686830-36686830 on Build GRCh38
rs1040878666 Chr.6: 36686866-36686866 on Build GRCh38
rs1002289641 Chr.6: 36686541-36686541 on Build GRCh38

View Full Product Details