Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36687050-36687538 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDKN1A
Entrez Gene ID
1026
Gene Name
cyclin dependent kinase inhibitor 1A
Gene Aliases
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
UniGene
Hs.370771
Gene Chromosome Location
Chr.6: 36676460-36687339 on Build GRCh38
Transcripts
NM_000389.4 NM_001220777.1 NM_001220778.1 NM_001291549.1 NM_078467.2

SNP Detail

SNP ID Chromosome Location
rs952486975 Chr.6: 36687374-36687377 on Build GRCh38
rs568384836 Chr.6: 36687487-36687487 on Build GRCh38
rs903433616 Chr.6: 36687203-36687203 on Build GRCh38
rs73730143 Chr.6: 36687202-36687202 on Build GRCh38
rs982916069 Chr.6: 36687215-36687215 on Build GRCh38
rs182235412 Chr.6: 36687098-36687098 on Build GRCh38
rs941154808 Chr.6: 36687278-36687278 on Build GRCh38
rs549777937 Chr.6: 36687311-36687311 on Build GRCh38
rs749625919 Chr.6: 36687368-36687368 on Build GRCh38
rs1036363838 Chr.6: 36687251-36687251 on Build GRCh38
rs965836001 Chr.6: 36687402-36687402 on Build GRCh38
rs531369735 Chr.6: 36687230-36687230 on Build GRCh38
rs932472522 Chr.6: 36687477-36687477 on Build GRCh38
rs1013522366 Chr.6: 36687343-36687343 on Build GRCh38
rs908248631 Chr.6: 36687406-36687406 on Build GRCh38
rs1022686135 Chr.6: 36687354-36687354 on Build GRCh38
rs1802570 Chr.6: 36687095-36687095 on Build GRCh38
rs555547784 Chr.6: 36687510-36687510 on Build GRCh38
rs190930058 Chr.6: 36687448-36687448 on Build GRCh38
rs185976780 Chr.6: 36687099-36687099 on Build GRCh38
rs917897545 Chr.6: 36687475-36687475 on Build GRCh38
rs527383432 Chr.6: 36687188-36687188 on Build GRCh38
rs745953295 Chr.6: 36687179-36687179 on Build GRCh38
rs778257166 Chr.6: 36687231-36687231 on Build GRCh38
rs961036608 Chr.6: 36687428-36687428 on Build GRCh38
rs566907275 Chr.6: 36687403-36687403 on Build GRCh38
rs766360274 Chr.6: 36687260-36687261 on Build GRCh38
rs908447221 Chr.6: 36687072-36687072 on Build GRCh38
rs1136646 Chr.6: 36687282-36687282 on Build GRCh38
rs963715824 Chr.6: 36687160-36687160 on Build GRCh38
rs548775480 Chr.6: 36687193-36687193 on Build GRCh38
rs1050054641 Chr.6: 36687504-36687504 on Build GRCh38
rs564442732 Chr.6: 36687272-36687272 on Build GRCh38
rs11550973 Chr.6: 36687187-36687187 on Build GRCh38
rs544851636 Chr.6: 36687103-36687103 on Build GRCh38
rs569616015 Chr.6: 36687347-36687347 on Build GRCh38
rs558338089 Chr.6: 36687384-36687384 on Build GRCh38
rs922527555 Chr.6: 36687453-36687453 on Build GRCh38
rs932480638 Chr.6: 36687185-36687185 on Build GRCh38
rs1011571683 Chr.6: 36687196-36687196 on Build GRCh38
rs540125889 Chr.6: 36687373-36687373 on Build GRCh38

View Full Product Details