Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 37482830-37483328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC167
Entrez Gene ID
154467
Gene Name
coiled-coil domain containing 167
Gene Aliases
C6orf129, HSPC265
UniGene
Hs.284207
Gene Chromosome Location
Chr.6: 37482920-37499924 on Build GRCh38
Transcripts
NM_138493.2 XM_005248955.3

Gene Symbol
CMTR1
Entrez Gene ID
23070
Gene Name
cap methyltransferase 1
Gene Aliases
FTSJD2, KIAA0082, MTr1, hMTr1
UniGene
Hs.520102
Gene Chromosome Location
Chr.6: 37433131-37482844 on Build GRCh38
Transcripts
NM_138493.2 XM_005248955.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5807603 c.263T>A p.L88Q Chr.6: 37483217-37483217 on Build GRCh38
7289091 c.159G>A p.S53S Chr.6: 37483268-37483268 on Build GRCh38
1078678 c.290T>C p.M97T Chr.6: 37483190-37483190 on Build GRCh38
5870540 c.259C>T p.L87F Chr.6: 37483221-37483221 on Build GRCh38
6331528 c.191-5C>A p.? Chr.6: 37483294-37483294 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933469743 Chr.6: 37483157-37483157 on Build GRCh38
rs185138437 Chr.6: 37483014-37483014 on Build GRCh38
rs991850545 Chr.6: 37482898-37482898 on Build GRCh38
rs776904706 Chr.6: 37483084-37483084 on Build GRCh38
rs140128298 Chr.6: 37482857-37482857 on Build GRCh38
rs146381766 Chr.6: 37483108-37483108 on Build GRCh38
rs899273132 Chr.6: 37483188-37483188 on Build GRCh38
rs747968759 Chr.6: 37482973-37482973 on Build GRCh38
rs778239529 Chr.6: 37483229-37483229 on Build GRCh38
rs371542950 Chr.6: 37483305-37483305 on Build GRCh38
rs924087889 Chr.6: 37482956-37482956 on Build GRCh38
rs767172012 Chr.6: 37483210-37483210 on Build GRCh38
rs753491069 Chr.6: 37483291-37483291 on Build GRCh38
rs769608195 Chr.6: 37482981-37482981 on Build GRCh38
rs925065077 Chr.6: 37483126-37483126 on Build GRCh38
rs933564085 Chr.6: 37483004-37483004 on Build GRCh38
rs1050218358 Chr.6: 37483296-37483296 on Build GRCh38
rs573565030 Chr.6: 37483195-37483195 on Build GRCh38
rs922211351 Chr.6: 37483000-37483000 on Build GRCh38
rs772580256 Chr.6: 37483160-37483160 on Build GRCh38
rs562528230 Chr.6: 37482882-37482882 on Build GRCh38
rs748552996 Chr.6: 37483153-37483153 on Build GRCh38
rs774412083 Chr.6: 37483275-37483275 on Build GRCh38
rs1005392544 Chr.6: 37482856-37482856 on Build GRCh38
rs761869302 Chr.6: 37483277-37483277 on Build GRCh38
rs565515174 Chr.6: 37482992-37482992 on Build GRCh38
rs747105669 Chr.6: 37482916-37482916 on Build GRCh38
rs776696014 Chr.6: 37483174-37483174 on Build GRCh38
rs373363248 Chr.6: 37482901-37482901 on Build GRCh38
rs138745679 Chr.6: 37483209-37483209 on Build GRCh38
rs780990597 Chr.6: 37483139-37483139 on Build GRCh38
rs1024852288 Chr.6: 37483095-37483095 on Build GRCh38
rs148157970 Chr.6: 37483285-37483285 on Build GRCh38
rs148927861 Chr.6: 37483256-37483256 on Build GRCh38
rs766049728 Chr.6: 37483290-37483290 on Build GRCh38
rs773299048 Chr.6: 37483061-37483061 on Build GRCh38
rs755810495 Chr.6: 37483213-37483213 on Build GRCh38
rs188157505 Chr.6: 37483248-37483248 on Build GRCh38
rs772097066 Chr.6: 37483059-37483059 on Build GRCh38
rs762743334 Chr.6: 37483204-37483204 on Build GRCh38
rs1013527739 Chr.6: 37483089-37483089 on Build GRCh38
rs556185318 Chr.6: 37483214-37483214 on Build GRCh38
rs3734325 Chr.6: 37483075-37483075 on Build GRCh38
rs953008410 Chr.6: 37482911-37482911 on Build GRCh38
rs749689778 Chr.6: 37483303-37483303 on Build GRCh38
rs7771002 Chr.6: 37482879-37482879 on Build GRCh38
rs551581753 Chr.6: 37482984-37482984 on Build GRCh38
rs574881427 Chr.6: 37483048-37483048 on Build GRCh38
rs184295314 Chr.6: 37483133-37483133 on Build GRCh38
rs541234466 Chr.6: 37483269-37483269 on Build GRCh38
rs750000322 Chr.6: 37483212-37483212 on Build GRCh38
rs778151727 Chr.6: 37483304-37483304 on Build GRCh38
rs768532759 Chr.6: 37482932-37482932 on Build GRCh38
rs533701063 Chr.6: 37483148-37483148 on Build GRCh38
rs959163879 Chr.6: 37483077-37483077 on Build GRCh38
rs771056654 Chr.6: 37483169-37483169 on Build GRCh38
rs758740406 Chr.6: 37483295-37483295 on Build GRCh38
rs1028777350 Chr.6: 37482902-37482902 on Build GRCh38
rs762646331 Chr.6: 37483040-37483040 on Build GRCh38
rs1036297164 Chr.6: 37483047-37483047 on Build GRCh38
rs780259137 Chr.6: 37483237-37483238 on Build GRCh38
rs902866093 Chr.6: 37483136-37483136 on Build GRCh38
rs764902665 Chr.6: 37483190-37483190 on Build GRCh38
rs1033634150 Chr.6: 37482886-37482886 on Build GRCh38
rs775083695 Chr.6: 37483268-37483268 on Build GRCh38
rs201463577 Chr.6: 37483161-37483161 on Build GRCh38
rs758827762 Chr.6: 37483217-37483217 on Build GRCh38
rs969838894 Chr.6: 37483255-37483255 on Build GRCh38
rs894982062 Chr.6: 37483236-37483236 on Build GRCh38
rs568845679 Chr.6: 37483107-37483107 on Build GRCh38
rs973924157 Chr.6: 37482884-37482884 on Build GRCh38
rs180956135 Chr.6: 37483289-37483289 on Build GRCh38
rs1050168480 Chr.6: 37483081-37483081 on Build GRCh38
rs1000156841 Chr.6: 37483137-37483137 on Build GRCh38
rs775380039 Chr.6: 37483200-37483200 on Build GRCh38
rs201946781 Chr.6: 37483260-37483260 on Build GRCh38
rs764135458 Chr.6: 37483205-37483205 on Build GRCh38
rs13216875 Chr.6: 37483027-37483027 on Build GRCh38
rs908087487 Chr.6: 37483163-37483163 on Build GRCh38
rs191706434 Chr.6: 37483068-37483068 on Build GRCh38
rs995360844 Chr.6: 37483071-37483071 on Build GRCh38
rs755263071 Chr.6: 37483146-37483146 on Build GRCh38
rs374683329 Chr.6: 37483279-37483279 on Build GRCh38
rs959255269 Chr.6: 37482891-37482891 on Build GRCh38
rs763650695 Chr.6: 37483130-37483130 on Build GRCh38
rs146517218 Chr.6: 37483208-37483208 on Build GRCh38
rs10692 Chr.6: 37483138-37483138 on Build GRCh38
rs368185026 Chr.6: 37483177-37483177 on Build GRCh38
rs576020127 Chr.6: 37483216-37483216 on Build GRCh38
rs747086703 Chr.6: 37483167-37483167 on Build GRCh38
rs1049498286 Chr.6: 37483010-37483010 on Build GRCh38
rs1004848103 Chr.6: 37483018-37483018 on Build GRCh38
rs143824551 Chr.6: 37483273-37483273 on Build GRCh38
rs1051938356 Chr.6: 37483009-37483009 on Build GRCh38
rs765130399 Chr.6: 37483098-37483098 on Build GRCh38
rs925023808 Chr.6: 37482934-37482934 on Build GRCh38
rs997240573 Chr.6: 37482883-37482883 on Build GRCh38
rs916686112 Chr.6: 37482979-37482979 on Build GRCh38
rs766884902 Chr.6: 37483281-37483281 on Build GRCh38
rs746029085 Chr.6: 37483261-37483261 on Build GRCh38
rs759238516 Chr.6: 37483092-37483092 on Build GRCh38
rs531720298 Chr.6: 37482914-37482914 on Build GRCh38
rs374390175 Chr.6: 37482933-37482933 on Build GRCh38
rs189266240 Chr.6: 37483036-37483036 on Build GRCh38
rs960872899 Chr.6: 37482975-37482977 on Build GRCh38
rs992189211 Chr.6: 37482976-37482976 on Build GRCh38
rs770070407 Chr.6: 37483267-37483267 on Build GRCh38
rs745596438 Chr.6: 37483144-37483144 on Build GRCh38

View Full Product Details