Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41067123-41067618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OARD1
Entrez Gene ID
221443
Gene Name
O-acyl-ADP-ribose deacylase 1
Gene Aliases
C6orf130, TARG1, dJ34B21.3
UniGene
Hs.227457
Gene Chromosome Location
Chr.6: 41066792-41072449 on Build GRCh38
Transcripts
NM_145063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4006501 c.421G>C p.E141Q Chr.6: 41067373-41067373 on Build GRCh38
1444465 c.376C>T p.R126C Chr.6: 41067418-41067418 on Build GRCh38
5061779 c.363A>C p.G121G Chr.6: 41067431-41067431 on Build GRCh38
1079164 c.409G>A p.E137K Chr.6: 41067385-41067385 on Build GRCh38
1079163 c.445G>A p.V149M Chr.6: 41067349-41067349 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182802798 Chr.6: 41067273-41067273 on Build GRCh38
rs374588660 Chr.6: 41067351-41067351 on Build GRCh38
rs776232037 Chr.6: 41067309-41067309 on Build GRCh38
rs768301317 Chr.6: 41067489-41067489 on Build GRCh38
rs975462284 Chr.6: 41067562-41067562 on Build GRCh38
rs1011375498 Chr.6: 41067148-41067148 on Build GRCh38
rs769634633 Chr.6: 41067531-41067531 on Build GRCh38
rs1019582206 Chr.6: 41067543-41067543 on Build GRCh38
rs567307625 Chr.6: 41067575-41067575 on Build GRCh38
rs565039431 Chr.6: 41067174-41067174 on Build GRCh38
rs531858470 Chr.6: 41067496-41067496 on Build GRCh38
rs753143818 Chr.6: 41067596-41067596 on Build GRCh38
rs758596299 Chr.6: 41067463-41067463 on Build GRCh38
rs759594221 Chr.6: 41067455-41067455 on Build GRCh38
rs1011259725 Chr.6: 41067479-41067479 on Build GRCh38
rs759274647 Chr.6: 41067311-41067311 on Build GRCh38
rs960192467 Chr.6: 41067589-41067589 on Build GRCh38
rs367707236 Chr.6: 41067362-41067362 on Build GRCh38
rs965458188 Chr.6: 41067165-41067165 on Build GRCh38
rs780849655 Chr.6: 41067397-41067397 on Build GRCh38
rs958141022 Chr.6: 41067591-41067591 on Build GRCh38
rs751332139 Chr.6: 41067467-41067467 on Build GRCh38
rs201526617 Chr.6: 41067359-41067359 on Build GRCh38
rs965776282 Chr.6: 41067551-41067551 on Build GRCh38
rs770594243 Chr.6: 41067307-41067307 on Build GRCh38
rs774383017 Chr.6: 41067453-41067453 on Build GRCh38
rs746965444 Chr.6: 41067392-41067392 on Build GRCh38
rs779073979 Chr.6: 41067370-41067370 on Build GRCh38
rs545345635 Chr.6: 41067151-41067151 on Build GRCh38
rs749762738 Chr.6: 41067320-41067320 on Build GRCh38
rs768257950 Chr.6: 41067298-41067313 on Build GRCh38
rs113683283 Chr.6: 41067593-41067593 on Build GRCh38
rs773520397 Chr.6: 41067443-41067443 on Build GRCh38
rs1041324072 Chr.6: 41067433-41067433 on Build GRCh38
rs769331533 Chr.6: 41067418-41067418 on Build GRCh38
rs969969125 Chr.6: 41067150-41067150 on Build GRCh38
rs770933579 Chr.6: 41067393-41067393 on Build GRCh38
rs551637427 Chr.6: 41067524-41067524 on Build GRCh38
rs960104768 Chr.6: 41067271-41067271 on Build GRCh38
rs766805550 Chr.6: 41067452-41067452 on Build GRCh38
rs764724990 Chr.6: 41067329-41067329 on Build GRCh38
rs527468988 Chr.6: 41067256-41067256 on Build GRCh38
rs984306036 Chr.6: 41067345-41067345 on Build GRCh38
rs762138770 Chr.6: 41067341-41067341 on Build GRCh38
rs377306368 Chr.6: 41067472-41067472 on Build GRCh38
rs1046023373 Chr.6: 41067396-41067396 on Build GRCh38
rs371749521 Chr.6: 41067283-41067283 on Build GRCh38
rs752891997 Chr.6: 41067461-41067461 on Build GRCh38
rs549309208 Chr.6: 41067473-41067473 on Build GRCh38
rs369156425 Chr.6: 41067243-41067243 on Build GRCh38
rs138048940 Chr.6: 41067568-41067568 on Build GRCh38
rs745434047 Chr.6: 41067411-41067411 on Build GRCh38
rs773862839 Chr.6: 41067441-41067441 on Build GRCh38
rs534117319 Chr.6: 41067530-41067530 on Build GRCh38
rs779579661 Chr.6: 41067476-41067476 on Build GRCh38
rs775201701 Chr.6: 41067336-41067336 on Build GRCh38
rs921407628 Chr.6: 41067241-41067241 on Build GRCh38
rs762638487 Chr.6: 41067422-41067422 on Build GRCh38
rs569177361 Chr.6: 41067491-41067491 on Build GRCh38
rs113592386 Chr.6: 41067425-41067425 on Build GRCh38
rs761187398 Chr.6: 41067444-41067444 on Build GRCh38
rs1035643452 Chr.6: 41067555-41067555 on Build GRCh38
rs898950564 Chr.6: 41067484-41067484 on Build GRCh38
rs775051149 Chr.6: 41067419-41067419 on Build GRCh38
rs111574127 Chr.6: 41067558-41067558 on Build GRCh38
rs777581809 Chr.6: 41067386-41067386 on Build GRCh38
rs765339126 Chr.6: 41067459-41067459 on Build GRCh38
rs767417957 Chr.6: 41067441-41067442 on Build GRCh38
rs1009238993 Chr.6: 41067537-41067537 on Build GRCh38
rs756577080 Chr.6: 41067350-41067350 on Build GRCh38
rs761255866 Chr.6: 41067440-41067441 on Build GRCh38
rs1021377551 Chr.6: 41067149-41067149 on Build GRCh38
rs945657788 Chr.6: 41067357-41067357 on Build GRCh38
rs139819793 Chr.6: 41067437-41067437 on Build GRCh38
rs773034083 Chr.6: 41067444-41067445 on Build GRCh38
rs573675949 Chr.6: 41067379-41067379 on Build GRCh38
rs371492851 Chr.6: 41067471-41067471 on Build GRCh38
rs767904715 Chr.6: 41067343-41067343 on Build GRCh38
rs777723136 Chr.6: 41067466-41067466 on Build GRCh38
rs942834095 Chr.6: 41067373-41067373 on Build GRCh38
rs750970469 Chr.6: 41067348-41067348 on Build GRCh38
rs187871594 Chr.6: 41067526-41067526 on Build GRCh38
rs779948050 Chr.6: 41067208-41067208 on Build GRCh38
rs529411165 Chr.6: 41067385-41067385 on Build GRCh38
rs748390599 Chr.6: 41067380-41067380 on Build GRCh38
rs111740039 Chr.6: 41067339-41067339 on Build GRCh38
rs371647903 Chr.6: 41067328-41067328 on Build GRCh38
rs746910477 Chr.6: 41067296-41067296 on Build GRCh38
rs112260512 Chr.6: 41067306-41067306 on Build GRCh38
rs973134412 Chr.6: 41067281-41067281 on Build GRCh38
rs1026143875 Chr.6: 41067509-41067509 on Build GRCh38
rs879044321 Chr.6: 41067352-41067352 on Build GRCh38
rs773083175 Chr.6: 41067285-41067285 on Build GRCh38
rs1053985513 Chr.6: 41067263-41067263 on Build GRCh38

View Full Product Details