Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 41071416-41071957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OARD1
Entrez Gene ID
221443
Gene Name
O-acyl-ADP-ribose deacylase 1
Gene Aliases
C6orf130, TARG1, dJ34B21.3
UniGene
Hs.227457
Gene Chromosome Location
Chr.6: 41066792-41072449 on Build GRCh38
Transcripts
NM_145063.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6888520 c.32G>A p.G11E Chr.6: 41071603-41071603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs893390800 Chr.6: 41071896-41071896 on Build GRCh38
rs962385596 Chr.6: 41071733-41071733 on Build GRCh38
rs911138847 Chr.6: 41071516-41071516 on Build GRCh38
rs776183225 Chr.6: 41071685-41071685 on Build GRCh38
rs962746243 Chr.6: 41071476-41071476 on Build GRCh38
rs111383690 Chr.6: 41071706-41071706 on Build GRCh38
rs930555641 Chr.6: 41071827-41071827 on Build GRCh38
rs763272928 Chr.6: 41071591-41071591 on Build GRCh38
rs920515640 Chr.6: 41071447-41071447 on Build GRCh38
rs950146481 Chr.6: 41071927-41071927 on Build GRCh38
rs766733649 Chr.6: 41071644-41071644 on Build GRCh38
rs969446641 Chr.6: 41071722-41071722 on Build GRCh38
rs370593319 Chr.6: 41071549-41071549 on Build GRCh38
rs1013196839 Chr.6: 41071924-41071924 on Build GRCh38
rs146938733 Chr.6: 41071883-41071883 on Build GRCh38
rs772080242 Chr.6: 41071668-41071668 on Build GRCh38
rs918353071 Chr.6: 41071505-41071505 on Build GRCh38
rs200507230 Chr.6: 41071573-41071573 on Build GRCh38
rs1033230612 Chr.6: 41071523-41071524 on Build GRCh38
rs535838516 Chr.6: 41071693-41071693 on Build GRCh38
rs369741906 Chr.6: 41071544-41071544 on Build GRCh38
rs575041917 Chr.6: 41071832-41071832 on Build GRCh38
rs759331264 Chr.6: 41071600-41071600 on Build GRCh38
rs754069983 Chr.6: 41071656-41071656 on Build GRCh38
rs1024431533 Chr.6: 41071687-41071687 on Build GRCh38
rs377637453 Chr.6: 41071507-41071507 on Build GRCh38
rs746573123 Chr.6: 41071679-41071679 on Build GRCh38
rs971857833 Chr.6: 41071799-41071799 on Build GRCh38
rs563746506 Chr.6: 41071849-41071849 on Build GRCh38
rs961907120 Chr.6: 41071760-41071761 on Build GRCh38
rs925220232 Chr.6: 41071875-41071875 on Build GRCh38
rs149681565 Chr.6: 41071612-41071612 on Build GRCh38
rs148899131 Chr.6: 41071833-41071833 on Build GRCh38
rs546893366 Chr.6: 41071886-41071886 on Build GRCh38
rs1053570556 Chr.6: 41071902-41071902 on Build GRCh38
rs1034766221 Chr.6: 41071756-41071756 on Build GRCh38
rs986591217 Chr.6: 41071784-41071784 on Build GRCh38
rs1025383616 Chr.6: 41071755-41071755 on Build GRCh38
rs1011006542 Chr.6: 41071648-41071648 on Build GRCh38
rs200985692 Chr.6: 41071864-41071864 on Build GRCh38
rs762258563 Chr.6: 41071626-41071626 on Build GRCh38
rs979700471 Chr.6: 41071449-41071449 on Build GRCh38
rs557395792 Chr.6: 41071520-41071520 on Build GRCh38
rs777429290 Chr.6: 41071675-41071675 on Build GRCh38
rs1039018639 Chr.6: 41071528-41071528 on Build GRCh38
rs568612296 Chr.6: 41071501-41071501 on Build GRCh38
rs542091873 Chr.6: 41071742-41071742 on Build GRCh38
rs556128294 Chr.6: 41071844-41071844 on Build GRCh38
rs752037663 Chr.6: 41071604-41071604 on Build GRCh38
rs955338719 Chr.6: 41071766-41071766 on Build GRCh38
rs778733470 Chr.6: 41071660-41071660 on Build GRCh38
rs1016564339 Chr.6: 41071461-41071461 on Build GRCh38
rs972836536 Chr.6: 41071736-41071736 on Build GRCh38
rs933824421 Chr.6: 41071506-41071506 on Build GRCh38
rs114941369 Chr.6: 41071630-41071630 on Build GRCh38
rs913279173 Chr.6: 41071911-41071911 on Build GRCh38
rs756184305 Chr.6: 41071640-41071640 on Build GRCh38
rs972526586 Chr.6: 41071487-41071487 on Build GRCh38
rs750433749 Chr.6: 41071637-41071637 on Build GRCh38
rs944782481 Chr.6: 41071480-41071480 on Build GRCh38
rs911064861 Chr.6: 41071803-41071803 on Build GRCh38
rs561812016 Chr.6: 41071800-41071800 on Build GRCh38
rs3087927 Chr.6: 41071602-41071602 on Build GRCh38
rs748062501 Chr.6: 41071664-41071664 on Build GRCh38
rs573363036 Chr.6: 41071697-41071697 on Build GRCh38
rs773467744 Chr.6: 41071529-41071529 on Build GRCh38
rs375112845 Chr.6: 41071582-41071582 on Build GRCh38
rs780313500 Chr.6: 41071649-41071649 on Build GRCh38
rs375286710 Chr.6: 41071682-41071682 on Build GRCh38
rs776374702 Chr.6: 41071549-41071551 on Build GRCh38
rs115107332 Chr.6: 41071603-41071603 on Build GRCh38
rs367798709 Chr.6: 41071592-41071592 on Build GRCh38
rs942570928 Chr.6: 41071824-41071824 on Build GRCh38
rs41273366 Chr.6: 41071690-41071690 on Build GRCh38
rs1040468122 Chr.6: 41071870-41071870 on Build GRCh38
rs754769713 Chr.6: 41071659-41071659 on Build GRCh38

View Full Product Details