Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152314559-152315059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1 NR_103778.1

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NM_002016.1 NR_103778.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474091 c.226C>T p.L76L Chr.1: 152314660-152314660 on Build GRCh38
377810 c.154G>C p.D52H Chr.1: 152314732-152314732 on Build GRCh38
5881522 c.293G>C p.G98A Chr.1: 152314593-152314593 on Build GRCh38
5607404 c.222T>A p.F74L Chr.1: 152314664-152314664 on Build GRCh38
5041000 c.308A>G p.K103R Chr.1: 152314578-152314578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781611045 Chr.1: 152314759-152314759 on Build GRCh38
rs552075786 Chr.1: 152315025-152315025 on Build GRCh38
rs1048532873 Chr.1: 152314893-152314893 on Build GRCh38
rs565004911 Chr.1: 152314844-152314844 on Build GRCh38
rs989936250 Chr.1: 152314589-152314589 on Build GRCh38
rs148520067 Chr.1: 152314735-152314735 on Build GRCh38
rs367585948 Chr.1: 152314630-152314630 on Build GRCh38
rs931474879 Chr.1: 152314678-152314678 on Build GRCh38
rs769813864 Chr.1: 152314767-152314767 on Build GRCh38
rs769046914 Chr.1: 152314679-152314679 on Build GRCh38
rs201998175 Chr.1: 152314708-152314708 on Build GRCh38
rs190549059 Chr.1: 152314877-152314877 on Build GRCh38
rs780251042 Chr.1: 152314752-152314752 on Build GRCh38
rs998243845 Chr.1: 152314932-152314932 on Build GRCh38
rs753416194 Chr.1: 152314731-152314731 on Build GRCh38
rs370019111 Chr.1: 152314993-152314993 on Build GRCh38
rs199918968 Chr.1: 152314608-152314608 on Build GRCh38
rs749996335 Chr.1: 152314742-152314742 on Build GRCh38
rs1055893727 Chr.1: 152314658-152314658 on Build GRCh38
rs753608358 Chr.1: 152314654-152314654 on Build GRCh38
rs931505931 Chr.1: 152314883-152314883 on Build GRCh38
rs34224823 Chr.1: 152314866-152314866 on Build GRCh38
rs901213894 Chr.1: 152314960-152314960 on Build GRCh38
rs370154185 Chr.1: 152314660-152314660 on Build GRCh38
rs747451295 Chr.1: 152314682-152314682 on Build GRCh38
rs1021814998 Chr.1: 152314762-152314762 on Build GRCh38
rs749328294 Chr.1: 152314685-152314685 on Build GRCh38
rs75611484 Chr.1: 152314837-152314837 on Build GRCh38
rs543139174 Chr.1: 152314768-152314768 on Build GRCh38
rs1056042662 Chr.1: 152314822-152314822 on Build GRCh38
rs755737937 Chr.1: 152314632-152314632 on Build GRCh38
rs115489580 Chr.1: 152314737-152314737 on Build GRCh38
rs145346832 Chr.1: 152314692-152314692 on Build GRCh38
rs759189084 Chr.1: 152314628-152314628 on Build GRCh38
rs1007383691 Chr.1: 152314864-152314864 on Build GRCh38
rs776679330 Chr.1: 152314691-152314691 on Build GRCh38
rs980887460 Chr.1: 152315019-152315019 on Build GRCh38
rs561041384 Chr.1: 152314957-152314957 on Build GRCh38
rs1022537019 Chr.1: 152314964-152314964 on Build GRCh38
rs529787677 Chr.1: 152314962-152314962 on Build GRCh38
rs182426113 Chr.1: 152314966-152314966 on Build GRCh38
rs540838582 Chr.1: 152314758-152314758 on Build GRCh38
rs761432475 Chr.1: 152314799-152314799 on Build GRCh38
rs770009613 Chr.1: 152314693-152314693 on Build GRCh38
rs186922625 Chr.1: 152314976-152314976 on Build GRCh38
rs369354507 Chr.1: 152314756-152314756 on Build GRCh38
rs908634502 Chr.1: 152314939-152314939 on Build GRCh38
rs768672802 Chr.1: 152314601-152314601 on Build GRCh38
rs80252243 Chr.1: 152314599-152314599 on Build GRCh38
rs887696202 Chr.1: 152314849-152314849 on Build GRCh38
rs916151255 Chr.1: 152314879-152314879 on Build GRCh38
rs767524129 Chr.1: 152314629-152314629 on Build GRCh38
rs778369045 Chr.1: 152314663-152314663 on Build GRCh38
rs759758332 Chr.1: 152314773-152314774 on Build GRCh38
rs749437039 Chr.1: 152314771-152314771 on Build GRCh38
rs774514736 Chr.1: 152314688-152314688 on Build GRCh38
rs773267945 Chr.1: 152314696-152314696 on Build GRCh38
rs760681209 Chr.1: 152314717-152314717 on Build GRCh38
rs759395508 Chr.1: 152314698-152314698 on Build GRCh38
rs939127528 Chr.1: 152314784-152314784 on Build GRCh38
rs751384298 Chr.1: 152314614-152314614 on Build GRCh38
rs374663403 Chr.1: 152314591-152314591 on Build GRCh38
rs762018918 Chr.1: 152314603-152314603 on Build GRCh38
rs770755074 Chr.1: 152314827-152314827 on Build GRCh38
rs113543010 Chr.1: 152314797-152314797 on Build GRCh38
rs926822287 Chr.1: 152315026-152315026 on Build GRCh38
rs111373725 Chr.1: 152314942-152314942 on Build GRCh38
rs760493006 Chr.1: 152314782-152314782 on Build GRCh38
rs1017475005 Chr.1: 152314866-152314866 on Build GRCh38
rs76438926 Chr.1: 152314602-152314602 on Build GRCh38
rs3126082 Chr.1: 152314831-152314831 on Build GRCh38
rs748256354 Chr.1: 152314761-152314761 on Build GRCh38
rs397864332 Chr.1: 152314877-152314877 on Build GRCh38
rs780631848 Chr.1: 152314681-152314681 on Build GRCh38
rs763759026 Chr.1: 152314721-152314721 on Build GRCh38
rs767191408 Chr.1: 152314701-152314701 on Build GRCh38
rs775791704 Chr.1: 152314587-152314587 on Build GRCh38
rs901764347 Chr.1: 152314736-152314736 on Build GRCh38
rs758847783 Chr.1: 152314747-152314747 on Build GRCh38
rs764135477 Chr.1: 152314786-152314786 on Build GRCh38
rs372637471 Chr.1: 152314789-152314789 on Build GRCh38
rs993586993 Chr.1: 152314834-152314834 on Build GRCh38
rs1028911515 Chr.1: 152314750-152314750 on Build GRCh38
rs763670992 Chr.1: 152314646-152314646 on Build GRCh38
rs758648098 Chr.1: 152314680-152314680 on Build GRCh38
rs577436533 Chr.1: 152314990-152314990 on Build GRCh38
rs1041445745 Chr.1: 152314952-152314952 on Build GRCh38
rs556436445 Chr.1: 152314775-152314775 on Build GRCh38
rs964155866 Chr.1: 152314667-152314667 on Build GRCh38
rs746246378 Chr.1: 152314671-152314671 on Build GRCh38

View Full Product Details