Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152348614-152349135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

SNP Detail

SNP ID Chromosome Location
rs901454711 Chr.1: 152349108-152349108 on Build GRCh38
rs948521721 Chr.1: 152348857-152348857 on Build GRCh38
rs879135352 Chr.1: 152348921-152348921 on Build GRCh38
rs145057374 Chr.1: 152348762-152348762 on Build GRCh38
rs897850510 Chr.1: 152348938-152348938 on Build GRCh38
rs879051145 Chr.1: 152348769-152348769 on Build GRCh38
rs913913141 Chr.1: 152348829-152348829 on Build GRCh38
rs955939700 Chr.1: 152348810-152348810 on Build GRCh38
rs367545860 Chr.1: 152348983-152348983 on Build GRCh38
rs1007742713 Chr.1: 152348740-152348740 on Build GRCh38
rs372037751 Chr.1: 152348718-152348718 on Build GRCh38
rs374786608 Chr.1: 152348982-152348982 on Build GRCh38
rs1027804569 Chr.1: 152348692-152348692 on Build GRCh38
rs993777221 Chr.1: 152348690-152348690 on Build GRCh38
rs140262842 Chr.1: 152348961-152348961 on Build GRCh38
rs534165664 Chr.1: 152348942-152348942 on Build GRCh38
rs778042176 Chr.1: 152348697-152348697 on Build GRCh38
rs1019909533 Chr.1: 152348687-152348687 on Build GRCh38
rs928581087 Chr.1: 152348879-152348879 on Build GRCh38
rs147146061 Chr.1: 152348939-152348939 on Build GRCh38
rs747373359 Chr.1: 152348713-152348713 on Build GRCh38
rs1031752416 Chr.1: 152349068-152349068 on Build GRCh38
rs12043049 Chr.1: 152348924-152348924 on Build GRCh38
rs183767339 Chr.1: 152349036-152349036 on Build GRCh38
rs1049309961 Chr.1: 152348943-152348943 on Build GRCh38
rs2065954 Chr.1: 152348868-152348868 on Build GRCh38
rs534388172 Chr.1: 152348773-152348773 on Build GRCh38
rs1056483939 Chr.1: 152348650-152348650 on Build GRCh38
rs558784400 Chr.1: 152348808-152348808 on Build GRCh38
rs1004474683 Chr.1: 152348663-152348663 on Build GRCh38
rs569278777 Chr.1: 152348775-152348775 on Build GRCh38
rs1048138037 Chr.1: 152348990-152348990 on Build GRCh38
rs979962100 Chr.1: 152348865-152348865 on Build GRCh38
rs939462103 Chr.1: 152348881-152348881 on Build GRCh38
rs760051516 Chr.1: 152348954-152348954 on Build GRCh38
rs1017785385 Chr.1: 152349009-152349010 on Build GRCh38
rs989933412 Chr.1: 152348815-152348815 on Build GRCh38
rs966716148 Chr.1: 152348841-152348841 on Build GRCh38
rs898040726 Chr.1: 152348677-152348677 on Build GRCh38
rs976126277 Chr.1: 152348792-152348792 on Build GRCh38
rs4550048 Chr.1: 152348845-152348845 on Build GRCh38
rs529917712 Chr.1: 152348755-152348755 on Build GRCh38
rs886733956 Chr.1: 152348642-152348642 on Build GRCh38
rs887981164 Chr.1: 152348722-152348722 on Build GRCh38
rs1040714076 Chr.1: 152349055-152349055 on Build GRCh38
rs574412094 Chr.1: 152349020-152349020 on Build GRCh38

View Full Product Details