Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152352632-152353129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG-AS1
Entrez Gene ID
339400
Gene Name
FLG antisense RNA 1
Gene Aliases
-
UniGene
Hs.23783
Gene Chromosome Location
Chr.1: 152313459-152366692 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Gene Symbol
FLG2
Entrez Gene ID
388698
Gene Name
filaggrin family member 2
Gene Aliases
IFPS
UniGene
Hs.156124
Gene Chromosome Location
Chr.1: 152348735-152360028 on Build GRCh38
Transcripts
NR_103778.1 NR_103779.1 NM_001014342.2 XM_011509531.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3474123 c.4876G>A p.D1626N Chr.1: 152352910-152352910 on Build GRCh38
260956 c.5073A>G p.R1691R Chr.1: 152352713-152352713 on Build GRCh38
7368399 c.5021A>C p.H1674P Chr.1: 152352765-152352765 on Build GRCh38
3474125 c.4784G>A p.R1595Q Chr.1: 152353002-152353002 on Build GRCh38
3671400 c.4774C>A p.P1592T Chr.1: 152353012-152353012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756477942 Chr.1: 152352885-152352885 on Build GRCh38
rs751052229 Chr.1: 152352862-152352862 on Build GRCh38
rs756082008 Chr.1: 152352863-152352863 on Build GRCh38
rs760412490 Chr.1: 152352681-152352681 on Build GRCh38
rs777050203 Chr.1: 152353067-152353067 on Build GRCh38
rs756857022 Chr.1: 152352691-152352691 on Build GRCh38
rs778979548 Chr.1: 152352897-152352897 on Build GRCh38
rs749120671 Chr.1: 152353022-152353023 on Build GRCh38
rs199839343 Chr.1: 152352935-152352935 on Build GRCh38
rs754337153 Chr.1: 152352893-152352893 on Build GRCh38
rs769645229 Chr.1: 152352837-152352837 on Build GRCh38
rs747252009 Chr.1: 152352993-152352993 on Build GRCh38
rs188198910 Chr.1: 152352834-152352834 on Build GRCh38
rs774405765 Chr.1: 152353005-152353005 on Build GRCh38
rs780182557 Chr.1: 152352988-152352988 on Build GRCh38
rs758485650 Chr.1: 152352981-152352981 on Build GRCh38
rs775177169 Chr.1: 152352943-152352943 on Build GRCh38
rs757083866 Chr.1: 152353098-152353098 on Build GRCh38
rs1039295082 Chr.1: 152352793-152352793 on Build GRCh38
rs749934736 Chr.1: 152352767-152352767 on Build GRCh38
rs200981503 Chr.1: 152353077-152353077 on Build GRCh38
rs758650635 Chr.1: 152352902-152352902 on Build GRCh38
rs773093326 Chr.1: 152352915-152352915 on Build GRCh38
rs777339480 Chr.1: 152352743-152352744 on Build GRCh38
rs779058950 Chr.1: 152353099-152353099 on Build GRCh38
rs1033522882 Chr.1: 152352689-152352689 on Build GRCh38
rs749375907 Chr.1: 152352815-152352815 on Build GRCh38
rs73007787 Chr.1: 152352980-152352980 on Build GRCh38
rs757767944 Chr.1: 152352973-152352973 on Build GRCh38
rs751228005 Chr.1: 152352769-152352769 on Build GRCh38
rs199599567 Chr.1: 152352664-152352664 on Build GRCh38
rs765155173 Chr.1: 152352766-152352766 on Build GRCh38
rs150364809 Chr.1: 152352892-152352892 on Build GRCh38
rs1045780308 Chr.1: 152352777-152352777 on Build GRCh38
rs151207521 Chr.1: 152352765-152352765 on Build GRCh38
rs777715863 Chr.1: 152352737-152352737 on Build GRCh38
rs765986373 Chr.1: 152352859-152352859 on Build GRCh38
rs767154074 Chr.1: 152352864-152352864 on Build GRCh38
rs755256022 Chr.1: 152353079-152353079 on Build GRCh38
rs761012281 Chr.1: 152352951-152352951 on Build GRCh38
rs987204431 Chr.1: 152353031-152353031 on Build GRCh38
rs950538621 Chr.1: 152353009-152353009 on Build GRCh38
rs368471567 Chr.1: 152352677-152352677 on Build GRCh38
rs1000774803 Chr.1: 152353016-152353016 on Build GRCh38
rs763587904 Chr.1: 152353038-152353038 on Build GRCh38
rs139512884 Chr.1: 152353070-152353070 on Build GRCh38
rs746075054 Chr.1: 152352825-152352825 on Build GRCh38
rs973231523 Chr.1: 152352692-152352692 on Build GRCh38
rs764067255 Chr.1: 152352690-152352690 on Build GRCh38
rs747946138 Chr.1: 152352907-152352907 on Build GRCh38
rs754123597 Chr.1: 152352698-152352698 on Build GRCh38
rs771163168 Chr.1: 152352741-152352741 on Build GRCh38
rs769605210 Chr.1: 152352908-152352908 on Build GRCh38
rs376175147 Chr.1: 152353011-152353011 on Build GRCh38
rs957682383 Chr.1: 152352747-152352747 on Build GRCh38
rs755533237 Chr.1: 152352716-152352716 on Build GRCh38
rs774534850 Chr.1: 152352753-152352753 on Build GRCh38
rs987556345 Chr.1: 152352748-152352748 on Build GRCh38
rs974360389 Chr.1: 152353105-152353105 on Build GRCh38
rs767121785 Chr.1: 152352937-152352937 on Build GRCh38
rs564525196 Chr.1: 152352804-152352804 on Build GRCh38
rs762594513 Chr.1: 152352838-152352838 on Build GRCh38
rs757992022 Chr.1: 152352700-152352700 on Build GRCh38
rs781511680 Chr.1: 152353093-152353093 on Build GRCh38
rs1020806897 Chr.1: 152352660-152352660 on Build GRCh38
rs779816742 Chr.1: 152352705-152352705 on Build GRCh38
rs747254831 Chr.1: 152352989-152352989 on Build GRCh38
rs760502199 Chr.1: 152352756-152352756 on Build GRCh38
rs779172100 Chr.1: 152352819-152352819 on Build GRCh38
rs771605398 Chr.1: 152353014-152353014 on Build GRCh38
rs768435002 Chr.1: 152352764-152352764 on Build GRCh38
rs774027082 Chr.1: 152352934-152352934 on Build GRCh38
rs760388364 Chr.1: 152353035-152353035 on Build GRCh38
rs141953432 Chr.1: 152353030-152353030 on Build GRCh38
rs750900984 Chr.1: 152353061-152353061 on Build GRCh38
rs771013080 Chr.1: 152353003-152353003 on Build GRCh38
rs140553142 Chr.1: 152352906-152352906 on Build GRCh38
rs756606921 Chr.1: 152352801-152352801 on Build GRCh38
rs1032581380 Chr.1: 152353028-152353028 on Build GRCh38
rs751998573 Chr.1: 152352710-152352710 on Build GRCh38
rs551341511 Chr.1: 152352791-152352791 on Build GRCh38
rs753022760 Chr.1: 152352870-152352870 on Build GRCh38
rs757383528 Chr.1: 152352896-152352896 on Build GRCh38
rs776296902 Chr.1: 152352835-152352835 on Build GRCh38
rs777012604 Chr.1: 152353043-152353043 on Build GRCh38
rs745802989 Chr.1: 152353101-152353101 on Build GRCh38
rs755923979 Chr.1: 152352992-152352992 on Build GRCh38
rs754143018 Chr.1: 152352972-152352972 on Build GRCh38
rs763095519 Chr.1: 152352653-152352653 on Build GRCh38
rs770274998 Chr.1: 152352729-152352729 on Build GRCh38
rs752892733 Chr.1: 152352774-152352774 on Build GRCh38
rs74127928 Chr.1: 152352979-152352979 on Build GRCh38
rs753732309 Chr.1: 152353095-152353095 on Build GRCh38
rs770966726 Chr.1: 152352816-152352816 on Build GRCh38
rs745883660 Chr.1: 152352898-152352898 on Build GRCh38
rs749344747 Chr.1: 152352740-152352740 on Build GRCh38
rs781775435 Chr.1: 152352727-152352727 on Build GRCh38
rs749228510 Chr.1: 152352930-152352930 on Build GRCh38
rs905831995 Chr.1: 152352968-152352968 on Build GRCh38
rs764641471 Chr.1: 152352695-152352695 on Build GRCh38
rs764504174 Chr.1: 152352953-152352953 on Build GRCh38
rs267598033 Chr.1: 152352742-152352742 on Build GRCh38
rs575700744 Chr.1: 152352997-152352997 on Build GRCh38
rs755374237 Chr.1: 152352719-152352719 on Build GRCh38
rs201802558 Chr.1: 152353002-152353002 on Build GRCh38
rs544813438 Chr.1: 152352802-152352802 on Build GRCh38
rs765784034 Chr.1: 152353048-152353048 on Build GRCh38
rs142337570 Chr.1: 152353046-152353046 on Build GRCh38

View Full Product Details