Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153807995-153808537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATAD2B
Entrez Gene ID
57459
Gene Name
GATA zinc finger domain containing 2B
Gene Aliases
MRD18, P66beta, p68
UniGene
Hs.4779
Gene Chromosome Location
Chr.1: 153804725-153922975 on Build GRCh38
Transcripts
XM_005245364.4

SNP Detail

SNP ID Chromosome Location
rs896404887 Chr.1: 153808092-153808092 on Build GRCh38
rs567897881 Chr.1: 153808156-153808156 on Build GRCh38
rs917718762 Chr.1: 153808480-153808480 on Build GRCh38
rs760035749 Chr.1: 153808069-153808069 on Build GRCh38
rs1006087426 Chr.1: 153808501-153808501 on Build GRCh38
rs964126447 Chr.1: 153808350-153808350 on Build GRCh38
rs979383449 Chr.1: 153808356-153808356 on Build GRCh38
rs935239630 Chr.1: 153808372-153808372 on Build GRCh38
rs988967713 Chr.1: 153808357-153808357 on Build GRCh38
rs1056158982 Chr.1: 153808390-153808390 on Build GRCh38
rs923486654 Chr.1: 153808341-153808341 on Build GRCh38
rs942714419 Chr.1: 153808062-153808062 on Build GRCh38
rs911309361 Chr.1: 153808323-153808323 on Build GRCh38
rs771697814 Chr.1: 153808088-153808088 on Build GRCh38
rs947606317 Chr.1: 153808366-153808366 on Build GRCh38
rs539410481 Chr.1: 153808288-153808288 on Build GRCh38
rs1052216023 Chr.1: 153808116-153808116 on Build GRCh38
rs755387277 Chr.1: 153808143-153808143 on Build GRCh38
rs549156846 Chr.1: 153808054-153808054 on Build GRCh38
rs571864729 Chr.1: 153808179-153808179 on Build GRCh38
rs547233449 Chr.1: 153808166-153808166 on Build GRCh38
rs911187559 Chr.1: 153808047-153808047 on Build GRCh38
rs1034899577 Chr.1: 153808089-153808089 on Build GRCh38
rs935272988 Chr.1: 153808115-153808115 on Build GRCh38
rs986881402 Chr.1: 153808309-153808309 on Build GRCh38
rs528684210 Chr.1: 153808157-153808157 on Build GRCh38
rs1052312271 Chr.1: 153808409-153808409 on Build GRCh38
rs1013302378 Chr.1: 153808123-153808123 on Build GRCh38
rs896343303 Chr.1: 153808118-153808118 on Build GRCh38
rs955733739 Chr.1: 153808305-153808305 on Build GRCh38
rs994457665 Chr.1: 153808280-153808280 on Build GRCh38
rs964675492 Chr.1: 153808258-153808258 on Build GRCh38
rs1043855693 Chr.1: 153808374-153808374 on Build GRCh38
rs1045216528 Chr.1: 153808125-153808125 on Build GRCh38
rs904831105 Chr.1: 153808153-153808153 on Build GRCh38
rs779219112 Chr.1: 153808310-153808310 on Build GRCh38
rs754296207 Chr.1: 153808110-153808110 on Build GRCh38
rs904948322 Chr.1: 153808387-153808387 on Build GRCh38
rs896373925 Chr.1: 153808408-153808408 on Build GRCh38
rs1001901930 Chr.1: 153808060-153808061 on Build GRCh38
rs1017682517 Chr.1: 153808247-153808247 on Build GRCh38
rs925325304 Chr.1: 153808367-153808367 on Build GRCh38
rs368396933 Chr.1: 153808315-153808315 on Build GRCh38
rs937620096 Chr.1: 153808388-153808388 on Build GRCh38
rs887604526 Chr.1: 153808483-153808487 on Build GRCh38
rs962956070 Chr.1: 153808202-153808202 on Build GRCh38
rs569750010 Chr.1: 153808406-153808406 on Build GRCh38
rs1017610331 Chr.1: 153808504-153808504 on Build GRCh38
rs987300371 Chr.1: 153808032-153808032 on Build GRCh38
rs934084315 Chr.1: 153808030-153808030 on Build GRCh38
rs557812401 Chr.1: 153808313-153808313 on Build GRCh38

View Full Product Details