Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153930454-153930955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4B
Entrez Gene ID
9909
Gene Name
DENN domain containing 4B
Gene Aliases
KIAA0476
UniGene
Hs.744878
Gene Chromosome Location
Chr.1: 153929501-153946681 on Build GRCh38
Transcripts
NM_014856.2 XM_005245678.4 XM_005245679.2 XM_011510219.1 XM_011510220.2 XM_011510221.2 XM_017002993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1195151 c.3956G>A p.R1319H Chr.1: 153930592-153930592 on Build GRCh38
7211888 c.3897C>T p.L1299L Chr.1: 153930739-153930739 on Build GRCh38
6351201 c.3975A>G p.T1325T Chr.1: 153930573-153930573 on Build GRCh38
6642428 c.3955C>T p.R1319C Chr.1: 153930593-153930593 on Build GRCh38
6642427 c.4291C>T p.R1431C Chr.1: 153930593-153930593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs967468008 Chr.1: 153930716-153930716 on Build GRCh38
rs747128049 Chr.1: 153930626-153930629 on Build GRCh38
rs376544331 Chr.1: 153930800-153930800 on Build GRCh38
rs770098919 Chr.1: 153930721-153930721 on Build GRCh38
rs756983556 Chr.1: 153930654-153930654 on Build GRCh38
rs761366115 Chr.1: 153930816-153930816 on Build GRCh38
rs763953270 Chr.1: 153930639-153930639 on Build GRCh38
rs375119207 Chr.1: 153930521-153930521 on Build GRCh38
rs372135633 Chr.1: 153930478-153930478 on Build GRCh38
rs1014890098 Chr.1: 153930708-153930708 on Build GRCh38
rs749527610 Chr.1: 153930902-153930902 on Build GRCh38
rs775503712 Chr.1: 153930780-153930780 on Build GRCh38
rs201557402 Chr.1: 153930525-153930525 on Build GRCh38
rs781633857 Chr.1: 153930699-153930699 on Build GRCh38
rs373864007 Chr.1: 153930684-153930684 on Build GRCh38
rs752010921 Chr.1: 153930850-153930850 on Build GRCh38
rs373354599 Chr.1: 153930722-153930722 on Build GRCh38
rs201317481 Chr.1: 153930479-153930479 on Build GRCh38
rs201922757 Chr.1: 153930841-153930841 on Build GRCh38
rs753706326 Chr.1: 153930807-153930807 on Build GRCh38
rs778254667 Chr.1: 153930655-153930655 on Build GRCh38
rs368266568 Chr.1: 153930477-153930477 on Build GRCh38
rs780167408 Chr.1: 153930511-153930513 on Build GRCh38
rs375939242 Chr.1: 153930879-153930879 on Build GRCh38
rs768859684 Chr.1: 153930689-153930689 on Build GRCh38
rs928642716 Chr.1: 153930873-153930873 on Build GRCh38
rs886840412 Chr.1: 153930585-153930585 on Build GRCh38
rs995307056 Chr.1: 153930555-153930555 on Build GRCh38
rs748299957 Chr.1: 153930714-153930714 on Build GRCh38
rs549408887 Chr.1: 153930757-153930757 on Build GRCh38
rs752525614 Chr.1: 153930627-153930627 on Build GRCh38
rs768980561 Chr.1: 153930662-153930662 on Build GRCh38
rs377296406 Chr.1: 153930650-153930650 on Build GRCh38
rs55874907 Chr.1: 153930653-153930653 on Build GRCh38
rs943692438 Chr.1: 153930544-153930544 on Build GRCh38
rs773471488 Chr.1: 153930618-153930618 on Build GRCh38
rs1018437069 Chr.1: 153930918-153930918 on Build GRCh38
rs965049029 Chr.1: 153930631-153930631 on Build GRCh38
rs769081956 Chr.1: 153930587-153930587 on Build GRCh38
rs777229163 Chr.1: 153930672-153930672 on Build GRCh38
rs764926061 Chr.1: 153930822-153930822 on Build GRCh38
rs879274651 Chr.1: 153930527-153930527 on Build GRCh38
rs769897190 Chr.1: 153930881-153930881 on Build GRCh38
rs370247245 Chr.1: 153930922-153930922 on Build GRCh38
rs759145171 Chr.1: 153930511-153930511 on Build GRCh38
rs760699475 Chr.1: 153930783-153930783 on Build GRCh38
rs746353179 Chr.1: 153930536-153930536 on Build GRCh38
rs369439497 Chr.1: 153930827-153930827 on Build GRCh38
rs771150123 Chr.1: 153930777-153930777 on Build GRCh38
rs755553463 Chr.1: 153930860-153930860 on Build GRCh38
rs755821699 Chr.1: 153930518-153930518 on Build GRCh38
rs2297897 Chr.1: 153930573-153930573 on Build GRCh38
rs762129698 Chr.1: 153930607-153930607 on Build GRCh38
rs774595759 Chr.1: 153930913-153930913 on Build GRCh38
rs765187119 Chr.1: 153930609-153930609 on Build GRCh38
rs938565359 Chr.1: 153930495-153930495 on Build GRCh38
rs561754834 Chr.1: 153930796-153930796 on Build GRCh38
rs373474714 Chr.1: 153930842-153930842 on Build GRCh38
rs1004395072 Chr.1: 153930586-153930586 on Build GRCh38
rs771095154 Chr.1: 153930903-153930903 on Build GRCh38
rs914742010 Chr.1: 153930709-153930709 on Build GRCh38
rs750088015 Chr.1: 153930825-153930825 on Build GRCh38
rs915804176 Chr.1: 153930915-153930915 on Build GRCh38
rs767394823 Chr.1: 153930515-153930515 on Build GRCh38
rs778085882 Chr.1: 153930892-153930892 on Build GRCh38
rs755914043 Chr.1: 153930634-153930634 on Build GRCh38
rs748615683 Chr.1: 153930880-153930880 on Build GRCh38
rs759511767 Chr.1: 153930621-153930621 on Build GRCh38
rs367842375 Chr.1: 153930549-153930549 on Build GRCh38
rs1027883236 Chr.1: 153930810-153930810 on Build GRCh38
rs531277377 Chr.1: 153930679-153930679 on Build GRCh38
rs758891726 Chr.1: 153930663-153930663 on Build GRCh38
rs764764155 Chr.1: 153930531-153930531 on Build GRCh38
rs760498000 Chr.1: 153930921-153930921 on Build GRCh38
rs1012110960 Chr.1: 153930629-153930629 on Build GRCh38
rs577830224 Chr.1: 153930532-153930532 on Build GRCh38
rs897840491 Chr.1: 153930721-153930721 on Build GRCh38
rs767627604 Chr.1: 153930623-153930623 on Build GRCh38
rs776653329 Chr.1: 153930597-153930597 on Build GRCh38
rs745448760 Chr.1: 153930662-153930662 on Build GRCh38
rs1055813617 Chr.1: 153930911-153930911 on Build GRCh38

View Full Product Details