Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153932746-153933251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DENND4B
Entrez Gene ID
9909
Gene Name
DENN domain containing 4B
Gene Aliases
KIAA0476
UniGene
Hs.744878
Gene Chromosome Location
Chr.1: 153929501-153946681 on Build GRCh38
Transcripts
NM_014856.2 XM_005245678.4 XM_005245679.2 XM_011510219.1 XM_011510220.2 XM_011510221.2 XM_017002993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
896445 c.3158C>T p.S1053L Chr.1: 153932990-153932990 on Build GRCh38
1195182 c.3281G>A p.R1094Q Chr.1: 153932867-153932867 on Build GRCh38
3474530 c.3137C>T p.S1046F Chr.1: 153933011-153933011 on Build GRCh38
7222096 c.3508G>A p.E1170K Chr.1: 153932976-153932976 on Build GRCh38
3801958 c.3227C>T p.P1076L Chr.1: 153932921-153932921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914577201 Chr.1: 153933142-153933145 on Build GRCh38
rs916272298 Chr.1: 153933212-153933212 on Build GRCh38
rs563201689 Chr.1: 153932803-153932803 on Build GRCh38
rs200973111 Chr.1: 153933070-153933070 on Build GRCh38
rs751552787 Chr.1: 153933041-153933041 on Build GRCh38
rs1030716451 Chr.1: 153933175-153933175 on Build GRCh38
rs768986209 Chr.1: 153933185-153933185 on Build GRCh38
rs967947625 Chr.1: 153933103-153933103 on Build GRCh38
rs764056629 Chr.1: 153932994-153932994 on Build GRCh38
rs371400116 Chr.1: 153933062-153933062 on Build GRCh38
rs746290102 Chr.1: 153932950-153932950 on Build GRCh38
rs551077275 Chr.1: 153933113-153933113 on Build GRCh38
rs978334002 Chr.1: 153933170-153933170 on Build GRCh38
rs1044938665 Chr.1: 153932935-153932935 on Build GRCh38
rs375614366 Chr.1: 153933146-153933146 on Build GRCh38
rs763897068 Chr.1: 153932905-153932905 on Build GRCh38
rs774694318 Chr.1: 153932949-153932949 on Build GRCh38
rs771469533 Chr.1: 153932943-153932943 on Build GRCh38
rs775535143 Chr.1: 153932792-153932792 on Build GRCh38
rs368196022 Chr.1: 153932870-153932870 on Build GRCh38
rs991799685 Chr.1: 153933206-153933206 on Build GRCh38
rs766478095 Chr.1: 153933038-153933038 on Build GRCh38
rs752244422 Chr.1: 153933049-153933049 on Build GRCh38
rs200058194 Chr.1: 153932774-153932774 on Build GRCh38
rs1056149377 Chr.1: 153932838-153932838 on Build GRCh38
rs757749443 Chr.1: 153933067-153933067 on Build GRCh38
rs998106311 Chr.1: 153933039-153933039 on Build GRCh38
rs1022542478 Chr.1: 153933088-153933088 on Build GRCh38
rs888649183 Chr.1: 153932945-153932945 on Build GRCh38
rs764265099 Chr.1: 153932811-153932811 on Build GRCh38
rs375041368 Chr.1: 153933002-153933002 on Build GRCh38
rs755543682 Chr.1: 153932920-153932920 on Build GRCh38
rs753456800 Chr.1: 153933063-153933063 on Build GRCh38
rs538623653 Chr.1: 153933071-153933071 on Build GRCh38
rs777367578 Chr.1: 153932926-153932926 on Build GRCh38
rs755848120 Chr.1: 153933053-153933053 on Build GRCh38
rs370715705 Chr.1: 153932989-153932989 on Build GRCh38
rs201848699 Chr.1: 153933048-153933048 on Build GRCh38
rs917476815 Chr.1: 153932880-153932880 on Build GRCh38
rs896136715 Chr.1: 153932847-153932847 on Build GRCh38
rs1538641 Chr.1: 153933099-153933099 on Build GRCh38
rs774395308 Chr.1: 153933202-153933202 on Build GRCh38
rs191677069 Chr.1: 153932800-153932800 on Build GRCh38
rs753835933 Chr.1: 153933034-153933035 on Build GRCh38
rs780515115 Chr.1: 153933179-153933179 on Build GRCh38
rs1122186 Chr.1: 153932990-153932990 on Build GRCh38
rs776557290 Chr.1: 153933188-153933188 on Build GRCh38
rs760235216 Chr.1: 153932844-153932846 on Build GRCh38
rs762792313 Chr.1: 153933037-153933037 on Build GRCh38
rs367833186 Chr.1: 153933003-153933003 on Build GRCh38
rs199987020 Chr.1: 153932914-153932914 on Build GRCh38
rs749739721 Chr.1: 153932927-153932927 on Build GRCh38
rs747167311 Chr.1: 153933183-153933183 on Build GRCh38
rs867778095 Chr.1: 153932898-153932898 on Build GRCh38
rs767966211 Chr.1: 153933098-153933098 on Build GRCh38
rs769996084 Chr.1: 153933196-153933196 on Build GRCh38
rs569210097 Chr.1: 153933147-153933147 on Build GRCh38
rs759572971 Chr.1: 153933203-153933203 on Build GRCh38
rs772429688 Chr.1: 153932980-153932980 on Build GRCh38
rs1538640 Chr.1: 153933083-153933083 on Build GRCh38
rs376949568 Chr.1: 153932815-153932815 on Build GRCh38
rs368610454 Chr.1: 153933153-153933153 on Build GRCh38
rs375597225 Chr.1: 153932867-153932867 on Build GRCh38
rs147284413 Chr.1: 153932819-153932819 on Build GRCh38
rs754659759 Chr.1: 153932868-153932868 on Build GRCh38
rs766242197 Chr.1: 153932837-153932837 on Build GRCh38
rs566126367 Chr.1: 153932794-153932794 on Build GRCh38
rs187284284 Chr.1: 153932783-153932783 on Build GRCh38
rs761922514 Chr.1: 153933189-153933189 on Build GRCh38

View Full Product Details