Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153952484-153952893 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRTC2
Entrez Gene ID
200186
Gene Name
CREB regulated transcription coactivator 2
Gene Aliases
TORC-2, TORC2
UniGene
Hs.406392
Gene Chromosome Location
Chr.1: 153947672-153958656 on Build GRCh38
Transcripts
NM_181715.2 XM_005244946.1 XM_005244949.2 XM_017000575.1 XM_017000576.1 XM_017000577.1 XM_017000578.1 XM_017000579.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3376736 c.638-4C>T p.? Chr.1: 153952639-153952639 on Build GRCh38
4430137 c.694G>A p.A232T Chr.1: 153952579-153952579 on Build GRCh38
896469 c.641C>T p.P214L Chr.1: 153952632-153952632 on Build GRCh38
122368 c.699G>C p.K233N Chr.1: 153952574-153952574 on Build GRCh38
3418091 c.657C>A p.N219K Chr.1: 153952616-153952616 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749709858 Chr.1: 153952802-153952802 on Build GRCh38
rs952846857 Chr.1: 153952526-153952526 on Build GRCh38
rs781354022 Chr.1: 153952540-153952540 on Build GRCh38
rs186634744 Chr.1: 153952669-153952669 on Build GRCh38
rs1018347381 Chr.1: 153952864-153952864 on Build GRCh38
rs747872123 Chr.1: 153952801-153952801 on Build GRCh38
rs757479190 Chr.1: 153952776-153952776 on Build GRCh38
rs772994822 Chr.1: 153952832-153952832 on Build GRCh38
rs752974063 Chr.1: 153952664-153952664 on Build GRCh38
rs1050752488 Chr.1: 153952623-153952623 on Build GRCh38
rs959952131 Chr.1: 153952743-153952743 on Build GRCh38
rs61804848 Chr.1: 153952604-153952604 on Build GRCh38
rs1035460327 Chr.1: 153952737-153952737 on Build GRCh38
rs776237648 Chr.1: 153952575-153952575 on Build GRCh38
rs543689284 Chr.1: 153952630-153952630 on Build GRCh38
rs750147849 Chr.1: 153952606-153952606 on Build GRCh38
rs778903807 Chr.1: 153952782-153952782 on Build GRCh38
rs200473751 Chr.1: 153952788-153952788 on Build GRCh38
rs764763783 Chr.1: 153952600-153952600 on Build GRCh38
rs889069446 Chr.1: 153952629-153952629 on Build GRCh38
rs756426576 Chr.1: 153952726-153952726 on Build GRCh38
rs777013796 Chr.1: 153952560-153952562 on Build GRCh38
rs781739093 Chr.1: 153952653-153952653 on Build GRCh38
rs758020239 Chr.1: 153952523-153952523 on Build GRCh38
rs112474121 Chr.1: 153952660-153952660 on Build GRCh38
rs113190401 Chr.1: 153952661-153952661 on Build GRCh38
rs755391766 Chr.1: 153952639-153952639 on Build GRCh38
rs969665797 Chr.1: 153952766-153952766 on Build GRCh38
rs775406065 Chr.1: 153952559-153952559 on Build GRCh38
rs761927721 Chr.1: 153952619-153952621 on Build GRCh38
rs762537967 Chr.1: 153952616-153952616 on Build GRCh38
rs762627230 Chr.1: 153952860-153952860 on Build GRCh38
rs777961219 Chr.1: 153952685-153952685 on Build GRCh38
rs201092523 Chr.1: 153952555-153952555 on Build GRCh38
rs1011357551 Chr.1: 153952645-153952645 on Build GRCh38
rs531910512 Chr.1: 153952578-153952578 on Build GRCh38
rs915365226 Chr.1: 153952676-153952676 on Build GRCh38
rs903029218 Chr.1: 153952697-153952697 on Build GRCh38
rs897094789 Chr.1: 153952785-153952785 on Build GRCh38
rs771060705 Chr.1: 153952556-153952556 on Build GRCh38
rs985041099 Chr.1: 153952772-153952772 on Build GRCh38
rs181685527 Chr.1: 153952528-153952528 on Build GRCh38
rs1006675081 Chr.1: 153952810-153952810 on Build GRCh38
rs769868515 Chr.1: 153952552-153952552 on Build GRCh38
rs370583662 Chr.1: 153952561-153952561 on Build GRCh38
rs191461887 Chr.1: 153952750-153952750 on Build GRCh38
rs138937027 Chr.1: 153952574-153952574 on Build GRCh38
rs529627884 Chr.1: 153952709-153952709 on Build GRCh38
rs201851117 Chr.1: 153952542-153952542 on Build GRCh38
rs1024298258 Chr.1: 153952760-153952760 on Build GRCh38
rs765269194 Chr.1: 153952837-153952838 on Build GRCh38
rs376899918 Chr.1: 153952690-153952690 on Build GRCh38
rs776667899 Chr.1: 153952798-153952798 on Build GRCh38
rs755388570 Chr.1: 153952533-153952533 on Build GRCh38
rs780683630 Chr.1: 153952527-153952527 on Build GRCh38
rs769687063 Chr.1: 153952807-153952807 on Build GRCh38
rs200315514 Chr.1: 153952554-153952554 on Build GRCh38
rs745981936 Chr.1: 153952707-153952707 on Build GRCh38
rs761696504 Chr.1: 153952592-153952592 on Build GRCh38

View Full Product Details