Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153967758-153968262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB3L4
Entrez Gene ID
148327
Gene Name
cAMP responsive element binding protein 3 like 4
Gene Aliases
AIBZIP, ATCE1, CREB3, CREB4, JAL, hJAL
UniGene
Hs.372924
Gene Chromosome Location
Chr.1: 153967679-153974364 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255979.1 NM_001255980.1 NM_001255981.1 NM_130898.3 NR_045658.1 XM_006711172.2 XM_017000372.1 NM_001271960.1

Gene Symbol
SLC39A1
Entrez Gene ID
27173
Gene Name
solute carrier family 39 member 1
Gene Aliases
ZIP1, ZIRTL
UniGene
Hs.743291
Gene Chromosome Location
Chr.1: 153959099-153968184 on Build GRCh38
Transcripts
NM_001255978.1 NM_001255979.1 NM_001255980.1 NM_001255981.1 NM_130898.3 NR_045658.1 XM_006711172.2 XM_017000372.1 NM_001271960.1

SNP Detail

SNP ID Chromosome Location
rs540942118 Chr.1: 153967917-153967917 on Build GRCh38
rs566819861 Chr.1: 153967961-153967961 on Build GRCh38
rs564387750 Chr.1: 153968071-153968071 on Build GRCh38
rs760152152 Chr.1: 153968061-153968061 on Build GRCh38
rs527562890 Chr.1: 153967975-153967975 on Build GRCh38
rs562543656 Chr.1: 153967780-153967780 on Build GRCh38
rs1026500933 Chr.1: 153967943-153967943 on Build GRCh38
rs938177713 Chr.1: 153967892-153967892 on Build GRCh38
rs1014019554 Chr.1: 153967845-153967845 on Build GRCh38
rs950919562 Chr.1: 153967948-153967948 on Build GRCh38
rs537977186 Chr.1: 153968103-153968103 on Build GRCh38
rs189773134 Chr.1: 153968059-153968059 on Build GRCh38
rs987580269 Chr.1: 153967952-153967952 on Build GRCh38
rs529873751 Chr.1: 153967871-153967871 on Build GRCh38
rs533100793 Chr.1: 153968086-153968086 on Build GRCh38
rs1045568159 Chr.1: 153968219-153968219 on Build GRCh38
rs941293308 Chr.1: 153968011-153968011 on Build GRCh38
rs949884732 Chr.1: 153968217-153968217 on Build GRCh38
rs969754655 Chr.1: 153967863-153967863 on Build GRCh38
rs935923186 Chr.1: 153968148-153968148 on Build GRCh38
rs550193808 Chr.1: 153968113-153968113 on Build GRCh38
rs535694199 Chr.1: 153968218-153968218 on Build GRCh38
rs1001193282 Chr.1: 153967887-153967887 on Build GRCh38
rs973604986 Chr.1: 153967939-153967939 on Build GRCh38
rs905652788 Chr.1: 153968221-153968221 on Build GRCh38
rs386635613 Chr.1: 153967881-153967882 on Build GRCh38
rs916979521 Chr.1: 153967916-153967916 on Build GRCh38
rs745472078 Chr.1: 153968105-153968105 on Build GRCh38
rs113218877 Chr.1: 153968034-153968034 on Build GRCh38
rs1046690846 Chr.1: 153967963-153967963 on Build GRCh38
rs1024442342 Chr.1: 153967846-153967846 on Build GRCh38
rs568328368 Chr.1: 153968180-153968180 on Build GRCh38
rs1038291977 Chr.1: 153968037-153968037 on Build GRCh38
rs780979491 Chr.1: 153967984-153967984 on Build GRCh38
rs891637087 Chr.1: 153968183-153968183 on Build GRCh38
rs552324449 Chr.1: 153968033-153968033 on Build GRCh38
rs926825684 Chr.1: 153967882-153967882 on Build GRCh38
rs184126855 Chr.1: 153967814-153967814 on Build GRCh38
rs548395358 Chr.1: 153967894-153967894 on Build GRCh38
rs925926113 Chr.1: 153968119-153968119 on Build GRCh38
rs573520504 Chr.1: 153968111-153968111 on Build GRCh38
rs751690831 Chr.1: 153967867-153967867 on Build GRCh38
rs968287048 Chr.1: 153967783-153967783 on Build GRCh38
rs867540286 Chr.1: 153967805-153967805 on Build GRCh38
rs535231181 Chr.1: 153967923-153967923 on Build GRCh38
rs998640762 Chr.1: 153967781-153967781 on Build GRCh38

View Full Product Details